Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 50
Type J Thermocouple Temperature Controller
to
Specific Gravity Instrument
Image
Product Detail
More Detail
Type J Thermocouple Temperature Controller
Buy Type J Thermocouple Temperature Controller Items Including:
Oakton Temp-9000 Thermocouple Temperature Controller, 115V
Oakton Temp-9000 Thermocouple Temperature Controller, 230V
Autotuning Benchtop Temperature Controllers; type J; 100-240V
Digi-Sense® Standard Temperature Controller, 110V (CPTEMCTPSTD01).
More Details
Inclined Manometer Instrument
Buy Inclined Manometer Instrument Items Including:
200.5: Inclin Mnmtr.1/0/1inW Durablock Stat Inclined Manometer 10/0/1i
209:Incline Mnmtr.2/0/3 Inw Durablock Stat Inclined Manometer .2/0/3.0
202.5:Inclin Mnmtr.2/0/2inW Durablock Stat Inclined Mnometer 0.2/0/2.0
Red Gauge Oil 4 oz.
More Details
10 Ml Pipettes
Buy 10 Ml Pipettes Items Including:
Thermo Scientific Finntip 10 mL Pipette Tips, 2 to 10 mL, 100/bag
Disposable Tips, 2-10 mL, 200/bg (BMT-Z)
Thermo Scientific Finntip 10 mL Sterile Pipette Tips, 2 to 10 mL, 5x24
CORNING Stripette Serological Pipettes, 1.0 mL, Individually Wrapped,.
More Details
Meriam Instruments Manometer
Buy Meriam Instruments Manometer Items Including:
30EBX25TM : Table Mount Manometer, 30in
Well Type Manometer, 303 Stai
30EBX25TM : Table Mount Manometer, 40in
Well Type Manometer, 303 Stai
Manometer 0 To 28" H2o Differential
More Details
Druck Dpi 104 is Pressure Gauge
Buy Druck Dpi 104 is Pressure Gauge Items Including:
PV210104ISHP204G : IS Test Kit,10psig DPI 104 Instrinsically Safe Pneu
PV210104 ISHP207G : IS Test Kit, 30psig DPI 104 Instrinsically Safe Pn
PV211104 ISHP210G : IS Test Kt, 100psig DPI 104 Instrinsically Safe Pn.
More Details
Weighing Paper
Buy Weighing Paper Items Including:
Glassine Weighing Paper 3x3" 500/pk (YO-01338-00)
Glassine Weighing Paper 4x4" 500/pk (YO-01338-02)
Glassine Weighing Paper 6x6" 500/pk (YO-01338-04).
More Details
Digital Inline Air Flow Meter
Buy Digital Inline Air Flow Meter Items Including:
Hydronic Manometer (HM680) Water/Air Flow Meter
Fluke 922 Air Flow / Digital Manometer (FLUKE-922/KIT)
Precision Digital ProtEX-MAX PD8-6200-6H0 Dual-line, Analog input flow
Precision Digital ProtEX-MAX PD8-6200-6H5 Dual-line, Analog input flow.
More Details
Thermo Scientific Nalgene Vacuum Desiccator
Buy Thermo Scientific Nalgene Vacuum Desiccator Items Including:
Thermo Scientific Nalgene Vacuum Desiccator, PC cover and body, 280 mm (5311-0250)
Thermo Scientific Nalgene vacuum desiccator PC cover, PP body 330 mm O (5310-0250)
Thermo Scientific Nalgene Autoclavable plastic desiccator; 140mm plate (5315-0150).
More Details
Portable Oxygen Meter with Glp
Buy Portable Oxygen Meter with Glp Items Including:
Thermo Scientific Orion Star A123 Dissolved Oxygen Portable Meter Kit
Thermo Scientific Orion Star A223 DO Portable Meter Kit
Thermo Scientific Orion Star A223 DO Portable Meter Only
Thermo Scientific Orion Star A323 RDO/DO Portable Meter Kit.
More Details
Brass T Compression Fittings 1 8
Buy Brass T Compression Fittings 1 8 Items Including:
Compression fitting; probe diameter, 3/16"; Brass; 1/8"NPT (M) (3-2 FH4BZ-B)
Ferrule and nut assembly, brass, 1/8" (YO-31212-69)
Tee union, 1/8", Brass, each (YO-31412-17)
Compression Union, Brass, 1/8", Each (YO-31412-51).
More Details
Fluke 789 Process Meter Manual
Buy Fluke 789 Process Meter Manual Items Including:
Fluke 789 Process Meter
Fluke 771 Process mA Clamp Meter (FLUKE-771)
Fluke 353 Clamp Meter, True-rms, 2000A (FLUKE-353)
Fluke 355 Clamp Meter, True-rms, 2000A (FLUKE-355)
Fluke 321 Clamp Meter (FLUKE-321)
More Details
5 Ml Volumetric Flask Tolerance
Buy 5 Ml Volumetric Flask Tolerance Items Including:
PYREX Brand 5640 Volumetric Flask; 100 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 200 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 1000 mL, pack of 1
PYREX Brand 5640 Volumetric Flask; 2000 mL, pack of 1.
More Details
Indoor Air Quality Meters
Buy Indoor Air Quality Meters Items Including:
Extech Indoor Air Quality Meter
Extech Desktop Indoor Air Quality CO2 Monitor with Memory
TSI™ IAQ-Calc™ CO2 meter
TSI™ IAQ-Calc™ datalogging CO and CO2 meter, with hum
GE (Telaire) Dual-Position CO2/Temperature Meter
More Details
Laboratory Pumps
Buy Laboratory Pumps Items Including:
Complete Laboratory Drum Pump Set, 27" in length, 18.5 GPM, Polypropyl
Complete Laboratory Drum Pumps, 27" in length, 17 GPM, Stainless Steel
Complete Laboratory Drum Pump Set, 27" in length, 17 GPM, Stainless St.
More Details
Stackable Acid Storage Cabinet
Buy Stackable Acid Storage Cabinet Items Including:
SciMatCo Full-Size Acid Storage Cabinet, twenty four 2.5 L bottle capa
SciMatCo Full-Size Acid Storage Cabinet with Full Lining, thirty 2.5 L
SciMatCo HDPE Benchtop Acid Storage Cabinet, six 2.5 L bottle capacity.
More Details
Lab Companion
Buy Lab Companion Items Including:
Lab Companion Hot Plate, Ceramic top 7"x7", 120V
Lab
Companion® Stirring Hot Plate, Ceramic Top, 240V (BDH571065K)
Lab
Companion Dual-Action Shaker, 14" x 14", 120V
More Details
Hose Barb X Female Npt Adapter
Buy Hose Barb X Female Npt Adapter Items Including:
Hose Barb Adapter, PTFE, 1/8" NPT(F) X 1/8" ID (F-FH0101)
Hose Barb Adapter, PTFE, 1/8" NPT(F) X 1/4" ID (F-FH0102)
Hose Barb Adapter, PTFE, 1/4" NPT(F) X 1/8" ID (F-FH0201).
More Details
Asco Pinch Valve
Buy Asco Pinch Valve Items Including:
Two-way normally closed pinch valve, 24 VDC, 1/16" x 1/8" (SCH284A003- 24VDC)
Two-way normally closed pinch valve, 24 VDC, 1/8" x 3/16" (SCH284A005- 24VDC)
3-way pinch valve, 12 VDC, 1/16" x 1/8" (SCH384A002- 12VDC).
More Details
Plate Centrifuge
Buy Plate Centrifuge Items Including:
Compact PCR Plate Centrifuge, 120VAC
PCR Support Plate for 17416-00 and -05, PCR strips, tubes and plates/2
Pony Utility Pump for Intermittent Use, Nickel Plated Brass, 110VAC
Pony Utility Pump for Intermittent Use, Nickel Plated Brass, 12VDC.
More Details
Cole Flex
Buy Cole Flex Items Including:
Cole-Parmer Polystat low-temperature immersion chiller; flex probe; 12.
More Details
Vial Storage Boxes
Buy Vial Storage Boxes Items Including:
Polypropylene cryogenic vial rack, holds 50 round bottom vials (985800)
Grid dividers holds 12-mm vials for Thermo Scientific Revco® Freez (5958)
Grid dividers holds 13-mm vials for Thermo Scientific Revco® Freez (6212).
More Details
3 Mm Diameter Stainless Steel Tube
Buy 3 Mm Diameter Stainless Steel Tube Items Including:
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 3/8" (YO-31528-29)
160-18 Stainless Steel Pitot Tube 18" Insertion Length 5/16" Diameter
160-48 Stainless Steel Pitot Tube 48" Insertion Length 5/16" Diameter.
More Details
Low Rpm Ac Motor
Buy Low Rpm Ac Motor Items Including:
56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,.
More Details
Gf Signet Ph Orp
Buy Gf Signet Ph Orp Items Including:
+GF+ Signet DryLoc Preamp with ISO threads, 15-ft cable (3-2760-2)
+GF+ Signet DryLoc Connector with ISO threads, 15-ft cable (3-2760-4)
pH electronic, Digital
Wall Mount Kit
Submersible Sensor electronics with ISO threads, 15-ft cable (3-2750-4).
More Details
Orion Ph Meter 230a Manuals
Buy Orion Ph Meter 230a Manuals Items Including:
Thermo Scientific Orion 3-star benchtop pH meter, 115/220 VAC
Thermo Scientific Orion 3-star benchtop pH meter kit, 115/220 VAC
Thermo Scientific Orion 2-star benchtop pH meter, convenience kit 115/
Thermo Scientific Orion 3-Star handheld pH meter.
More Details
Large Clear Plastic Cylinder Containers
Buy Large Clear Plastic Cylinder Containers Items Including:
Rubbermaid Storage Container with Handles, Clear PP, 2 quart
Rubbermaid Storage Container with Handles, Clear PP, 4 quart
Rubbermaid Storage Container with Handles, Clear PP, 12 quart
Rubbermaid Storage Container with Handles, Clear PP, 18 quart.
More Details
High School Chemistry Lab Equipment
Buy High School Chemistry Lab Equipment Items Including:
High-intensity, long-wave UV lamp; 20-ft lead, 115 VAC/60 Hz (YO-97600-10)
High-intensity, long-wave UV lamp; 20-ft lead, 220 VAC/50 Hz (YO-97600-15)
Size 1 high-flow polypropylene filter bag; 200°F (93°C) maximu (255042-03).
More Details
Men's Extra Large Rubber Gloves
Buy Men's Extra Large Rubber Gloves Items Including:
Cryo-Industrial gloves, wrist style, extra large, 12" length
Cryo-Industrial gloves, mid-arm style, extra large, 15" length
Best Viton Gloves, extra-large, 12-mil thickness, 1 pair
Best Viton Gloves, extra-large, 28-mil thickness, 1 pair.
More Details
High Pressure Flexible Tubing
Buy High Pressure Flexible Tubing Items Including:
High-pressure valve, PTFE, 4 port, 90° adjacent ports connection, (YO-06471-76)
Masterflex High-Pressure PharMed BPT Tubing, L/S 14HP, 25 ft/pack (YO-95664-14)
Masterflex High-Pressure PharMed BPT Tubing, L/S 15HP, 25' (YO-95664-15).
More Details
Specific Gravity Instrument
Buy Specific Gravity Instrument Items Including:
H-B Instrument Hydrometer, 1.000 to 1.225 Specific Gravity, 0.005 Divi
H-B Instrument Hydrometer, 1.200 to 1.450 Specific Gravity, 0.005 Divi
H-B Instrument Hydrometer, 0.700 to 0.770 Specific Gravity, 0.001 Divi.
More Details
70618
results
<<<Previous
46
47
48
49
50
51
52
53
54
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
55
56
57
58
59
60
61
62
63
64
70
80
90
100
110
120
130
140
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Type J Thermocouple Temperature Controller to Specific Gravity Instrument
>
31125329-s-toc_2 31125329stoc2