Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 500
Pressure Clamps
to
Iotech Personal Daq 3000 <
Image
Product Detail
More Detail
Pressure Clamps
Buy Pressure Clamps Items Including:
Digital Sanitary Gauge, 1.5" Tri-Clamp, 30 to 0" Hg, battery powered
Digital Sanitary Gauge, 1.5" Tri-Clamp, 0 to 100 psi, battery powered
Digital Sanitary Gauge, 1.5" Tri-Clamp, 30 to 0" Hg, loop powered.
More Details
Electronic Tools
Buy Electronic Tools Items Including:
Fowler PolyCal Electronic Caliper (54101175)
Electronic depth micrometer (54-225-456)
Electronic depth gauge (54-125-777)
Fowler Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) (54422444)
TSI/Alnor EBT731 Electronic Balancing Capture Hood.
More Details
2 Stainless Steel Pipe Adapter
Buy 2 Stainless Steel Pipe Adapter Items Including:
Compression pipe adapter elbow, 1/2" x 1/2", Stainless Steel, Each (YO-30486-21)
Compression pipe adapter elbow, 1/16" x 1/2", Stainless Steel, Each (YO-30486-60).
More Details
Tds Probes
Buy Tds Probes Items Including:
Oakton TDS 6+ handheld TDS meter with probe
WTW Cond 3110 meter kit and TetraCon probe, 1-m cable
WTW Cond 3110 meter kit and TetraCon probe, 3-m cable
WTW Cond 3110 meter kit and KLE probe
WTW Cond 3210 meter kit and TetraCon probe, 1-m cable.
More Details
Two-Way Pvc Ball Valves
Buy Two-Way Pvc Ball Valves Items Including:
Low-Cost PVC Two-Way Ball Valve, 1" NPT(F) connectors (TJH-003)
Ball valve, 2-way, 1/8" NPT(F) x 1/8" NPT(M), PVC with Buna N seals (PVC 226-2F2M-B)
Ball valve, 2-way right angled, 1/4" NPT(F) x 1/4" NPT(M) (PVC-657-4M4F-B).
More Details
Graphical Neoprene Full Face Masks
Buy Graphical Neoprene Full Face Masks Items Including:
North Full-Face Respirator, Silicone
3M 6000-series Full-face respirators, Medium
Cartridge for North Full-Face and Half-Mask Respirators, Defender™ (YO-81606-50)
Cartridge for North Full-Face and Half-Mask Respirators, Organic vapor (YO-81606-56).
More Details
Cole-Parmer Conical Graduates, Pp
Buy Cole-Parmer Conical Graduates, Pp Items Including:
Polypropylene conical graduate, 100 mL (212205)
Polypropylene conical graduate, 250 mL (212215)
Polypropylene conical graduate, 500 mL (212225).
More Details
Recirculating Refrigerated Bath
Buy Recirculating Refrigerated Bath Items Including:
Air-cooled solid-state recirculating chiller, 8-oz reservoir, 600-watt (YO-12751-00)
PolyScience Recirculator; -10 to 70°C; 1 hp; centrifugal pump with (YO-12910-21)
PolyScience Recirculator; -10 to 70°C; 1 hp; centrifugal pump with (YO-12910-22).
More Details
Mesh Strainers
Buy Mesh Strainers Items Including:
Stainless Steel Strainer, 304 SS (heavy duty), Single Mesh, 10-1/4" Bo
Cole-Parmer Low-Cost In-Line Strainer Systems, 304 ss Strainer Mesh fo (YO-29595-55)
Cole-Parmer Low-Cost In-Line Strainer Systems, 304 ss Strainer mesh fo (YO-29595-57).
More Details
Variable Speed Mixer
Buy Variable Speed Mixer Items Including:
Heavy-Duty, Variable Speed Mixer, 120 VAC (YO-04340-10)
Talboys low-cost laboratory mixer; light-duty, variable speed, single (YO-04335-00)
Waring variable-speed blender, 1-liter, SS, 120v
Variable-Speed Homogenizer; 18,000 rpm; 115V (YO-04728-00).
More Details
Noshok Pressure Transmitters and Pressure Transducers
Buy Noshok Pressure Transmitters and Pressure Transducers Items Including:
Noshok Series 100 Pressure Transmitter, 1/4" NPT(M), 1000 psi
Noshok Series 100 Pressure Transmitter, 1/4" NPT(M), 2000 psi
Noshok Series 100 Pressure Transmitter, 1/4" NPT(M), 3000 psi.
More Details
Watts Stainless Steel Manifold Box
Buy Watts Stainless Steel Manifold Box Items Including:
304 Stainless Steel manifold; 2 outlets; 1/4" inlet NPT(F) (YO-31521-31)
304 Stainless Steel manifold; 2 outlets; 3/8" inlet NPT(F) (YO-31521-33)
304 Stainless Steel manifold; 2 outlets; 1/2" inlet NPT(F) (YO-31521-35).
More Details
Cryogenic Accessories
Buy Cryogenic Accessories Items Including:
Stainless steel cryogenic transfer hose, 4-ft long (8CF0403D21S4A-4FT)
Stainless steel cryogenic transfer hose, 6-ft long (8CF0403D21S4A-6FT)
Thermo Scientific Nalgene Stepped Cryocane, Aluminum, 290 mL (holds 5
CryoClaw, 22"length (AY509X18).
More Details
Centrifuge 12 Places
Buy Centrifuge 12 Places Items Including:
Fixed-angle Rotor; 12 place x 15mL
Thermo Scientific Medilite 12-place centrifuge, 120 VAC, 60 Hz
Thermo Scientific Medilite 12-place centrifuge, 220/240 VAC, 50/60 Hz
Rotor, 6-place for 12-mL centrifuge tubes - for use with 17414-series (C880-01).
More Details
Cole-Parmer Plastic Fittings
Buy Cole-Parmer Plastic Fittings Items Including:
Plastic hose clamps, black, 0.569" x 0.650", 100/pack (YO-95613-08)
Plastic hose clamps, black, 0.859" x 0.989", 100/pack (YO-95613-13)
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61).
More Details
Cole-Parmer Collapsible Containers, Ldpe
Buy Cole-Parmer Collapsible Containers, Ldpe Items Including:
Collapsible Container, Low-Density Polyethylene (LDPE), 1 quart
Collapsible Container, Low-Density Polyethylene (LDPE), 1 gallon
Collapsible Container, Low-Density Polyethylene (LDPE), 2.5 gallon.
More Details
Circular Recording Charts
Buy Circular Recording Charts Items Including:
Chart Paper for 8" Circular Recorders; 7 day, 0 to 200, 100 sheets/pk
Chart Paper for 8" Circular Recorders; 24 hour, 0 to 200°, 60 shee
Chart Paper for 8" Circular Recorders; 24 hour, 0 to 2000°F, 60/pk.
More Details
3 Way Pneumatic Valve
Buy 3 Way Pneumatic Valve Items Including:
Hayward pneumatically actuated 3-way ball valve; 1" PVC
Pneumatically Actuated 2-Way Elliptical Valve System; PP body, 3/8" NP (32-PP-RA4)
Two-Way Elliptical Valve, PP, 115 VAC, 3/8"
Two-Way Elliptical Valve, PP, 115 VAC, 3/4".
More Details
4 Stainless Quick Disconnect
Buy 4 Stainless Quick Disconnect Items Including:
Quick-disconnect insert, stainless steel, 1" sanitary connector (SQCX222416M)
Quick-Disconnect NPT(M) Pipe Adapter; Valved, 1/4 in
NPT(M), 1/4 in
(PLCD10004)
Quick-Disconnect NPT(M) Pipe Adapter; Valved, 1/4 in
More Details
Shore D
Buy Shore D Items Including:
Shore A digital hardness tester
Phase II PHT-980 Durometer, Shore D scale
Phase II PHT-960 Durometer, Shore A scale
Portable Shore Durometer Mdl:53-762-002
Portable Shore Durometer Model 53-762-001
More Details
Hach Water Analysis Accessories
Buy Hach Water Analysis Accessories Items Including:
Replacement DO Probe for Hach Oxygen Meters, 3-ft Cable
DO Probe for Hach Dissoved Oxygen Meters, 10-ft Cable
BOD Accessory Kit for Hach Dissolved Oxygen Meters
Replacement DO Cap for Hach DO Probes, 2/pack
DO Probe Service Kit for Hach DO Probes.
More Details
Infrared Thermometer Review Industrial
Buy Infrared Thermometer Review Industrial Items Including:
80PK-27 SureGrip Industrial Surface Temperature Probe (YO-20007-79)
Mi3 Sensor, Standard Model, 10:1, -40-600°C, 3m Cable
Mi3 Sensor, Standard Model, 10:1, -40-600°C, 8m Cable
Mi3 Sensor, Standard Model, 10:1, -40-600°C, 1m Cable
Air purge jacket.
More Details
250 Ml Polypropylene Centrifuge Bottle
Buy 250 Ml Polypropylene Centrifuge Bottle Items Including:
Thermo Scientific Nalgene Centrifuge Bottle; 250-mL; PC; 61.5 mm dia x
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; FEP; 1/pack
Azlon solvent venting wash bottle for acetone, 250 mL
Azlon solvent venting wash bottle for isopropanol, 250 mL.
More Details
Air Line Tubing Quick Disconnect
Buy Air Line Tubing Quick Disconnect Items Including:
Metal Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 5/16" Tube (LCD16005)
Quick-Disconnect Hose Barb Fitting Body; Valved, 1/16 in
Tube ID, 1/8 (PMCD1701)
Quick-Disconnect Hose Barb Fitting Body; Valved, 1/8 in
Tube ID, 1/8 (PMCD1702).
More Details
Adam Equipment Adam Nimbus Analytical Balances
Buy Adam Equipment Adam Nimbus Analytical Balances Items Including:
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-98)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-99).
More Details
Temperature Clamps
Buy Temperature Clamps Items Including:
Sanitary Ball Valve, 3/4" TRI-CLAMP
Sanitary Ball Valve, 1" TRI-CLAMP
Sanitary Ball Valve, 2" TRI-CLAMP
Sanitary Butterfly Valve, 1" TRI-CLAMP
Temperature Meter with Pipe Clamp for diameters from 1/4 to 3/8" and r (4005I).
More Details
Cole-Parmer Bag Filter Systems
Buy Cole-Parmer Bag Filter Systems Items Including:
Polypropylene bag filter housing, 1-1/2" NPT(F) (150338)
Polypropylene bag filter housing, 1-1/2" NPT(F) (150337)
Polypropylene felt filter bag, 10"L, 10 µm (155387-03)
Polypropylene felt filter bag, 10"L, 25 µm (155389-03).
More Details
Labpette Fluid & Liquid Handling
Buy Labpette Fluid & Liquid Handling Items Including:
Labpette Discovery Pipette 0.5 - 10.0 uL (P3940-10)
Labpette Discovery Pipette 10 - 100 uL (P3940-100)
Labpette Discovery Pipette 20 - 200 uL (P3940-200)
Labpette Discovery Pipette 100- 1000 uL (P3940-1000)
Labpette Discovery Pipette 0.1 - 2.0 uL.
More Details
Test Scale
Buy Test Scale Items Including:
Atago Digital Pocket Ethylene Glycol Refractometer (°C scale)
Atago Digital Pocket Propylene Glycol Refractometer (°C scale)
Atago Digital Pocket Propylene Glycol Refractometer (°F scale)
Atago Special-scale Refractometer, Salinity of Seawater.
More Details
Iotech Personal Daq 3000 <
Buy Iotech Personal Daq 3000 < Items Including:
IOtech Personal Daq 3000™ USB, 1-MHz, 16-bit Multifunction Data Acq
IOtech Personal Daq 3005™ USB, 1-MHz, 16-bit Multifunction Data Acq
IOtech Personal Daq 3001™ USB, 1-MHz, 16-bit Multifunction Data Ac.
More Details
70618
results
<<<Previous
496
497
498
499
500
501
502
503
504
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
410
420
430
440
450
460
470
480
485
486
487
488
489
490
491
492
493
494
495
496
497
498
499
500
501
502
503
504
505
506
507
508
509
510
511
512
513
514
520
530
540
550
560
570
580
590
600
700
800
900
1,000
1,100
1,200
1,300
1,400
2,000
2,354
Pressure Clamps to Iotech Personal Daq 3000 <
>
31125329-s-toc_2 31125329stoc2