Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 6
Peristaltic Pump 7520 00
to
High Temperature Laboratory Oven Thermometer
Image
Product Detail
More Detail
Peristaltic Pump 7520 00
Buy Peristaltic Pump 7520 00 Items Including:
Low Flow Peristaltic Pump, Variable Speed, 1.5 - 5 RPM, 12 VDC, Manual
Cole-Parmer One Channel precision micro peristaltic pump
Pump, Peristaltic Metering, Fixed Speed, 2.3 GPD, 115 VAC
Pump, Peristaltic Metering, Variable Speed, 2.3 GPD, 115 VAC.
More Details
Dispensette Iii Bottle Top Dispenser
Buy Dispensette Iii Bottle Top Dispenser Items Including:
Adjustable-volume digital dispensers (4701321)
Adjustable-volume digital dispensers (4701331)
Adjustable-volume digital dispensers (4701341)
Adjustable-volume digital dispensers (4701351)
Adjustable-Volume analog dispenser - 0.05-.05mL (4701101).
More Details
Taylor Wharton Dewar
Buy Taylor Wharton Dewar Items Including:
Taylor Wharton Storage Dewars Tipping Stand (D025-8C00)
Dewars with Canister Storage Systems, 20.7 Liters, 2.0" Neck ID (20XTB-11M)
Dewars with Canister Storage Systems, 34 Liters, 2.0" Neck ID (34XTB-11M)
Storage Dewars, 4 Liters (4LDB).
More Details
Peristaltic Pump Tubing Sizes
Buy Peristaltic Pump Tubing Sizes Items Including:
Tube Assembly Kit for the peristaltic pump 74130-60 and 74130-61 pumps
Masterflex two-stop pump tubing, Tygon, L/S 15
Pack of 4.
Masterflex two-stop pump tubing, Tygon, L/S 24
Pack of 4.
More Details
Orion Quikchek Conductivity Meter Manual
Buy Orion Quikchek Conductivity Meter Manual Items Including:
Thermo Scientific Orion® 3-Star Handheld Conductivity Meter Kit
Thermo Scientific Orion 3-Star Benchtop Conductivity Meter Only
Thermo Scientific Orion 3-Star Benchtop Conductivity Meter Kit
Thermo Scientific Orion 4-Star pH/Conductivity Meter Kit.
More Details
Micro Flow Meter 10 Ml Minute
Buy Micro Flow Meter 10 Ml Minute Items Including:
Piston Metering Pump, 1/8" stainless head, 0.01 to 10 mL/min range, 60 (2SMP)
High Pressure Piston Pump, Stainless Steel construction, 0.01 to 10 mL
Economical High-Pressure Dual-Piston Pump, 10.0 mL/min Max Flow, 230 V (AA-100-S-2-CE (#1444)).
More Details
Koflo Static Mixer
Buy Koflo Static Mixer Items Including:
PVC static mixer without injection port; 1-1/2" dia (YO-04668-63)
Pipe Mixer, 316 Stainless Steel, 3/8 inch NPT(M), 12 elements (YO-04669-11)
Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 6 elements (YO-04669-12).
More Details
Fluke Rtd Thermometer
Buy Fluke Rtd Thermometer Items Including:
HART 1552A-12 Handheld Thermometer, Fixed RTD, –80 To 300C, 1/4x12
Contact RTD probe for 35629-20 & -30
Temp range -40 to 500°F; 40" (YO-35629-50)
Fluke Calibration (formerly Hart Scientific) 1502A: Tweener Thermistor (1502A (115 VOLT)).
More Details
Brookfield Digital Viscometer Model Lvdve
Buy Brookfield Digital Viscometer Model Lvdve Items Including:
Replacement stand for viscometers (MODEL A)
Small sample adaptor for models 98945-00 and -05; 3 to 10 K cp
Small sample adapter for models 98945-10, -15, -20 and -25; 250 to 830
Replacement spindle for models 98945-00, -05, 50 to 100,000 cp (LV-2).
More Details
Benchtop Temperature Controller
Buy Benchtop Temperature Controller Items Including:
Autotuning Benchtop Temperature Controllers; type J; 100-240V
Oakton Temp-9500 Multiparameter Temperature Controller with USB Output (Temp-9500) (YO-89800-03)
Digi-Sense 1-Zone benchtop dual display temperature control, type J in (YO-36225-61).
More Details
Flow Meter 0 1000 Cc Min
Buy Flow Meter 0 1000 Cc Min Items Including:
Thermal Gas Mass Flow Controller, to 1000 L/min Air / N2, 3
Thermal Non-Invasive Low-Flow Flowmeters, 200 to 1000 µL/min (YO-32611-08)
Thermal Gas Mass Flowmeter, to 1000 L/min Ar, Aluminum body w/ display
Flowmeter 100 - 1000 mL/min (YO-32550-03).
More Details
Phase Ii Hardness Testers
Buy Phase Ii Hardness Testers Items Including:
Phase II PHT-3000 Multi-Function Digital PDA Hardness Tester w/ IR Pri
Phase II Ultrasonic Hardness Tester, with 15N handheld probe
Phase-II MET-U1A50 Ultrasonic Hardness Tester, with 50N handheld probe
Phase II Economical surface roughness tester.
More Details
100 Ml Graduated Cylinder
Buy 100 Ml Graduated Cylinder Items Including:
Cylinder, Pyrex Brand 3025, graduated, 100 mL
Cylinder, Pyrex Brand 3025, graduated, 100 mL, case of 12
Pyrex Brand 3024 cylinder; 100 mL, case of 12
Pyrex Brand 3042 cylinder; 100 mL, case of 24
Pyrexplus Brand 63024 cylinder; 100 mL, pack of 1.
More Details
Cole-Parmer International
Buy Cole-Parmer International Items Including:
Cole-Parmer 4-place Electrophoresis System without internal cooling
Cole-Parmer LabGEN Homogenizer stand for 04727-series
Data View Datalogger, Temperature/Humidity Datalogger with Graphic Dis (DVTH)
Mini Temperature/Humidity Dataloggers (SL300TH).
More Details
Horiba Conductivity Meter B 173 Manu
Buy Horiba Conductivity Meter B 173 Manu Items Including:
Replacement conductivity cell for Horiba conductivity and multiparamet
Horiba DS-72 conductivity benchtop meter
Horiba F-74 pH/Conductivity benchtop meter
Horiba F-72 pH/ORP/ION benchtop meter
Horiba B-171 Twin Conductivity/Salinity Pocket Tester.
More Details
Flow Meter 10 Ml Min
Buy Flow Meter 10 Ml Min Items Including:
Piston Metering Pump, 1/8" stainless head, 0.01 to 10 mL/min range, 60 (2SM)
Digital mass Flowmeter, Aluminum, For Air, 0 To 10 mL/min (XFM17A-VADL9A5-01-AIR/N2)
Digital mass Flowmeter, Aluminum, For Co2, 0 To 10 mL/min (XFM17A-VADL9A5-01-CO2).
More Details
Accumet Ph Meter
Buy Accumet Ph Meter Items Including:
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Accumet pH Electrode, Combination, General Purpose, Long-neck, Gel-fil
Accumet Benchtop Meter Replacement pH/ATC Probe
Replacement Self-Stirring BOD Probe for accumet Meters.
More Details
Inline Stainless Liquid Filter Process
Buy Inline Stainless Liquid Filter Process Items Including:
General-purpose 304 stainless steel filter bag housing, 2" NPT(F) (156121)
Filter Housing; 50 GPM (189 LPM); Stainless Steel
316 stainless steel adapter head; 3/4" NPT(F) (156094)
316 stainless steel adapter head; 1" NPT(F) (156095).
More Details
04644 Series Digital Hot Plate Stirrer
Buy 04644 Series Digital Hot Plate Stirrer Items Including:
IKA Digital Stirring Hot Plate, 115v (3581201)
IKA Ceramic Stirring Hot Plate, 10" x 10", 115v (3581401)
IKA Ceramic Stirring Hot Plate, 10" x 10", 230v (3581400)
EchoTherm Programmable Digital Stirring Hot Plate, 230V (HS40-2).
More Details
Positive Displacement Pipette Ptfe
Buy Positive Displacement Pipette Ptfe Items Including:
BRAND Positive Displacement Pipettors, 100-500 µL (2702904)
BRAND Positive Displacement Pipettors, 2-10 mL (2702912)
Brand Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/pk
Brand Positive Displacement Pipettor Tips, PP, 200-1000 µL, 10/pk.
More Details
Air Cadet Vacuum Pump 07350
Buy Air Cadet Vacuum Pump 07350 Items Including:
Air Cadet® Vacuum/Pressure Pump, Diaphragm, single head, 0.8 cfm,
Air Cadet® Vacuum/Pressure Pump, Diaphragm, dual head, 0.9 cfm, 11
Air Cadet® Vacuum/Pressure Pump, Diaphragm, 0.5 cfm, 115 VAC
Nitrile diaphragm service kit for all Air Cadet pumps.
More Details
Liquid Flow Rate Meter
Buy Liquid Flow Rate Meter Items Including:
Flow Rate Indicator with miniature 3-1/2 Digit LCD Display
Flow Sensor,50-500ml/min (105-5)
Flow Sensor,0.1-2l/min (102-7T)
Flow Sensor,100-1000ml/min (101-6T)
Flow Sensor,0.1-2l/min (105-7)
More Details
High Precision Digital Pressure Gauge
Buy High Precision Digital Pressure Gauge Items Including:
Ashcroft Digital Pressure Gauge, 0 to 1000 psi, 1/4" NPT(M) Process co (D1005PS2)
High-accuracy digital test gauge, 0 to 30" Hg range
High-accuracy digital test gauge, 0 to 15 psi range
High-accuracy digital test gauge, 0 to 30 psi range.
More Details
Extech 407510 Dissolved Oxygen Meter
Buy Extech 407510 Dissolved Oxygen Meter Items Including:
Extech DO600 Dissolved Oxygen Pocket Tester Kit
YSI splashproof dissolved oxygen meter with probe, 25-ft cable, model
YSI Model 550A Dissolved Oxygen Meter, 12-ft (3.7-m) probe cables
YSI DO 200 Dissolved Oxygen Meter Kit, 13-ft (4-m) probe cables.
More Details
Gf Signet Ph Meters
Buy Gf Signet Ph Meters Items Including:
+GF+ Signet Conductivity/Resistivity Cell, 316SS, K = 0.1, 1.5" sanita (3-2820-S1C)
+GF+ Signet Conductivity/Resistivity Cell, 316SS, K = 0.1, 2" sanitary (3-2820-S2C)
+GF+ Signet Conductivity Cell with Integral Electronics, K = 0.1, 4 to.
More Details
Large Lab Hot Plate
Buy Large Lab Hot Plate Items Including:
Lab Companion Hot Plate, Ceramic top 7"x7", 120V
Lab
Companion® Stirring Hot Plate, Ceramic Top, 240V (BDH571065K)
Lab
Companion® Stirring Hot Plate, Ceramic top, 120V (BDH571066U)
More Details
Portable Flammable Gas Leak Detector
Buy Portable Flammable Gas Leak Detector Items Including:
Leak hunter plus™ gas leak detector, 220 VAC
UEI™ Combustible Gas Leak Detector (CD100A)
Thermo Scientific GLD Pro Leak Detector
Portable Hydrocarbon (non-methane) Detector, 0 to 100% LEL
Portable Hydrocarbon (non-methane) Detector, 0 to 1000 ppm.
More Details
Electronic Desiccator Cabinet
Buy Electronic Desiccator Cabinet Items Including:
Electronic Desiccator with Flexboard Shelves; interior dim., 11-1/2"W (134041-2)
Electronic Desiccator with Stainless Steel Shelves; interior dim.,11-1 (134241)
Electronic Desiccator with Stainless Steel Shelves; interior dim., 11- (134241-2).
More Details
Vacuum Desiccator Cabinet
Buy Vacuum Desiccator Cabinet Items Including:
Scienceware Lab Companion Vacuum Desiccator Cabinet, 11L.
Scienceware Lab Companion Vacuum Desiccator Cabinet, 23L.
Scienceware Gas Purge Extra-Large Desiccator Cabinet, 3.6 cu
ft.
Scienceware Automatic Extra-Large Desiccator Cabinet, 3.6 cu
More Details
High Temperature Laboratory Oven Thermometer
Buy High Temperature Laboratory Oven Thermometer Items Including:
Electronic-controlled high-temperature vacuum oven, 0.67 cu ft, 120 VA (YO-05017-00)
Microprocessor-controlled high-temperature vacuum oven, 1.50 cu ft, 12 (YO-05017-10)
Durac Oven Thermometer, 20/130°C, 135mm Length, Total Immersion, P.
More Details
70618
results
<<<Previous
2
3
4
5
6
7
8
9
10
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Peristaltic Pump 7520 00 to High Temperature Laboratory Oven Thermometer
>
31125329-s-toc_2 31125329stoc2