Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 700

Lab Bead Mill   to   First Aid and Safety

Image
  Product Detail
More Detail

Lab Bead Mill

Buy Lab Bead Mill Items Including:
  • Mini bead beater, 115 VAC 60 Hz (YO-36270-02)
  • Mini bead beater, 230 VAC 50 Hz (YO-36270-07)
  • Glass Beads, 0.1 to 0.15 mm (YO-36270-52)
  • Zirconia-Silicate Beads, 0.5 mm (YO-36270-61)
  • 100 Sterile Vials with caps in foam rack, 2-mL (YO-36270-70).

Accuform Signs Biohazard Sign

Buy Accuform Signs Biohazard Sign Items Including:
  • Safety Sign, Biohazard, 7 X 10, Adhesive Vinyl
  • Safety Sign, Biohazard, 7 X 10, Aluminum
  • Safety Sign, Biohazard, 10 X 14, Adhesive Vinyl
  • Safety Sign, Biohazard, 10 X 14, Aluminum
  • Safety Sign, Warning - Biohazard, 7" X 10", Adhesive Vinyl.

Advantec Filter Membranes

Buy Advantec Filter Membranes Items Including:
  • Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.6 (YO-06644-06)
  • Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.4 (YO-06644-08).

Cole-Parmer Toroidal Conductivity Transmitters

Buy Cole-Parmer Toroidal Conductivity Transmitters Items Including:
  • Cole-Parmer Toroidal Conductivity transmitter, CPVC, 0 to 100 mS
  • Cole-Parmer Toroidal Conductivity transmitter, CPVC, 0 to 1000 mS
  • Cole-Parmer Toroidal Conductivity transmitter, PVDF, 0 to 100 mS.

Brady Lockout/Tagout Danger Tags

Buy Brady Lockout/Tagout Danger Tags Items Including:
  • Brady Personal Valve and Electrical Lockout Kit.

Petroleum Flash Point

Buy Petroleum Flash Point Items Including:
  • Pensky-Martens closed-cup flash tester, 115 VAC/60 Hz
  • Pensky-Martens gas-heated closed-cup flash tester
  • Koehler tag gas heated closed-cup flash tester
  • Cleveland open-cup flash tester, 115 VAC/60 Hz
  • Koehler tag open-cup flash tester, 115 VAC/60 Hz.

Silicone (Platinum-Cured) Autoanalysis/Microbore Tubing

Buy Silicone (Platinum-Cured) Autoanalysis/Microbore Tubing Items Including:
  • Three-stop silicone (platinum cured) tubing, 0.64 mm ID, 6/pack (95603-22)
  • Three-stop silicone (platinum cured) tubing, 0.76 mm ID, 6/pack (95603-24)
  • Three-stop silicone (platinum cured) tubing, 0.89 mm ID, 6/pack (95603-26).

3m Air Mate Belt Mounted (Papr)

Buy 3m Air Mate Belt Mounted (Papr) Items Including:
  • 3M Air-Mate™ Respirator Hood, Pkg/3
  • 3M Air-Mate™ Respirator Hood, Extended Length Pkg/3
  • 3M Air-Mate™ Respirator Breathing Tube
  • 3M Air-Mate™ Powered Air Purifying Resprirator Blower Assy
  • 3M Air-Mate™ HEPA Respirator Filter.

M Micro Ph Electrode Meter

Buy M Micro Ph Electrode Meter Items Including:
  • Cole-Parmer self-cleaning pH electrode, PPS housing, 100 Ω Pt RT (YO-27003-23)
  • Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
  • Cole-Parmer 3-ft pH electrode, submersible, single-junction, 100 ohm R (YO-05993-85).

Griffin Beakers no 1003

Buy Griffin Beakers no 1003 Items Including:
  • Pyrex Brand 1003 Griffin Heavy-Duty Beaker, 150 mL, 12/pk (1003-150)
  • Pyrex Brand 1003 Griffin Heavy-Duty Beaker, 250 mL, 12/pk (1003-250)
  • Pyrex Brand 1003 Griffin Heavy-Duty Beaker, 400 mL, 12/pk (1003-400).

Cole-Parmer Ultra-Ware Hplc Economy Cap Systems

Buy Cole-Parmer Ultra-Ware Hplc Economy Cap Systems Items Including:
  • 1 L Reservoir System (YO-02017-45)
  • 2 L Reservoir System (YO-02017-50).

Cole-Parmer Flexible Tubing

Buy Cole-Parmer Flexible Tubing Items Including:
  • Cole-Parmer Non-DEHP Lab Tubing 1/16" x 1/8", 50' package
  • Cole-Parmer Non-DEHP Lab Tubing 3/32" x 5/32", 50' pack
  • Pvc Tubing, 3/16 X 5/16", 50-ft/pk (YO-96605-02)
  • Pvc Tubing, 5/16 X 7/16", 50-ft/pk (YO-96605-06).

Corning Cell Plate 96 Well

Buy Corning Cell Plate 96 Well Items Including:
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Clear-Bottom Plates; Black, Cell Culture Surface
  • Corning 96-Well Black Solid Plates, Nontreated
  • Plate,round,white,100/cs (YO-01728-60).

Thermo Scientific Nalgene Wide-Mouth Bottles, Fep

Buy Thermo Scientific Nalgene Wide-Mouth Bottles, Fep Items Including:
  • Thermo Scientific Nalgene Bottle, Wide Mouth, FEP 125mL, 1/pk
  • Thermo Scientific Nalgene Bottle, Wide Mouth, FEP 250mL, 1/pk
  • Thermo Scientific Nalgene Bottle, Wide Mouth, FEP 500mL, 1/pk.

Peristaltic Pump Fittings and Accessories

Buy Peristaltic Pump Fittings and Accessories Items Including:
  • Tris 3-channel peristaltic pump (YO-01470-54)
  • 90° Elbow Fittings, 63 ISO-K, Aluminum (C100-07-203)
  • 90° Elbow Fittings, 100 ISO-K, Aluminum (C100-09-203)
  • Aluminum Tee Fittings, 63 ISO-K (C100-07-207)
  • Tee Fittings, 100 ISO-K, Aluminum (C100-09-207).

Crincle Finish Rubber Coated Gloves

Buy Crincle Finish Rubber Coated Gloves Items Including:
  • Sperian® Atlas Fit™ Rubber Coated Cotton/Poly Glove, Medium
  • Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Sm (320-S).

1 Od White Epdm Tubing

Buy 1 Od White Epdm Tubing Items Including:
  • Mullite process tube, open both ends, 1" OD, 12" L
  • Mullite process tube, open both ends, 1" OD, 18" L
  • Mullite process tube, open both ends, 1" OD, 24" L
  • Mullite process tube, open both ends, 1" OD, 36" L.

Fixed Infrared Thermometer with Laser Targeting

Buy Fixed Infrared Thermometer with Laser Targeting Items Including:
  • Seeker Infrared Thermometer and Scanner with Star Burst Laser Target (
  • Cole-Parmer scope and laser sighting infrared thermometer; sighting ty (YO-39800-00)
  • Cole-Parmer scope and laser sighting infrared thermometer; sighting ty (YO-39800-02).

Positive Displacement Rotary Gear Pumps

Buy Positive Displacement Rotary Gear Pumps Items Including:
  • Moderate Flow Positive Displacement Bypass Diaphragm Pump, 1.4 GPM, 11
  • Moderate Flow Positive Displacement Bypass Diaphragm Pump, 1.8 GPM, 12
  • Moderate Flow Positive Displacement Flexible Impeller Pump, 12 VDC, 5..

Stainless Steel Mixing Process Mixers

Buy Stainless Steel Mixing Process Mixers Items Including:
  • Aerosol-Sealed Assembly with 303 stainless steel mixing chamber; sampl (YO-04729-02)
  • 304 stainless steel housing for 4 filters (156025-02)
  • 304 stainless steel housing for 8 filters (156026-02)
  • 304 stainless steel housing for 12 filters (156027-02).

Fend all Emergency Personal Eyewash Station

Buy Fend all Emergency Personal Eyewash Station Items Including:
  • Fend-all Primary Eyewash Station (32-000200-0000)
  • Scienceware Plum Double Sterile Saline Eyewash Station, two 1000 mL bo
  • Scienceware Plum Covered Double Sterile Saline DUO Eyewash Station, tw
  • Scienceware Plum Sterile Saline Eyewash Station, 500 mL.

Ph Meter Probes Platinum

Buy Ph Meter Probes Platinum Items Including:
  • Conductivity probe, 2-cell, K = 0.1, epoxy body/platinum electrode, 2-
  • Conductivity probe, 2-cell, K = 1.0, glass body/platinum electrode, 2-
  • Conductivity probe, 2-cell, K = 10.0, epoxy body/platinum electrode, 2.

Labconco Precise Controlled Atmosphere Glove Boxes

Buy Labconco Precise Controlled Atmosphere Glove Boxes Items Including:
  • Controlled Atmosphere Glove Box, 115V (5220100)
  • Controlled Atmosphere Glove Box, 230V (5220120)
  • Adjustable Height Mobile Base Stand, 33-40" High (5235500).

Inlet Liquid Filter Screens

Buy Inlet Liquid Filter Screens Items Including:
  • Disposable solvent inlet filter; 1/16" OD stem, for 1/16" ID tubing (A-302)
  • Disposable Solvent Inlet Filter; Includes Flangeless Fittings For 1/8 (A-302)
  • Replaceable solvent inlet filter assembly, 2 µ, 10 mL/min max
  • flow (A-551).

Tissue Culture Tubes

Buy Tissue Culture Tubes Items Including:
  • Culture tube closure, 13 mm, stainless steel (2005-00013)
  • Culture tube closure, 16 mm, stainless steel (2005-00016)
  • Culture tube closure, 18 mm, stainless steel (2005-00018)
  • Cole-Parmer Sterile Tissue Culture 12- Well Plate, 100/case.

Pneumatics Valves

Buy Pneumatics Valves Items Including:
  • Pneumatically Actuated 2-Way Elliptical Valve System; PP body, 3/8" NP (32-PP-RA4)
  • Hayward pneumatically actuated 3-way ball valve; 1" PVC
  • Vacuum solenoid valve switch (S070C-6CC0-32)
  • Two-Way Elliptical Valve, PP, 115 VAC, 1/8".

Blunt Needles

Buy Blunt Needles Items Including:
  • Removable-type needle, 90° blunt tip, 26s gauge, for syringes 100 (YO-25702-60)
  • Removable-type needle, 90° blunt tip, 22s gauge, for syringes 100 (YO-25702-61)
  • Removable-type needle, 90° blunt tip, 22s gauge, for syringes 250 (YO-25702-63).

Siemens Energy & Automation Vacuum Pumps

Buy Siemens Energy & Automation Vacuum Pumps Items Including:
  • Automated self-cleaning dry vacuum system, 115 VAC.

Filter Paper Tube Holder

Buy Filter Paper Tube Holder Items Including:
  • Holder filter paper f/Nitrate (3200053995)
  • PFA In-Line Filter Holder, 47-mm, 1/4" OD x 1/4" OD Tubing Connections.

First Aid and Safety

Buy First Aid and Safety Items Including:
  • Safety Sign, First Aid Station (symbol), 7" X 10", Aluminum
  • Safety Sign, First Aid Station (symbol), 10" X 14", Adhesive Vinyl
  • Safety Sign, Eye Wash Station, 7 X 10, Adhesive Vinyl
  • Safety Sign, Eye Wash Station, 7 X 10, Plastic.



Lab Bead Mill to First Aid and Safety

>