Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 8
Leybold Vacuum Pump Cold Traps
to
Stir-Pak Laboratory Mixers
Image
Product Detail
More Detail
Leybold Vacuum Pump Cold Traps
Buy Leybold Vacuum Pump Cold Traps Items Including:
Cold trap, glass, 40 mL, 29/42 joint size (7729-28)
Cold trap, glass, 70 mL, 40/50 joint size (7729-38)
Rotary Evaporator with Cold-trap glassware, vacuum regulator, vapor te (YO-28623-25)
Acid neutralization trap for vacuum pumps 79201-00 and -05.
More Details
Instech Peristaltic Pump Explosion Proof
Buy Instech Peristaltic Pump Explosion Proof Items Including:
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/2 Hp, 3600 RPM,.
More Details
Psia Pressure Transducers
Buy Psia Pressure Transducers Items Including:
AccuSense™ Pressure Transducer, 0-15 PSIA
AccuSense™ Pressure Transducer, 0-25 PSIA
AccuSense™ Pressure Transducer, 0-50 PSIA
Absolute pressure monitor, 0 to 20 psia
Absolute pressure monitor, 0 to 50 psia
More Details
Propeller Blades for Laboratory Mixers
Buy Propeller Blades for Laboratory Mixers Items Including:
Propeller stirrer, 4-bladed (YO-50703-25)
Propeller stirrer, 4-bladed (YO-50703-26)
304 SS four-blade propeller for shaft 04370-02, 75 mm propeller dia (YO-04370-11)
304 SS four-blade propeller for shaft 04370-02, 60 mm propeller dia (YO-04370-12).
More Details
Plastic Luer Lock Syringes
Buy Plastic Luer Lock Syringes Items Including:
Syringes with PTFE luer lock; syringe capacity; 50µL; needle gauge; (YO-07939-32)
Syringes with PTFE luer lock; syringe capacity; 100µL; needle gauge (YO-07939-33)
Syringes with PTFE luer lock; syringe capacity; 1 mL; needle gauge; 22 (YO-07939-80).
More Details
Scientific I Vortex Genie Shakers
Buy Scientific I Vortex Genie Shakers Items Including:
Multi Vortex-Genie Shaker 120V
Multi Vortex-Genie 240V
Vortex-Genie 2 Shaker, 120 VAC
Vortex-Genie 2 Shaker, 230 VAC
Vortex-Genie 1 Shaker, 120 VAC
More Details
Digital Vacuum Meter
Buy Digital Vacuum Meter Items Including:
Handheld Vacuum Meter, 0-1999 mbar
Vacuum Meter/Controller with Dogotal Dial and Reading, 810 to 0.1 Torr
Vacuum Meter/Controller with Digital Dial and Reading, 115 VAC (683,160.0)
Cole-Parmer Digital Gauge, 760 Torr; 1/4" NPT(M) w/ Backlight.
More Details
Wika Vacuum Gauge
Buy Wika Vacuum Gauge Items Including:
30 to 0" Hg Test Gauge, 4" Dial
0 to 400 psi Test Gauge, 4" Dial
0 to 600 psi Test Gauge, 4" Dial
30 to 0" Hg Test Gauge, 6" Dial
0 to 400 psi Test Gauge, 6" Dial
More Details
Handheld Portable Ph Meter
Buy Handheld Portable Ph Meter Items Including:
Oakton pH meter with probe
WTW pH 1970i Heavy-Duty pH Meter
Hach waterproof ISFET pH meter kit
Thermo Scientific Orion 3-Star handheld pH meter kit
Horiba D-51 Waterproof pH Meter
More Details
Flexible Stainless Steel Tubing for Liquids
Buy Flexible Stainless Steel Tubing for Liquids Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Accuracy of Cannon Fenske Routine Viscometer
Buy Accuracy of Cannon Fenske Routine Viscometer Items Including:
Cannon Cannon-Fenske routine viscometer, size 25
Cannon Cannon-Fenske routine viscometer, size 50
Cannon Cannon-Fenske routine viscometer, size 75
Cannon Cannon-Fenske routine viscometer, size 100
Cannon Cannon-Fenske routine viscometer, size 150.
More Details
Walchem High Stroke-Frequency Diaphragm Metering Pumps
Buy Walchem High Stroke-Frequency Diaphragm Metering Pumps Items Including:
Remote Control Pump with PVC head, 1.3 GPH, 230 VAC, 50 Hz
Remote Control Pump with GFRPP head, 2.3 GPH, 115 VAC, 50/60 Hz
Remote Control Pump with GFRPP head, 2.3 GPH, 230 VAC, 50 Hz
Remote Control Pump with GFRPP head, 4.3 GPH, 115 VAC, 50/60 Hz.
More Details
Circulating Refrigerated Water Bath
Buy Circulating Refrigerated Water Bath Items Including:
Air-cooled solid-state recirculating chiller, 8-oz reservoir, 600-watt (YO-12751-00)
Cole-Parmer Flow-through Refrgerated Chillers; range, -25 to 40°C; (YO-01283-40)
Cole-Parmer Flow-through Refrgerated Chillers; range, -25 to 40°C; (YO-01283-41).
More Details
High Speed Glass Centrifuge Tubes
Buy High Speed Glass Centrifuge Tubes Items Including:
Compact Variable-Speed Universal Centrifuge Tube Adapter for 15-mL Fix (YO-17306-60)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Concentrated Acid Bottle Top Dispenser
Buy Concentrated Acid Bottle Top Dispenser Items Including:
Cole-Parmer Bottletop Dispenser .25-2.5 mL
Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Glass Bottle.
More Details
Bipolar Dc Power Supply
Buy Bipolar Dc Power Supply Items Including:
Quad Output DC Power Supply (382270)
High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 10 A (GPR-1810HD)
High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 20 A (GPR-1820HD)
B&K Precision 1739 DC Power Supply, 30V, 1A.
More Details
Float Switch for Water Storage Tank
Buy Float Switch for Water Storage Tank Items Including:
Storage Tank for Thermo Scientific Barnstead Metal Classic Stills, 10 (B3043)
Hinged float switch; polypropylene, 1/2" x 1/2" conduit connection (160450)
Hinged float switch; polypropylene, 5/8"-11 bulkhead internal mounting (164520).
More Details
Low Pressure Vacuum Gauges
Buy Low Pressure Vacuum Gauges Items Including:
4 1/2 Low-pressure Bellows Gauge, 100 Inwc Vacuum, 1/4 NPT
4 1/2 Low-pressure Bellows Gauge, 0 To 10 Inwc, 1/4 NPT
4 1/2 Low-pressure Bellows Gauge, 0 To 100 Inwc, 1/4 NPT
4 1/2 Low-pressure Bellows Gauge, 0 To 150 Inwc, 1/4 NPT.
More Details
4000 Ml Erlenmeyer Flask with Stopper
Buy 4000 Ml Erlenmeyer Flask with Stopper Items Including:
PYREX Brand 4995 flask; 4000 mL, case of 1
Erlenmeyer flask mantle for 4000-ml flask, 330 watts, 230 VAC (100AO933)
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 1000 mL
Thermo Scientific Nalgene polypropylene Erlenmeyer flask, 2000 mL.
More Details
Hamilton Lab Furniture
Buy Hamilton Lab Furniture Items Including:
1 Drawer Wood Lab Table With Epoxy Resin Top 36in W X 30in H X 24in D (YO-41527-00)
11-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-00)
12-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-05).
More Details
Oakton Ph Con 10 Meter
Buy Oakton Ph Con 10 Meter Items Including:
Oakton Waterproof pH/Con 10 Meter with pH/conductivity probe and NIST-
Oakton pH Tutor Benchtop Meter
Oakton pH 700 Benchtop Meter, double-junction refillable glass pH elec (pH 700) (YO-35419-10)
Oakton pH 700 Benchtop Meter, meter and probe stand.
More Details
Aeromatic Low Pressure Air Gauge
Buy Aeromatic Low Pressure Air Gauge Items Including:
4 1/2 Low-pressure Bellows Gauge, 100 Inwc Vacuum, 1/4 NPT
4 1/2 Low-pressure Bellows Gauge, 0 To 10 Inwc, 1/4 NPT
4 1/2 Low-pressure Bellows Gauge, 0 To 100 Inwc, 1/4 NPT
4 1/2 Low-pressure Bellows Gauge, 0 To 150 Inwc, 1/4 NPT.
More Details
Ika T25 Digital Ultra Turrax
Buy Ika T25 Digital Ultra Turrax Items Including:
IKA T 25 ULTRA-TURRAX High-Speed Homogenizer 115VAC-unit only
IKA T 25 ULTRA-TURRAX High-Speed Homogenizer 220VAC-unit only
IKA T 50 ULTRA-TURRAX Heavy-Duty Homogenizer 115VAC-unit only
IKA T 50 ULTRA-TURRAX Heavy-Duty Homogenizer 230VAC-unit only.
More Details
Portable Centrifuge
Buy Portable Centrifuge Items Including:
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Sho (YO-17300-39)
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Sho (YO-17300-41)
Cole-Parmer FS-3500 Portable six-place clinical centrifuge with 18-pla.
More Details
200 Mesh Screen
Buy 200 Mesh Screen Items Including:
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 200 µm, pack of 3
Sieve, brass, 200 mm OD, 0.075 mm openings, full height, No
200 mesh
Mesh Screen, #200 screen size (YO-99035-54)
Standard Sieve, Stainless Steel Mesh, 0.075 mm sieve opening, No
More Details
Haake Falling Ball Viscometers
Buy Haake Falling Ball Viscometers Items Including:
Thermo Scientific HAAKE Falling Ball Viscometer
Gilmont Falling Ball Viscometer, 20 to 200 / 100 to 1000 centipoise
Optional tantalum ball for Gilmont falling ball viscometers
Gilmont Falling Ball Visometer, 2 to 20 / 10 to 100 centipoise.
More Details
Stainless Sanitary Ferrule 1 2
Buy Stainless Sanitary Ferrule 1 2 Items Including:
Barbed to sanitary adapter, 1/2" to mini sanitary, PVDF (YO-31513-63)
Barbed to sanitary adapter, 1/2" to mini sanitary, PVDF, pack of 10 (YO-31513-82)
Barbed to sanitary adapter, 1/2" to mini, animal-free polypropylene (YO-31513-14).
More Details
Tygon Tubing Sizes
Buy Tygon Tubing Sizes Items Including:
Masterflex® Tygon® tubing, nonphthlate/non-DEHP, 1.85mm ID, 1
Test tube racks for 1.5 mL tube size; PP (YO-06703-40)
Test tube racks for 0.5 mL tube size; PP (YO-06703-41)
Masterflex B/T Peroxide-Cured Silicone Tubing, Size 87, 3 ft (YO-96406-87).
More Details
Masterflex Pumps
Buy Masterflex Pumps Items Including:
L/S 16 GORE Style 100SC Tubing for Masterflex Pumps, 12" per pack
L/S 17 GORE Style 100SC Tubing for Masterflex Pumps, 12" per pack
L/S 18 GORE Style 100SC Tubing for Masterflex Pumps, 12" per pack.
More Details
Stir-Pak Laboratory Mixers
Buy Stir-Pak Laboratory Mixers Items Including:
Stir-Pak speed controller, 115 VAC
Stir-Pak High-speed, low-torque mixer system; 60-6,000 rpm, 115 VAC
Tank clamp for overhead mixers (YO-04552-65)
316 Stainless Steel Shaft, 3/8" dia x 24"L (accepts a 5/16" bore dia p (YO-04552-00).
More Details
70618
results
<<<Previous
4
5
6
7
8
9
10
11
12
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Leybold Vacuum Pump Cold Traps to Stir-Pak Laboratory Mixers
>
31125329-s-toc_2 31125329stoc2