Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 80
Self Acting Temperature Control Valve
to
Low Range Conductivity Calibration Solutions
Image
Product Detail
More Detail
Self Acting Temperature Control Valve
Buy Self Acting Temperature Control Valve Items Including:
Brass, Two-Way, Direct-Acting Valve (Normally Closed), 1/4" NPT(F), 5/ (8262H202-120/60)
Brass, Two-Way, Direct-Acting Valve (Normally Closed), 1/4" NPT(F), 7/ (8262H208-120/60).
More Details
100 Ml Test Tube
Buy 100 Ml Test Tube Items Including:
Pyrex Brand 9800 test tube; 9 mL, pack of 72
Pyrex Brand 9800 test tube; 9 mL, case of 720
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Lon (YO-17300-25)
Centrifuge Tube Insert for Automatic Heated Oil Test Rotor, 100 mL Con.
More Details
Plastic Hinged Lid Sample Containers
Buy Plastic Hinged Lid Sample Containers Items Including:
Hinged-lid vial, 1.25" OD, 1.5 mL, 1000/CS (YO-06097-02)
Hinged-lid vial, 1.25" OD, 3.3 mL, 1000/CS (YO-06097-04)
Hinged-lid vial, 1.25" OD, 5.3 mL, 1000/CS (YO-06097-06)
Hinged-lid vial, 1.25" OD, 7.7 mL, 1000/CS (YO-06097-08).
More Details
Female Threaded Tube Adapter
Buy Female Threaded Tube Adapter Items Including:
Compression threaded adapter, PFA, 1/8" tubing OD x 1/8" female NPT (YO-31320-50)
Compression threaded adapter, PFA, 1/8" tubing OD x 1/4" female NPT (YO-31320-51).
More Details
Branson Large Capacity Ultrasonic Cleaners
Buy Branson Large Capacity Ultrasonic Cleaners Items Including:
Ten-gallon heavy-duty ultrasonic cleaner, 120 VAC (000-914-506 DHA-1000)
Large Capacity Stainless Steel Ultrasonic Cleaners, 10 gal, 208VAC, 25 (CPN-908-011)
Large Capacity Stainless Steel Ultrasonic Cleaners, 10 gal, 208VAC, 40 (CPN-908-012).
More Details
Autoclavable Nalgene Bottle Lids
Buy Autoclavable Nalgene Bottle Lids Items Including:
Thermo Scientific Nalgene Wash bottle, FEP, 125mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 250mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 1000mL, 1/pk
Thermo Scientific Nalgene PPCO dispensing bottle, 125 mL.
More Details
Pyrex Amber Media Bottles
Buy Pyrex Amber Media Bottles Items Including:
500 mL Kimax Amber Media Bottles
1000 mL Kimax Amber Media Bottles
2000 mL Kimax Amber Media Bottles
5000 mL Kimax Amber Media Bottles
Pyrex Brand 1395 media storage bottle w/screw cap, 1 L
More Details
Stainless Steel Tubing Cooling System
Buy Stainless Steel Tubing Cooling System Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Luer Lock Fittings Valve
Buy Luer Lock Fittings Valve Items Including:
adapter; female luer lock to two-way valve; Kynar body and PP insert
Adapter, PP, Female luer lock to three-way valve; (Kynar body and PP i (YO-31200-80)
Panel mount lock ring, black nylon, 25/pk (YO-45509-00).
More Details
Cole-Parmer Digi Sense
Buy Cole-Parmer Digi Sense Items Including:
Cole-Parmer 100 ohm Platinum RTD Probes; 5"L
Cole-Parmer 100 ohm Platinum RTD Probes; 9"L
Type E High-termperature Thermocouple probes; 12"L (050797B-E)
Type K High-temperature Flexible thermocouple probe; 12"L (050797B-K).
More Details
Activated Carbon Capsule Filters
Buy Activated Carbon Capsule Filters Items Including:
Double open end granular activated carbon cartridge; 20 µm, 10"L (155155-43)
Activated Carbon After-Filter
Double open end activated carbon-impregnated paper cartridge; 5 µm, (155002-03)
Activated Carbon Refill for 33703-04, -05 (YO-33703-07).
More Details
Large Pipette
Buy Large Pipette Items Including:
Mla™ D-tipper Large Tip Pipette, Red, 200µL
Transfer Pipette, 5.8ml, Graduated, Large Bulb, 500/pk (YO-06226-61)
Transfer Pipette 7.5ml Graduated Large Bulb 500/pk (YO-06226-63)
10 mL Cole-Parmer large-tip opening serological pipette (YO-25562-26).
More Details
Small Simple Hydraulic Piston Pumps
Buy Small Simple Hydraulic Piston Pumps Items Including:
Hydraulic Piston Pump, 31 mL/min, 115/230 VAC (#405)
Hydraulic Piston Pump, 104 mL/min, 115/230 VAC (#425)
Hydraulic Piston Pump, 195 mL/min, 115/230 VAC (#429)
Hydraulic Piston Pump, 390 mL/min, 115/230 VAC (#437).
More Details
Low Volume Continuous Duty Pump
Buy Low Volume Continuous Duty Pump Items Including:
All-plastic, continous-duty double-diaphragm pump, polypropylene body/
industrial Hose Pump, Natural Rubber Hose, 57 GPM, continuous duty, 7.
industrial Hose Pump, Natural Rubber Hose, 3.1 GPM, continuous duty, 1.
More Details
Eye Protection Face Shield Net
Buy Eye Protection Face Shield Net Items Including:
Pro-Shield™ Face Shield Kit with helmet, safety glasses, protective
Sperian Protecto-Shield Replacement face shield only, Polycarbonate
Uvex Bionic Face Shield, Clear, Antifog coating
Thermo Scientific Nalgene Wraparound Face Shield, Polycarbonate.
More Details
High Voltage Function Generator
Buy High Voltage Function Generator Items Including:
High-Speed Function Generator, 5 MHz (GFG-8250A)
High-Speed Function Generator, 3 MHz, No Display (GFG-8215A)
B&K Precision 4005DDS Function Generator, 5 MHz
Tektronix® AFG3021B, Single Channel, Arbitrary/Function Generator, (AFG3021B).
More Details
Intrinsically Safe Anemometer
Buy Intrinsically Safe Anemometer Items Including:
Intrinsically safe barrier; for use with intrinsically safe transmitte (YO-43312-12)
GE (Druck) Intrinsically Safe Advanced Modular Calibrator with HART Co (DPI 620 IS) (YO-50015-02)
Thermo Scientific Intrinsically Safe Portable Conductivity Meter.
More Details
Roller Bottle Apparatus
Buy Roller Bottle Apparatus Items Including:
Thermo Scientific Nunc TufRol™ Roller Bottle, pleated surface, pac (YO-01836-23)
Wheaton Mini Bottle Benchtop Roller Culture System, 120V North America
Wheaton Mini Bottle Benchtop Roller Culture System, 230V Continental E.
More Details
Spirax Sarco Thermostatic Steam Traps
Buy Spirax Sarco Thermostatic Steam Traps Items Including:
Cast iron float and thermostatic steam trap (in-line horizontal config (FTI-15 1/2" 66664 )
Cast iron float and thermostatic steam trap (in-line horizontal config (FTI-15 3/4" 66669 ).
More Details
300 Ml Amber Volumetric Flask
Buy 300 Ml Amber Volumetric Flask Items Including:
PYREX Brand 5640 Volumetric Flask; 100 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 200 mL, pack of 6
PYREX Brand 5640 Volumetric Flask; 1000 mL, pack of 1
PYREX Brand 5640 Volumetric Flask; 2000 mL, pack of 1.
More Details
Threaded Brass Tube
Buy Threaded Brass Tube Items Including:
Threaded adapter, brass, 1/8" NPTM x 3/8" tubing ID, 5/pack (YO-30904-00)
Threaded adapter, brass, 1/8" NPTM x 1/4" tubing ID, 5/pack (YO-30904-01).
More Details
John Guest Push-to-Connect Fittings
Buy John Guest Push-to-Connect Fittings Items Including:
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD
Push-to-connect soft tube supports, 1/2" stem x 3/8" tube OD, 10/pack (YO-34007-39)
Push-to-connect soft tube supports, 5/16" stem x 1/4" tube OD, 10/pack (YO-34007-40).
More Details
Oakley Safety Glasses with Interchangeable Lenses
Buy Oakley Safety Glasses with Interchangeable Lenses Items Including:
Uvex Genesis X2 Safety Glasses, Silver/Navy frame glasses, clear lense
Uvex Genesis Translucent blue frame glasses with clear lenses
Uvex Genesis Translucent blue frame glasses with indoor/outdoor lenses.
More Details
Inline Explosion Proof Flow Meter
Buy Inline Explosion Proof Flow Meter Items Including:
Flowline EchoSafe Explosion-Proof Ultrasonic Level transmitter (2-wire (XP88-0)
Panel mount explosion proof with loop-powered backlight
Panel mount explosion proof with heater
Cole-Parmer® Low-Cost Loop-Powered Meter, Explosion Proof (YO-94799-04).
More Details
Type S Quartz Cuvettes
Buy Type S Quartz Cuvettes Items Including:
Suprasil Quartz Cuvettes for Turner Quantech™ Fluorometer
Standard cuvettes; cover top; quartz; 3.5 mL capacity, 2/pack
Fluorimeter cuvettes; cover top; quartz; 3.5 mL capcity, 2/pack
Standard cuvettes; stopper top; quartz; 3.5 mL capacity, 2/pack.
More Details
Allsafe Safety Glasses Z87
Buy Allsafe Safety Glasses Z87 Items Including:
Allsafe SMC™ Nemesis™ Safety Glasses, Black frame, Clear lens
Allsafe MonoGoggle, Regular, Clear Lens
Allsafe MonoGoggle, Large, Clear Lens
Allsafe MonoGoggle XTR™, Blue Frame, Clear Lens
Allsafe Shield/MonoGoggle XTR™, Blue Frame, Clear Lens.
More Details
Parker Hannifin Fittings
Buy Parker Hannifin Fittings Items Including:
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD x (YO-06444-10)
Parker compression union tee, stainless steel, 5/8" (YO-30486-31)
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD (YO-06444-00).
More Details
Amico Dial Type Flow Meter
Buy Amico Dial Type Flow Meter Items Including:
Paddle Wheel Flow Meter, Brass, 0.2 to 10 GPM (SEB-050)
Electromagnetic Flowmeter, PVC Flow Sensor And Tee Fitting- 1"; 0.2 To (EX81-P-EF81T-P-100)
Electromagnetic Flowmeter, PVC Flow Sensor And Tee Fitting- 1.5"; 0.2 (EX81-P-EF81T-P-150).
More Details
Npt Female Compression Fitting
Buy Npt Female Compression Fitting Items Including:
Compression fittings, female pipe adapter, straight, Black PP, 1/4" OD (YO-06444-11)
Compression fittings, female pipe adapter, straight, Black PP, 3/8" OD (YO-06444-12).
More Details
Low Range Conductivity Calibration Solutions
Buy Low Range Conductivity Calibration Solutions Items Including:
Conductivity calibration standard, 1000 µmhos (3161)
MYRON L conductivity calibration standard, 23.8 µS
MYRON L conductivity calibration standard, 46.7 µS
MYRON L conductivity calibration standard, 2060 µS.
More Details
70618
results
<<<Previous
76
77
78
79
80
81
82
83
84
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
100
110
120
130
140
150
160
170
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Self Acting Temperature Control Valve to Low Range Conductivity Calibration Solutions
>
31125329-s-toc_2 31125329stoc2