Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 800
Rubbermaid Laboratory Supplies & Consumables
to
Cole Density
Image
Product Detail
More Detail
Rubbermaid Laboratory Supplies & Consumables
Buy Rubbermaid Laboratory Supplies & Consumables Items Including:
Rubbermaid Yellow LDPE Lid for Storage Container with Handles 63511-01
Rubbermaid 2 cup scoop, PE (CPFG9G5100-CLR)
Rubbermaid 3 cup scoop, PE (CPFG9G5200-CLR)
Rubbermaid 4 cup scoop, PE (CPFG9G5300-CLR)
Rubbermaid 8 cup scoop, PE (CPFG9G5400-CLR).
More Details
Ika Heating Equipment
Buy Ika Heating Equipment Items Including:
IKA Ceramic Hot Plate, 4" x 4", 115v (YO-04671-60)
IKA Ceramic Hot Plate, 4" x 4", 230V (YO-04671-62)
IKA Ceramic Hot Plate, 10" x 10", 115v (YO-04671-68)
IKA Ceramic Hot Plate, 10" x 10", 230v (YO-04671-70).
More Details
Btx Electroporators
Buy Btx Electroporators Items Including:
Electroporation system, exponential decay (YO-02894-10)
Electroporation system, square wave (YO-02894-14)
Electroporation system, square wave (YO-02894-18)
Electroporation system (YO-02894-04)
cuvettes plus, 1 mm gap, sterile, 50/pk (YO-02894-30).
More Details
Electrothermal Extraction Apparatus
Buy Electrothermal Extraction Apparatus Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal Electromantle Extraction Heater - 3 positions, 1000 mL, (EME3 1000/CEBX6)
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0100/CEX1).
More Details
Labconco Replacements Accessories
Buy Labconco Replacements Accessories Items Including:
Labconco Replacement prefilter for 36" horizontal clean benches
Labconco detergent for FlaskScrubber Vantage (4522000)
Labconco HEPA Filter for 4 & 8 ft horizontal clean benches
Replacement Prefilters for 2' W Verticle Bench (YO-09114-64).
More Details
Buffer Solution, Ph 10.00
Buy Buffer Solution, Ph 10.00 Items Including:
Oakton pH buffer pouches; pH 10.00, 20/pk
Oakton buffer pack, 500-ml each of 4.01, 7.00, and 10.00 buffer
Oakton 10.00 pH buffer, 500-mL, case of 12 (YO-05942-61)
Oakton 10.00 pH buffer, 1 L (YO-05942-62)
Oakton 10.00 pH buffer, 4 L (YO-05942-64).
More Details
Personal Multigas Monitor
Buy Personal Multigas Monitor Items Including:
Rae Four-Gas Personal Protection Monitor, Datalogging Montor with Conf (027-0000-011)
PID sampling unit with datalogger for multiRAE personal multi gas moni (009-3121-010)
ToxiRAE Plus Pocket Monitor, standard model, pocket size (YO-86261-00).
More Details
Laboratory Reagents
Buy Laboratory Reagents Items Including:
METHANOL LABORATORY GRADE 4L (YO-88051-79)
2-PROPANOL LABORATORY GRADE 4L (YO-88052-13)
Spatula, "Reagent Digger", 304SS, 9" L, fits openings down to 15/16" d
SHUR/Clean™ Reagent Bowl rack.
More Details
Lab Filtration
Buy Lab Filtration Items Including:
Cole-Parmer Compact Filtration System, PP/PVC
Cole-Parmer Compact Filtration System, CPVC, 30 psi max pressure
Reusable water filtration element without housing; 100 µm (R10-2155)
Reusable water filtration element without housing; 50 µm (R10-2300).
More Details
Bower Digital Cleaning Solution Kit
Buy Bower Digital Cleaning Solution Kit Items Including:
Thermo Scientific Orion pH Electrode Cleaning Solution Kit
Needle cleaning kit; includes 70-mL of cleaning solution and five tube (YO-07938-74)
Bleach Spray System Kit, (includes six, 11-oz
bleach cartridges, two, (77062).
More Details
Delta Monitor Temperature Control Removable
Buy Delta Monitor Temperature Control Removable Items Including:
CALgrafix Process Monitoring and Configuration Software (Add a license (1005GB000)
Batch Controller, 12-24 VAC/dc (3-5600)
WSC-5-SDOR : Wind Speed Controller Single Set Pt.
Temperature Sensor Cable for Hose Barb Fitting, 1/4" phone jack, 10'.
More Details
Druck Cole-Parmer Modular Calibrators
Buy Druck Cole-Parmer Modular Calibrators Items Including:
DPI 620 Calibrator Carrying Case (IO620-CASE-1)
DPI 620 Modular system transit case (IO620-CASE-4)
IDOS to DPI 620 Adaptor
Comparator adaptor (IO620-COMP)
DPI 620 system carry case (IO620-CASE-2)
More Details
Mce Membranes
Buy Mce Membranes Items Including:
Mixed Cellulose Ester (MCE) membrane for asbestos monitoring; 25-mm di (YO-36230-30)
Mixed Cellulose Ester (MCE) membrane for asbestos monitoring; 37-mm di (YO-06639-72)
MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia (YO-29959-00).
More Details
Lab Flooring Products
Buy Lab Flooring Products Items Including:
250 L Floor Model Heated Shaking Incubator - 120VAC 60Hz
250 L Floor Model Refrigerated Shaking Incubator - 230VAC 60Hz
Floor Stand for 81781-00
Floor Stand for 81781-02
Hard floor caster for plastic base, pack of five
More Details
Drumquik Pur
Buy Drumquik Pur Items Including:
DrumQuik PUR Drum Adapter System Kit, 985 mm tube and 3/4" flare liqui (YO-31608-50)
DrumQuik PUR Drum Adapter System Kit, 985 mm tube and 3/4" flare liqui (YO-31608-51)
DrumQuik PUR Drum Coupler, PVDF with FKM seals (YO-31608-70).
More Details
Hm Digital Waterproof Orp Meter
Buy Hm Digital Waterproof Orp Meter Items Including:
Oakton Waterproof ORP Testr 10 BNC Pocket ORP Tester
Oakton Waterproof ORPTestr 10 Pocket ORP Tester
Compact Waterproof Digital Thermohygrometer (rh/temp/dew/wet) (1340-5621(TFH620+TPH100))
Horiba Waterproof Digital Conductivity Simulator (377778).
More Details
Circular Chart with Display
Buy Circular Chart with Display Items Including:
Dickson 8" Temperature/humidity Recorder With Digital Display (TH802) (YO-80001-52)
Dickson 8" Temperature/humidity Recorder With Probe, Dig
Display And (TH803) (YO-80001-54)
Dickson 6" Temperature/Humidity Recorder with LCD Display.
More Details
Lab Bags
Buy Lab Bags Items Including:
Replacement deionizer bag
Disposable Paper Bags, 5/pk
Polypropylene bag filter housing, 1" NPT(F) (150360)
Polypropylene bag filter housing, 1-1/2" NPT(F) (150338)
General-purpose aluminum filter bag housing, 1-1/4" NPT(F) (156109).
More Details
Fluorescent Tracers
Buy Fluorescent Tracers Items Including:
Flourescent FWT Red Dye Tablets, 200 tablets
Flourescent FLT Yellow/Green Dye Tablets, 200 tablets
Flourescent FLT Orange Dye Tablets, 200 tablets
Flourescent FLT Yellow/Green Dye Concentrate, Gallon
Flourescent FLT Orange Dye Concentrate, Gallon.
More Details
Ika Mixer T 50 I Ul
Buy Ika Mixer T 50 I Ul Items Including:
IKA Economical Analytical Mill, 115 VAC, 50/60 Hz
IKA Economical Analytical Mill, 230 VAC, 50/60 Hz
IKA Heavy-Duty Analytical Mill, 230 VAC/50 Hz
IKA ULTRA TURRAX Tube Disperser, 100-240 VAC, 50/60 Hz
IKA ULTRA TURRAX Stirring Tube 50 mL, (10/pk).
More Details
Roll on Adhesive
Buy Roll on Adhesive Items Including:
Handheld adhesive roll mop, 3"W roller (6-303)
Handheld adhesive roll mop; 9" wide (6-301)
Telescopic adhesive roll mop; 18" wide (6-300)
Cole-Parmer Extra-Thick PTFE Adhesive Tape, 2" W, 20mil, 15ft/roll (YO-08277-12).
More Details
Cole-Parmer Vibration Measurement
Buy Cole-Parmer Vibration Measurement Items Including:
Analog Vibration Meter (211-S-100)
Measuring Cable, Lemo to BNC Slip-on, 1.5 M
Vibration meter (6400-012)
Vibration meter kit with sensor pack (6400-011)
Vibration meter kit with sensor pack and software (6400-010)
More Details
Surgical Grade Forceps
Buy Surgical Grade Forceps Items Including:
Brown-Adson Tissue Forceps, Dissecting-grade (YO-10918-26)
Halsted mosquito forceps, Dissecting-grade, Straight (YO-10918-14)
Halsted mosquito forceps, Dissecting-grade, Curved (YO-10918-16)
Kelly forceps, Dissecting-grade, Straight (YO-10918-18).
More Details
Online Process Ph Meter
Buy Online Process Ph Meter Items Including:
HF Scientific Online Chlorine Monitor, 100 to 240 VAC, 47 to 63 Hz (20040)
HF Scientific 60 Day Supply of Total Chlorine Reagent, powder (09954)
HF Scientific 12 Month Supply of Free Chlorine Reagent, powder (09955).
More Details
Drying Tubes
Buy Drying Tubes Items Including:
Drierite drying tube; 4/pack (26930 6" DRYING TUBE)
Drying tube
Protects sensitive reagents from moisture or CO2/s (707930)
Dry Small Diameter Sampling Tube, 12" Length (PN007)
Thermo Scientific Heating block for 6 mm tubes, 30 openings.
More Details
Stainless Steel Diaphragm
Buy Stainless Steel Diaphragm Items Including:
Sandpiper Air-Powered Double-Diaphragm Stainless Steel Pump, 316 SS, 4 (E02B1SYSSNS000.)
Sandpiper Air-Powered Double-Diaphragm Stainless Steel Pump, 316 SS, 4 (SB1,SS4SS.)
Surge Suppressors, 1" Flange, Stainless Steel Body, PTFE-Neoprene Diap.
More Details
Humidity Controller with Multiple Input Points
Buy Humidity Controller with Multiple Input Points Items Including:
Digi-Sense 1-Zone benchtop dual display Limit control, type J input, a (YO-36225-76)
1/16 DIN controllers with universal input and fuzzy logic, two SPST re (16A2133)
1/16 DIN controllers with universal input and fuzzy logic, two SSR and (16A2111).
More Details
Round Tubing Caps
Buy Round Tubing Caps Items Including:
Thermo Scientific Nunc CryoTube cryogenic tube with round bottom; 1.8 (368632)
Thermo Scientific Nunc CryoTube cryogenic tube with round bottom; 1.8 (363401)
BIOSIMPLEX CARBOY EZ TOP ASSEMBLY, 20L, 2 PORTS, 3/8 X 5/8 TUBING, inc.
More Details
Pressure Meter Test
Buy Pressure Meter Test Items Including:
Accu-cal Digital Test Gauge, Vac To 5000 Psi, W/ Flange, Loop Powered
Accu-cal Digital Test Gauge, Vac To 10,000 Psi, W/ Flange, Loop Powere
High-accuracy digital test gauge, 0 to 100 psi range
High-accuracy digital test gauge, 0 to 1000 psi range.
More Details
Cole Density/Specific Gravity
Buy Cole Density/Specific Gravity Items Including:
Specific Gravity Cup, 83.2 mL capacity, 0.5% volume tolerance, for use (PV-9654)
Low-Cost Specific Gravity Meter, 0.750 to 1.000 SGU, °C model (DSG-11-075100-C-B)
Specific Gravity Cup with Tare Weight, 8.32 mL capacity, 1.2% volume t (WG-SS-8.32/T).
More Details
70618
results
<<<Previous
796
797
798
799
800
801
802
803
804
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
710
720
730
740
750
760
770
780
785
786
787
788
789
790
791
792
793
794
795
796
797
798
799
800
801
802
803
804
805
806
807
808
809
810
811
812
813
814
820
830
840
850
860
870
880
890
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
2,000
2,354
Rubbermaid Laboratory Supplies & Consumables to Cole Density
>
31125329-s-toc_2 31125329stoc2