Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 90

Rubber Tubing 2   to   Light Meter Calibration Standards

Image
  Product Detail
More Detail

Rubber Tubing 2.5 Mm

Buy Rubber Tubing 2.5 Mm Items Including:
  • Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
  • Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25')
  • Gum Rubber Tubing, 5/16" x 7/16", 25' per pack (138PXA-25').

Schott Ph/Ion Laboratory Electrodes

Buy Schott Ph/Ion Laboratory Electrodes Items Including:
  • Glass electrodes with removable cable
  • Epoxy electrodes with removable cable
  • Glass electrodes with BNC fixed cable (285129155(BLUE LINE#15PH))
  • Epoxy electrodes with DIN fixed cable
  • Epoxy pH electrodes with BNC connection.

Male to Male Luer Lock Connector

Buy Male to Male Luer Lock Connector Items Including:
  • Male luer lock plug, PP, 25/pk (YO-45503-56)
  • Male luer lock plug, Nylon, 25/pk (YO-45505-56)
  • Male luer with lock ring x female luer coupler, PP, 25/pk (YO-45500-80)
  • Luer fittings, Polycarbonate fittings, female x male lock, rotating, (YO-06464-95).

Dissolved Oxygen Meters Optical

Buy Dissolved Oxygen Meters Optical Items Including:
  • YSI ProODO™ Optical Dissolved Oxygen Meter
  • Thermo Scientific Orion 3-Star™ RDO® Optical Dissolved Oxygen M (1213301)
  • Thermo Scientific Orion 3-Star™ RDO® Optical Dissolved Oxygen M (1213302)
  • Replacement Optical Cap
  • Optical dissolved oxygen sensor.

30mm X 115 Mm Tube Racks

Buy 30mm X 115 Mm Tube Racks Items Including:
  • Scienceware test tube rack for 10- to 13-mm tubes
  • Scienceware polypropylene test tube rack for 26-30 mm tubes
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.

Polyethylene Tubing 1 8 X 170

Buy Polyethylene Tubing 1 8 X 170 Items Including:
  • Flexelene SFX Flexible Polyethylene Tubing, 1/8 x 1/4, 100' per pack (YO-96180-03)
  • Flexelene SFX Flexible Polyethylene Tubing, 1/4 x 3/8, 100' per pack (YO-96180-05)
  • Flexelene SFX Flexible Polyethylene Tubing, 3/8 x 1/2, 100' per pack (YO-96180-07).

Polystat Chiller

Buy Polystat Chiller Items Including:
  • Cole-Parmer Polystat low-temperature immersion chiller; cold-finger; 1
  • Cole-Parmer Polystat recirculator with force/suction pump, 21 lpm, 250 (YO-13042-05)
  • Cole-Parmer Polystat recirculator with force/suction pump, 21 lpm, 250 (YO-13042-07).

Ion Selective Electrode Principle

Buy Ion Selective Electrode Principle Items Including:
  • Cole-Parmer Combination Ion Selective Electrodes, Bromide (Br)
  • Cole-Parmer Combination Ion Selective Electrodes, Copper (Cu+2)
  • Cole-Parmer Combination Ion Selective Electrodes, Cyanide (CN)
  • Cole-Parmer Combination Ion Selective Electrodes, Nitrate.

Chemical Compatibility Chart

Buy Chemical Compatibility Chart Items Including:
  • byko-charts for Opacity, 5.50" x 10.00", 2A, 250 per box
  • byko-charts for Opacity, 7.63" x 10.25", 2C, 250 per box
  • Chart Paper; 7 day, graduations from 0 to 14, box of 100 sheets
  • 8" Chart Paper, -50 to 50 F/C, 31 Day, 60/pk.

Microbiology Agar Plates

Buy Microbiology Agar Plates Items Including:
  • Prepared agar plate, TSA, 10/pk
  • Prepared agar plate, Standard Methods agar, 10/pk
  • Prepared agar plate, Mannitol Salt agar, 10/pk
  • Prepared agar plate, Sab Dex, 10/pk
  • Prepared agar plate, XLD agar, 10/pk

Plastic Specific Gravity

Buy Plastic Specific Gravity Items Including:
  • Durac Specific Gravity Polycaronate Hydrometers, 1.000 to 1.180, 0.002
  • Durac Specific Gravity Polycaronate Hydrometers, 0.900 to 1.080, 0.002
  • Durac Specific Gravity Polycaronate Hydrometers, 1.050 to 1.220, 0.002.

Press Fit Hose Connector

Buy Press Fit Hose Connector Items Including:
  • Barbed Press-in Plug, 5/32", Polypropylene, 10/pk (YO-31220-04)
  • Barbed Press-in Plug, 1/4", Polypropylene, 10/pk (YO-31220-06)
  • Barbed Press-in Plug, 3/8", Polypropylene, 10/pk (YO-31220-07).

Ph Meter Computer Interface

Buy Ph Meter Computer Interface Items Including:
  • Data transfer adapter connects colorimeter to printer or your computer
  • RS232 Interface cable for Star series meters
  • RS-232 to USB interface, includes RS-232 cable, RS-232 to USB adapter, (YO-18003-83)
  • Star series printer with RS232 interface cable.

Corning Pc 100 Hot Plate

Buy Corning Pc 100 Hot Plate Items Including:
  • Stirring hot plate, 4" x 5", 120 VAC
  • Corning Digital Hot Plate, 5" x 7", 120 VAC
  • Corning Digital Hot Plate, 5" x 7", 220 VAC
  • Economical Hot Plate, 120 VAC, 60 Hz
  • Corning PC-171 Scholar Stirrer - 5x5, 120V (6795-171).

Stainless Steel Capillary Tube Fittings

Buy Stainless Steel Capillary Tube Fittings Items Including:
  • 160-18 Stainless Steel Pitot Tube 18" Insertion Length 5/16" Diameter
  • 166-12 Stainless Steel Pitot Tube 12" Insertion Length 1/8" Diameter
  • Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24).

K Type Thermocouple Pipe Clamp

Buy K Type Thermocouple Pipe Clamp Items Including:
  • Type T Pipe-fitting thermocouple probes; 12"L immersion depth (072997D-T)
  • Type T Pipe plug thermocouple probes; 1/4"NPT (M); 0.5"L (050597A-T)
  • Type K Pipe Plug Thermocouple Probes; 1/4" NPT (M); 0.5"L (050597A-K).

1 Double Barbed Brass Fitting

Buy 1 Double Barbed Brass Fitting Items Including:
  • Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
  • Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
  • Coupling bodies; hose barb valved fittings; 1/4" ID; 2.00" length (LCD17004).

Polycarbonate Containers

Buy Polycarbonate Containers Items Including:
  • Square polycarbonate container with handles, 12 quart
  • Polycarbonate wide-mouth container with handles, 12 quart
  • Round Container with Handle, PC, 1 Qt, 12 Pk
  • Round Container with Handle, PC, 2 Qt, 12 Pk
  • Round Container with Handle, PC, 4 Qt, 12 Pk.

Evaporative Sample Concentrator

Buy Evaporative Sample Concentrator Items Including:
  • Heated Evaporator/Concentrator; single block with heated manifold gas, (YO-28690-05)
  • Heated Evaporator/Concentrator; single block, 20 test tubes; 15- to 16 (YO-28690-10)
  • Heated Evaporator/Concentrator; single block for 96-well plates (YO-28690-30).

Cyanuric Acid Test Kit

Buy Cyanuric Acid Test Kit Items Including:
  • Liquid Sanitizer Strength Verification Kits, Peracetic Acid Test (7191-01)
  • Test Strips, Peracetic acid, 0-160ppm, 50 strips/vial (3000)
  • Total Phosphate and Acid Hydrolyzable PhosphateTube, 50 tests
  • On Guard Acid Detecting Paint, Gallon (AA-001-01).

Thick Walled Steel Tubing

Buy Thick Walled Steel Tubing Items Including:
  • 316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
  • 316 Stainless Steel Tubing; dimensions, 3/16" ID, 1/4" OD, 1/32" wall; (YO-03300-10).

Low Temperature Lab Freezer

Buy Low Temperature Lab Freezer Items Including:
  • 2" Box Rack for Revco 3 cuft CxF Ultra-Low Temperature Chest Freezers.
  • 6" Chart Recorder for 3 cuft Revco CxF Ultra-Low Temperature Chest Fre
  • Standard low-temperature cabinets; 9.4 cu ft; -40°C; 230V (40-9.4 230V/50HZ).

Temperature Probe for Chest Freezer

Buy Temperature Probe for Chest Freezer Items Including:
  • 2" Box Rack for Revco 3 cuft CxF Ultra-Low Temperature Chest Freezers.
  • Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-D)
  • Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-V).

Stirrer Motor

Buy Stirrer Motor Items Including:
  • Stirrer motor with electronic speed controller; 208/240 VAC, (CE Marke (099C K5424CE)
  • Koehler manual flash point tester stirrer motor, 115 VAC/60 Hz
  • Koehler manual flash point tester stirrer motor, 220 VAC/50 Hz
  • Motorless Magnetic Stirrer, 120 VAC.

Pcc Series Brass Quick Connects

Buy Pcc Series Brass Quick Connects Items Including:
  • Metal quick-disconnect coupling body, straight-through, 1/4" NPT (M) t (LC10004)
  • Barbed fittings, Metal quick-disconnect fittings, coupling bodies,valv (LCD13006)
  • Barbed fittings, Metal quick-disconnect fittings, coupling bodies,stra (LC13006).

Fisher Accumet Ph Probe

Buy Fisher Accumet Ph Probe Items Including:
  • accumet AB150 pH Benchtop Meter Only
  • Accumet AB200 pH/Conductivity Benchtop Meter Only
  • Accumet AB150 pH Benchtop Meter Kit
  • Accumet AB150B pH Benchtop Meter Kit
  • Accumet AB200 pH/Conductivity Benchtop Meter Kit

Female Luer Lock Cap

Buy Female Luer Lock Cap Items Including:
  • Adapter, PP, 1/2" mini to female luer lock (YO-31200-50)
  • Adapter, Kynar, 1/2" mini to 1/2" to female luer lock
  • Female luer lock x 1/8" NPT, 316 SS, each (YO-31507-46)
  • Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48).

Plastic Gear Pump

Buy Plastic Gear Pump Items Including:
  • Miniature gear pump, 0.43 GPM (1600 mL/min) non-enclosed OEM model, 11
  • Miniature gear pump, 0.43 GPM (1600 mL/min) non-enclosed OEM model, 23
  • Miniature gear pump, .32 GPM (1200 mL/min), fully enclosed lab model, (400000/LGP-115).

5 Gal Plastic Carboys Foodgrade

Buy 5 Gal Plastic Carboys Foodgrade Items Including:
  • Economical Square Carboy w/handle, 5 Gal - HDPE, 1/ea (YO-06031-82)
  • Economical Square Carboy w/handle, 5 Gal - HDPE, 4/cs (YO-06031-86)
  • Cylidrical Tank with Cover and Spigot, HDPE, 5 Gal., 3/16" Wall (YO-06317-51)
  • PE Tank, 5 gal (YO-06925-58).

Light Meter Calibration Standards

Buy Light Meter Calibration Standards Items Including:
  • Dual-Scale Light Meter with NIST-traceable certificate of calibration (840020C)
  • Conductivity calibration standard, 1000 µmhos (3161)
  • MYRON L conductivity calibration standard, 23.8 µS
  • MYRON L conductivity calibration standard, 46.7 µS.



Rubber Tubing 2 to Light Meter Calibration Standards

>