Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 900
Stopwatch Large Display
to
Progressing Cavity Pumps
Image
Product Detail
More Detail
Stopwatch Large Display
Buy Stopwatch Large Display Items Including:
Large Display Pocket Timer (810046).
More Details
Kimax Glass Cylinders
Buy Kimax Glass Cylinders Items Including:
100 mL Kimax class B graduated cylinders
250 mL Kimax class B graduated cylinders
500 mL Kimax class B graduated cylinders
1000 mL Kimax class B graduated cylinders
2000 mL Kimax class B graduated cylinders
More Details
Flexible Heating Mantles
Buy Flexible Heating Mantles Items Including:
60° heating mantle with 12" top diameter, 400 watts, 230 VAC (100AO727)
60° heating mantle with 10" top diameter, 400 watts, 115 VAC (100AO724)
60° heating mantle with 12" top diameter, 400 watts, 115 VAC (100AO726).
More Details
Contec Easysat Bucketless Floor Mop Heads
Buy Contec Easysat Bucketless Floor Mop Heads Items Including:
Contec EasySat™ Sterile Textured Mop Head for Bucketless Floor Mop
Contec EasySat™ Mop Head for Bucketless Floor Mop
Contec EasySat™ Textured Mop Head for Bucketless Floor Mop.
More Details
Thermo Scientific Electrophoresis Power Supply Accessories
Buy Thermo Scientific Electrophoresis Power Supply Accessories Items Including:
Power supply adapter
Power cord, 6 ft L
Replacement fuse
Pk of two
Temperature probe provides temperature control from 0° to 90°C
More Details
Chromair Air Sampling and Gas Analysis
Buy Chromair Air Sampling and Gas Analysis Items Including:
ChromAir Color Comparator; Carbon monoxide
ChromAir Color Comparator; Formaldehyde.
More Details
Cole-Parmer High-Efficiency Polypropylene Absolute-Rated Filter Bags
Buy Cole-Parmer High-Efficiency Polypropylene Absolute-Rated Filter Bags Items Including:
Size 1 high-efficiency polypropylene absolute-rated filter bag; 200de (255112-03)
Size 1 high-efficiency polypropylene absolute-rated filter bag; 200de (255114-03)
Size 1 high-efficiency polypropylene absolute-rated filter bag; 200de (255115-03).
More Details
Repetitive Syringe Pipettor
Buy Repetitive Syringe Pipettor Items Including:
Repetitive syringe dispenser (NSTP)
Varipet repetitive syringe dispensers; volume range, 0.0 - 1.0 mL; inc
Varipet repetitive syringe dispensers; volume range, 0.0 - 10.0mL; inc
Syringe, 0.05 mL tip volume (NS-V)
Syringe, 0.5 mL tip volume (NS-S).
More Details
Vacuum Pumps Parts
Buy Vacuum Pumps Parts Items Including:
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 1
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.55 cfm, 2
Replacement Valox pump head assembly for all air Cadet pumps
Eccentric assembly for all Air Cadet pumps.
More Details
Vacuum Pump Racing Parts
Buy Vacuum Pump Racing Parts Items Including:
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 1
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.55 cfm, 2
Replacement Valox pump head assembly for all air Cadet pumps
Eccentric assembly for all Air Cadet pumps.
More Details
Precision Vacuum Pumps Parts
Buy Precision Vacuum Pumps Parts Items Including:
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 1
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.55 cfm, 2
Replacement Valox pump head assembly for all air Cadet pumps
Eccentric assembly for all Air Cadet pumps.
More Details
Chemical Storage Cabinets
Buy Chemical Storage Cabinets Items Including:
Vented Storage Cabinet, 45L Compact, Organic Filter
Vented Storage Cabinet, 45L Compact, Inorganic Filter
Vented Storage Cabinet, 100L Single, Inorganic Filter
Vented Storage Cabinet, 200L Double, Inorganic Filter.
More Details
Polystyrene Slide Holders
Buy Polystyrene Slide Holders Items Including:
Polystyrene slide holder; holds one hundred 75 x 25 mm slides (44577-0000)
Blue Polystyrene 100-Slide Holder (HS15994M OR HEA1594M)
White Polystyrene 25-Slide Holder (HS15989R).
More Details
Pneumatically Actuated 2-Position, 3-Way Elliptical Valves
Buy Pneumatically Actuated 2-Position, 3-Way Elliptical Valves Items Including:
Pneumatically Actuated 2-Position, 3-Way Elliptical Valve System; PP b (123-PP-RA4)
Pneumatically Actuated 2-Position, 3-Way Elliptical Valve System; PP b (423-PP-RA4).
More Details
Viton Check Valves
Buy Viton Check Valves Items Including:
Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm (YO-98553-00)
Nylon Miniature Check Valve, 3/16" hose barb, fluorosilicone diaphragm (YO-98553-01)
PVDF Miniature Check Valve, 3/16" hose barb, Viton diaphragm.
More Details
Scienceware Write-on Tape and Dispenser
Buy Scienceware Write-on Tape and Dispenser Items Including:
Scienceware; Write-On Tape, 1/2"W x 120', Pack of 6
Scienceware; Write-On Tape, 3/4"W x 120', Pack of 4
Scienceware; Write-On Tape, 1"W x 120', Pack of 3
Scienceware® write-on tape, 1/2"W (YO-08277-42).
More Details
Disposable Gloves
Buy Disposable Gloves Items Including:
Best N-DEX PLUS Non-latex Industrial Gloves; size, medium; 50/pack
Best N-DEX PLUS Non-latex Industrial Gloves; size, extra-large; 50/pac
Best N-Dex Laboratory Grade Gloves, low-powder, small
Best N-Dex Laboratory Grade Gloves, low-powder, X-large.
More Details
Corning Pipette
Buy Corning Pipette Items Including:
CORNING Stripette Serological Pipettes, 5.0 mL, Individually Wrapped P
CORNING Stripette Serological Pipettes, 2.0 mL, Individually Wrapped P
IsoTip, 0.1 to 10 µL, G tip, sterile, 960/pk (4807)
IsoTip, 1 to 30 µL, sterile, 960/pk (4821).
More Details
Pyrex Vista Beakers no 20075
Buy Pyrex Vista Beakers no 20075 Items Including:
Pyrex Vista Griffin Style Beakers, 100mL 12/pk (70000-100)
Pyrex Vista Griffin Style Beakers, 150mL 12/pk (70000-150)
Pyrex Vista Griffin Style Beakers, 250mL 12/pk (70000-250)
Pyrex Vista Griffin Style Beakers, 400mL 12/pk (70000-400).
More Details
Pfa Sanitary Clamp 90° Elbows
Buy Pfa Sanitary Clamp 90° Elbows Items Including:
90° elbow, PFA, 1/2" sanitary clamp (YO-31530-01)
90° elbow, PFA, 3/4" sanitary clamp (YO-31530-02)
90° elbow, PFA, 1" sanitary clamp (YO-31530-03).
More Details
Rite in the Rain Bound Books
Buy Rite in the Rain Bound Books Items Including:
Horizontal line bound book (390N).
More Details
Digital Scales
Buy Digital Scales Items Including:
Atago Digital Pocket Ethylene Glycol Refractometer (°C scale)
Atago Digital Pocket Propylene Glycol Refractometer (°C scale)
Atago Digital Pocket Propylene Glycol Refractometer (°F scale)
Atago Special-scale Refractometer, Salinity of Seawater.
More Details
Cole-Parmer Hydraulic Piston Pumps
Buy Cole-Parmer Hydraulic Piston Pumps Items Including:
Hydraulic Piston Pump, 31 mL/min, 115/230 VAC (#405)
Hydraulic Piston Pump, 104 mL/min, 115/230 VAC (#425)
Hydraulic Piston Pump, 195 mL/min, 115/230 VAC (#429)
Hydraulic Piston Pump, 390 mL/min, 115/230 VAC (#437).
More Details
True Union Ball Valve Spears
Buy True Union Ball Valve Spears Items Including:
Thermoplastic True Union Ball Valve, PVC with EPDM seat and seals, 1/2 (V07101N)
Thermoplastic True Union Ball Valve, PVC with EPDM seat and seals, 3/4 (V08101N)
Thermoplastic True Union Ball Valve, PVC with EPDM seat and seals, 1" (V10101N).
More Details
Low Pressure Gauge
Buy Low Pressure Gauge Items Including:
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 3 psi and 0 to 20 kPa
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 5 psi and 0 to 35 kPa
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 10 psi and 0 to 70 kPa
Pressure Gauge for Manifold 59620-61.
More Details
Power Meter Calibration
Buy Power Meter Calibration Items Including:
NIST-Traceable Calibration; Power Meter (Low Frequency) (YO-17101-34).
More Details
Handle for Round Brush
Buy Handle for Round Brush Items Including:
Handle for 84550-series round tank brushes, 42" long.
More Details
Uv Black Lights
Buy Uv Black Lights Items Including:
Black (UV) light ring lights, for 48402-series stereomicroscopes; 115V (YO-41210-00)
Black (UV) light ring lights, for 48403-series stereomicroscopes; 115V (YO-41210-10)
Black (UV) light ring lights, for 48920-series steromicroscopes; 115V.
More Details
Straight Carbofib Tweezers
Buy Straight Carbofib Tweezers Items Including:
Carbofib Tweezers 5" long with Tapered precision point for bending (18767)
Carbofib Tweezers 5" long with Sharp Fine tip (18760)
Straight: Ultra Fine Tips 5CF (18.762)
Straight: Soft and Strong Tips 302 (18.702).
More Details
Progressing Cavity Pumps
Buy Progressing Cavity Pumps Items Including:
Moyno progressing cavity pump, phenolic housing and rotor/nitrile stat (20551)
Moyno progressing cavity pump, 316 stainless steel housing and rotor/n (3910000251)
Moyno progressing cavity pump, phenolic housing and rotor/nitrile stat (3910000252).
More Details
70618
results
<<<Previous
896
897
898
899
900
901
902
903
904
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
800
810
820
830
840
850
860
870
880
885
886
887
888
889
890
891
892
893
894
895
896
897
898
899
900
901
902
903
904
905
906
907
908
909
910
911
912
913
914
920
930
940
950
960
970
980
990
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
1,800
2,000
2,354
Stopwatch Large Display to Progressing Cavity Pumps
>
31125329-s-toc_2 31125329stoc2