Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Micro 90 Detergent
Image
Product Detail
SKU
Cole-Parmer® Micro-90® cleaning solution, case of twelve 1 qt (YO-18100-00)
Category: Lab and General Purpose Cleaners
Description: Cole-Parmer
®
Micro-90
®
Cleaning Solution
Brand:
Micro-90
YO-18100-00
More Details
Cole-Parmer® Micro-90® cleaning solution, 1 quart bottle (YO-18100-01)
Category: Lab and General Purpose Cleaners
Description: Cole-Parmer
®
Micro-90
®
Cleaning Solution
Brand:
Micro-90
YO-18100-01
More Details
Cole-Parmer® Micro-90® cleaning solution, case of four 1 gallo (YO-18100-10)
Category: Lab and General Purpose Cleaners
Description: Cole-Parmer
®
Micro-90
®
Cleaning Solution
Brand:
Micro-90
YO-18100-10
More Details
Cole-Parmer Micro-90 cleaning solution, 5 gallon bottle
Category: Surface Cleaners
Description: Cole-Parmer Micro-90 Cleaning Solution
Brand:
Micro-90
YO-18100-20
More Details
Cole-Parmer Micro-90 cleaning solution, 55 gallon drum
Category: Surface Cleaners
Description: Cole-Parmer Micro-90 Cleaning Solution
Brand:
Micro-90
YO-18100-30
More Details
Cole-Parmer Micro-90 cleaning solution, case of twelve 1 liter bottles
Category: Surface Cleaners
Description: Cole-Parmer Micro-90 Cleaning Solution
Brand:
Micro-90
YO-18100-04
More Details
Cole-Parmer Micro-90 cleaning solution, 1 liter bottle
Category: Surface Cleaners
Description: Cole-Parmer Micro-90 Cleaning Solution
Brand:
Micro-90
YO-18100-05
More Details
Cole-Parmer Micro-90 cleaning solution, case of four 4 Liter bottles
Category: Surface Cleaners
Description: Cole-Parmer Micro-90 Cleaning Solution
Brand:
Micro-90
YO-18100-11
More Details
Detergent 8 Cleaning Detergent, case of four 1-gallon bottles
Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand:
Detergent 8
Model Number:
1701
YO-17775-10
More Details
Fast-response thermocouple microprobe with 90°-angle tip (BT-1)
Category: Oakton General-Purpose Probes
Description: Micro-Probe Fast-Response with 90 deg-angle tip
Brand:
Oakton
Model Number:
BT-1
YO-08506-95
More Details
AEMC 6250 : Micro-Ohmmeter, 10A 1SHOT Continuous Multi-Test Manual/Au
Category: Resistance Testers
Description: AEMC Digital 10A Micro Ohmmeter - Model 6250
Brand:
AEMC
Model Number:
6250
YO-20018-28
More Details
AEMC 6240 10-Amp Micro-Ohmmeter
Category: Resistance Testers
Description: AEMC 6240 Micro-Ohmmeter
Brand:
AEMC
Model Number:
6240
YO-27443-45
More Details
Labconco CentriVap Micro IR Vacuum Centrifugal Concentrator, 115V 60Hz
Category: Microcentrifuges
Description: Labconco CentriVap Micro IR Vacuum Centrifugal Concentrators
Brand:
Labconco
YO-13040-11
More Details
Labconco CentriVap Micro IR Vacuum Centrifugal Concentrator, 230v 50/6 (YO-13040-13)
Category: Microcentrifuges
Description: Labconco CentriVap Micro IR Vacuum Centrifugal Concentrators
Brand:
Labconco
YO-13040-13
More Details
Labconco CentriVap Micro IR Vacuum Centrifugal Concentrator, 230v 50/6 (YO-13040-15)
Category: Microcentrifuges
Description: Labconco CentriVap Micro IR Vacuum Centrifugal Concentrators
Brand:
Labconco
YO-13040-15
More Details
Labconco CentriVap Micro IR Vacuum Centrifugal Concentrator, 230v 50/6 (YO-13040-17)
Category: Microcentrifuges
Description: Labconco CentriVap Micro IR Vacuum Centrifugal Concentrators
Brand:
Labconco
YO-13040-17
More Details
Micro Aperture Stopper Red Rubber 24pk
Category: Stoppers
Description: Cole-Parmer Micro Aperture Stoppers, Rubber or Neoprene
Brand:
Cole-Parmer
YO-62995-01
More Details
Micro Aperture Stopper Green Neoprene 24pk
Category: Stoppers
Description: Cole-Parmer Micro Aperture Stoppers, Rubber or Neoprene
Brand:
Cole-Parmer
YO-62995-03
More Details
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 20 microL, 12 VDC (120SP1220-4EE)
Category: Diaphragm Pumps
Description: Cole-Parmer Self-Priming Micro Pumps
Brand:
Cole-Parmer
Model Number:
120SP1220-4EE
YO-73120-10
More Details
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 20 microL, 24 VDC (120SP2420-4EE)
Category: Diaphragm Pumps
Description: Cole-Parmer Self-Priming Micro Pumps
Brand:
Cole-Parmer
Model Number:
120SP2420-4EE
YO-73120-12
More Details
Micro 90 Detergent
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Concentration of a Solution
|
Next: Micro Cleaning Solution
>
31125329-s-content_2 31125329scontent2