Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
Fume Hood Monitors
Alnor Airflow Monitors
Monitor Hoods
Fume Hood Accessories
Welding Fume Hoods
Fuming Hood
Alnor Furniture & Fume Hoods
Furniture & Fume Hoods
Product Type Fume Hood Accessories
Cole-Parmer Formaldehyde
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Monitor 70
Image
Product Detail
SKU
Digital Fume Hood monitor, 70 to 250 ft/min (YO-33556-06)
Category: Airflow Monitors
Description: Fume Hood Airflow Monitors with Visual/Audible Alarm
Brand:
Alnor
YO-33556-06
More Details
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital (YO-08895-70)
Category: Standard Ultrasonic Cleaners
Description: Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater, Digital Timer, Temperature Monitor, and Degas Feature
Brand:
Cole-Parmer
YO-08895-70
More Details
Air Flow Monitor for Bio Safety Cabinets, 30 to 400 FPM Monitoring Ran
Category: Airflow Monitors
Description: Wide-range Airflow Monitors for Fume Hoods
Brand:
TEL UK
YO-33556-70
More Details
M5PID-Xwqv:Rpdyn03PIDLEL O2 Cio2 Wrecharge Batt Motorized Pump Data Lo (YO-10107-70)
Category: PID Gas Detection
Description: GasAlertMicro Five-Gas Detectors
Brand:
GasAlertMicro
YO-10107-70
More Details
Interface cable for monitor 08330-20; length, 20-ft (YO-23000-70)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-23000-70
More Details
Primary Formaldehyde Filters for 30" Self-Monitoring Ductless Fume Hoo (YO-33713-70)
Category: Fume Hood Air Filters
Description: Filters for 30-Inch Fume Hoods
Brand:
Cole-Parmer
YO-33713-70
More Details
GasAlertQuattro 3-Gas Detector, rechargeable, %LEL, O2, CO
Category: Multigas Gas Detection
Description: GasAlertQuattro Multigas Monitors
Brand:
GasAlertQuattro
YO-86418-70
More Details
NO2 Filter Badges, pack of 10
Category: Personal Monitor Badges
Description: Advantec NO2 Filter Badge
Brand:
Cole-Parmer
YO-06649-70
More Details
Monitor 70
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Tel Airflow Monitors
|
Next: Alnor Furniture & Fume Hoods
>
31125329-s-content_2 31125329scontent2