Potassium Chloride 40 Lb Bag

Image
  Product Detail
SKU

Platinum (IV) Potassium Chloride (Laboratory) (1g)

Category: Platina to Potash
Description: Platinum (IV) Potassium Chloride
Brand: Cole-Parmer


Platinum (IV) Potassium Chloride (Laboratory) (5g)

Category: Platina to Potash
Description: Platinum (IV) Potassium Chloride
Brand: Cole-Parmer


Potassium Chloride (Crystalline/USP/FCC/EP/BP/JP) (2.5kg)

Category: Potassium
Description: Potassium Chloride
Brand: Cole-Parmer


Potassium Chloride (Cryst./USP/FCC) (3kg) (YO-88079-05)

Category: Potassium
Description: Potassium Chloride
Brand: Cole-Parmer


Potassium Chloride (Crystalline/USP/FCC/EP/BP/JP) (12kg) (P33512)

Category: Potassium
Description: Potassium Chloride
Brand: Cole-Parmer


Potassium Chloride Rgt Acs 1kg (AC424090010)

Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand: Cole-Parmer


Potassium Chloride Reagent 25g (AC424090250)

Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand: Cole-Parmer


PBS, 1X Solution (20 liter)

Category: Phenyls to Phospho
Description: Phosphate Buffered Saline, 1X Solution
Brand: Cole-Parmer


PBS, 1X Solution (4 liter)

Category: Phenyls to Phospho
Description: Phosphate Buffered Saline, 1X Solution
Brand: Cole-Parmer


Tris Buffered Saline (TBS), 10X Solution (pH 7.4/Molecular Biology) (100mL)

Category: Tris
Description: Tris Buffered Saline (TBS), 10X Solution
Brand: Cole-Parmer


Tris Buffered Saline (TBS), 10X Solution (pH 7.4/Molecular Biology) (500mL)

Category: Tris
Description: Tris Buffered Saline (TBS), 10X Solution
Brand: Cole-Parmer


Iron (III) Chloride Hexahydrate (Lumps/Technical) (10kg)

Category: Iron to Isopent
Description: Iron (III) Chloride Hexahydrate
Brand: Cole-Parmer


Copper (I) Chloride (Certified ACS) (500g)

Category: Chemicals Ca to Cy
Description: Copper (I) Chloride
Brand: Cole-Parmer


Iron (II) Chloride Tetrahydrate (Crystalline/Certified) (500g)

Category: Iron to Isopent
Description: Iron (II) Chloride Tetrahydrate
Brand: Cole-Parmer


Nickel (II) Chloride Hexahydrate (Crystalline/Certified) (500g)

Category: Napthy to Nickel
Description: Nickel (II) Chloride Hexahydrate
Brand: Cole-Parmer


Nickel (II) Chloride Hexahydrate (Crystalline/Certified) (3kg)

Category: Napthy to Nickel
Description: Nickel (II) Chloride Hexahydrate
Brand: Cole-Parmer


Potassium Phosphate Monobasic (Crystalline/NF/FCC) (2.5kg)

Category: Potassium
Description: Potassium Phosphate Monobasic
Brand: Cole-Parmer


Potassium Hydrogen Phthalate (Primary Standard) 2 (500g)

Category: Potassium
Description: Potassium Hydrogen Phthalate
Brand: Cole-Parmer


Potassium Phosphate Monobasic (Crystalline/Primary Standard) (500g)

Category: Potassium
Description: Potassium Phosphate Monobasic
Brand: Cole-Parmer


Barium Chloride Dihydrate (Cryst./Technical) (10kg)

Category: Ba to Benzene
Description: Barium Chloride Dihydrate
Brand: Cole-Parmer




Potassium Chloride 40 Lb Bag
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Cole-Parmer Potassium Chloride  |  Next: Potassium Chloride