Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
Qsonica Q55 Sonicator Ultrasonic Processor
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Probes for Q55 Sonicator Ultrasonic Processor
Image
Product Detail
SKU
Dual Horn (without probes) for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-86
More Details
Temperature Probe for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-94
More Details
Thermocouple probe for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-95
More Details
Microprobe, 5/64", 0.2 - 5ml, for use with Q55 Sonicator
Category: Accessories for Sonicator Ultrasonic Processors
Description: Probes for Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-53
More Details
Microprobe, 1/8", 0.5 - 15ml, for use with Q55 Sonicator
Category: Accessories for Sonicator Ultrasonic Processors
Description: Probes for Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-54
More Details
Microprobe, 1/4", 10 - 50ml, for use with Q55 Sonicator
Category: Accessories for Sonicator Ultrasonic Processors
Description: Probes for Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-55
More Details
Coupler (for Probe 04712-65) for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-64
More Details
Dual Horn (with two 3/4" solid probes) for use with ultrasonic process
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-83
More Details
Probe, 1", solid
Category: Qsonica Ultrasonic Processors
Description: Probes & Tips for Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-71
More Details
Probe, 1/2", solid
Category: Qsonica Ultrasonic Processors
Description: Probes & Tips for Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-67
More Details
Probe, 3/4", solid
Category: Qsonica Ultrasonic Processors
Description: Probes & Tips for Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-69
More Details
Probes for Q55 Sonicator Ultrasonic Processor
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Laboratory Sonicators
|
Next: Qsonica Accessories for Sonicator Ultrasonic Processors
>
31125329-s-content_2 31125329scontent2