Related Categories

Shop All Products
 

Sodium Hydroxide Solution 50

Image
  Product Detail
SKU

Sodium Hydroxide Solution (50% w/w/Certified) (200L) (SS254200)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide Solution
Brand: Hydroxide


Sodium Hydroxide Solution (0.02N (N/50)/Certified) (4 liter) (SS2824)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide Solution
Brand: Hydroxide


Sodium Hydroxide Solution (0.2N (N/5)/Certified) (1 liter)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide Solution
Brand: Hydroxide


Sodium Hydroxide Solution (6.25N) (500mL) (SS263500)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide Solution
Brand: Hydroxide


Sodium Hydroxide Solution (1N/Certified) (20 liter) (YO-88085-25)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide Solution
Brand: Hydroxide


Sodium Hydroxide Solution (0.25N (N/4)/Certified) (20 liter) (SS27220)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide Solution
Brand: Hydroxide


Sodium Hydroxide (NF/EP/BP/FCC) (2.5kg) (S399212)

Category: Sodiumh to Sodiumo
Description: Sodium Hydroxide
Brand: Hydroxide


Sodium Hydroxide 50 Wt% 25ml (AC259860250)

Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand: Hydroxide


Cesium hydroxide, 50 wt% solution in water 99% (500g) (YO-88228-15)

Category: Carboxy to Cesium
Description: Cesium hydroxide
Brand: Hydroxide


Cesium hydroxide, 50 wt% solution in water 99% (100g) (YO-88228-14)

Category: Carboxy to Cesium
Description: Cesium hydroxide
Brand: Hydroxide


Sodium cyanoacetate, 50 wt% aqueous solution (1kg) (AC250290010)

Category: Sodium to Sodiumg
Description: Sodium cyanoacetate
Brand: Cole-Parmer


2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water (25ml)

Category: Pya to Pyruvic
Description: 2-Pyrrolidone-5-carboxylic acid, sodium salt
Brand: Cole-Parmer


2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water (100ml)

Category: Pya to Pyruvic
Description: 2-Pyrrolidone-5-carboxylic acid, sodium salt
Brand: Cole-Parmer




Sodium Hydroxide Solution 50
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Tampa Fl Sodium Hydroxide Caustic Soda  |  Next: 50 Sodium Hydroxide Membrane Grade