Related Categories

Shop All Products
 

Tube 12

Image
  Product Detail
SKU

Microbore - Puri-Flex ™ Autoanalysis Tubing, -12, 50 feet

Category: Masterflex Pump Tubing for Masterflex C/L Pumps
Description: Puri-Flex Autoanalysis/Microbore Tubing
Brand: Masterflex


Conical-Bottom Glass Centrifuge Tubes, 12 mL, with Graduations; 12/case

Category: Centrifuge Tubes
Description: Conical-Bottom Glass Centrifuge Tubes
Brand: Cole-Parmer


Conical-Bottom Glass Centrifuge Tubes, 12 mL, with Graduations, Pack of 1

Category: Centrifuge Tubes
Description: Conical-Bottom Glass Centrifuge Tubes
Brand: Cole-Parmer


Conical-Bottom Glass Centrifuge Tubes, 12 mL, without Graduations; 12/case

Category: Centrifuge Tubes
Description: Conical-Bottom Glass Centrifuge Tubes
Brand: Cole-Parmer


166-12 Stainless Steel Pitot Tube 12" Insertion Length 1/8" Diameter

Category: Manometers
Description: Dwyer Stainless Steel Pitot Tubes
Brand: Dwyer Instruments


Dwyer 167-12 Stainless Steel Pitot Tube; 12" L, Short-Tip, 1/8" Dia

Category: Manometers
Description: Dwyer Stainless Steel Pitot Tubes
Brand: Dwyer Instruments


160-12 Stainless Steel Pitot Tube 12" Insertion Length 5/16" Diameter

Category: Manometers
Description: Dwyer Stainless Steel Pitot Tubes
Brand: Dwyer Instruments


Dry Small Diameter Sampling Tube, 12" Length (PN007)

Category: Soil Samplers
Description: Sampling Tubes
Brand: Cole-Parmer


Wet/dry soil sampling tube 12" long (PN012)

Category: Soil Samplers
Description: Sampling Tubes
Brand: Cole-Parmer


Thermo Scientific Nunc Round-bottom tubes; 12-mL; PC; 10/pack

Category: Centrifuge Tubes
Description: Thermo Scientific Nunc Round-Bottom Tubes
Brand: Nunc


Thermo Scientific Nunc Round-bottom tubes; 12-mL; PPCO; 10/pack

Category: Centrifuge Tubes
Description: Thermo Scientific Nunc Round-Bottom Tubes
Brand: Thermo Scientific Nunc


Masterflex, Gore Style 400, Precision Size L/S 17 Tubing, 12 In

Category: L/S Precision Pump Tubing for Masterflex L/S Pumps
Description: L/S Precision GORE High-Resilience Style 400 Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Spectra/Por 5 Standard RC Dialysis Tubing, 12-14 kDalton, 75 mm, 50ft/bx

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 5 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Spectra/Por 5 Standard RC Dialysis Tubing, 12-14 kDalton, 140 mm, 50ft/bx

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 5 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft. (YO-96143-12)

Category: Rubber Tubing
Description: Black Rubber Vacuum Tubing
Brand: Vacuum tubing


Coiled FEP tubing, 1/8"ID x 1/4"OD, 2 ft long (46034)

Category: Coiled FEP Tubing
Description: Coiled FEP tubing, 12" relaxed
Brand: Cole-Parmer


Coiled FEP tubing, 1/4"ID x 3/8"OD, 2 ft long (49126)

Category: Coiled FEP Tubing
Description: Coiled FEP tubing, 12" relaxed
Brand: Cole-Parmer


Coiled FEP tubing, 3/8"ID x 1/2"OD, 2 ft long (49125)

Category: Coiled FEP Tubing
Description: Coiled FEP tubing, 12" relaxed
Brand: Cole-Parmer


Thermo Scientific Heating block for 15 or 16 mm tubes, 12 openings

Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Heater, Blocks
Brand: Thermo Scientific


Thermo Scientific Heating block for 17 or 18 mm tubes, 12 openings

Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Heater, Blocks
Brand: Thermo Scientific