Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
Masterflex Tygon Tubing
Tygon Masterflex
Tygon R 3603
Tygon E - 3603 Lab Tubing
Tygon E-3603 Non-Dehp Vacuum Tubing
Tubing Formulation Tygon E-Lab (E-3603)
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Tygon Tubing R 3603
Image
Product Detail
SKU
Masterflex® Tygon® Lab tubing (R-3603), I/P® 73, 50 ft. (YO-06408-73)
Category: I/P® Precision Tubing for Masterflex® I/P® Pumps
Description: I/P® Precision Tygon® Lab (R-3603) Tubing for Masterflex® I/P® Pumps
Brand:
Masterflex
YO-06408-73
More Details
Masterflex Tygon® Lab tubing (R-3603), I/P® 82, 50 ft. (YO-06408-82)
Category: I/P® Precision Tubing for Masterflex® I/P® Pumps
Description: I/P® Precision Tygon® Lab (R-3603) Tubing for Masterflex® I/P® Pumps
Brand:
Masterflex
YO-06408-82
More Details
Masterflex® Tygon® Lab tubing (R-3603), L/S® 15, 50 ft. (YO-06409-15)
Category: L/S® High-Performance Precision Tubing for Masterflex® L/S® Pumps
Description: L/S® High-Performance Precision Tygon® Lab (R-3603) Tubing for Masterflex® L/S® Pumps
Brand:
Masterflex
YO-06409-15
More Details
Masterflex® Tygon® Lab tubing (R-3603), L/S® 24, 50 ft. (YO-06409-24)
Category: L/S® High-Performance Precision Tubing for Masterflex® L/S® Pumps
Description: L/S® High-Performance Precision Tygon® Lab (R-3603) Tubing for Masterflex® L/S® Pumps
Brand:
Masterflex
YO-06409-24
More Details
Masterflex® Tygon® Lab tubing (R-3603), I/P® 26, 50 ft. (YO-06409-26)
Category: I/P® Precision Tubing for Masterflex® I/P® Pumps
Description: I/P® Precision Tygon® Lab (R-3603) Tubing for Masterflex® I/P® Pumps
Brand:
Masterflex
YO-06409-26
More Details
Masterflex® Tygon® Lab tubing (R-3603), L/S® 35, 50 ft. (YO-06409-35)
Category: L/S® High-Performance Precision Tubing for Masterflex® L/S® Pumps
Description: L/S® High-Performance Precision Tygon® Lab (R-3603) Tubing for Masterflex® L/S® Pumps
Brand:
Masterflex
YO-06409-35
More Details
Masterflex® Tygon® Lab tubing (R-3603), L/S® 36, 50 ft. (YO-06409-36)
Category: L/S® High-Performance Precision Tubing for Masterflex® L/S® Pumps
Description: L/S® High-Performance Precision Tygon® Lab (R-3603) Tubing for Masterflex® L/S® Pumps
Brand:
Masterflex
YO-06409-36
More Details
TYGON LAB TUBING, Non-DEHP, 1/32"ID X 3/32"OD, 50 FT/PACK (YO-07407-70)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-70
More Details
TYGON LAB TUBING, Non-DEHP, 1/16"ID X 1/8"OD, 50 FT/PACK (YO-07407-71)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-71
More Details
Tygon lab tubing, Non-DEHP, 1/16"ID x 3/16"OD, 50 ft/pack (ACF00003-CP)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
Model Number:
ACF00003-CP
YO-07407-72
More Details
TYGON LAB TUBING, Non-DEHP, 3/32"ID X 5/32"OD, 50 FT/PACK (YO-07407-73)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-73
More Details
TYGON LAB TUBING, Non-DEHP, 3/32"ID X 7/32"OD, 50 FT/PACK (YO-07407-74)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-74
More Details
TYGON LAB TUBING, Non-DEHP, 1/8"ID X 3/16"OD, 50 FT/PACK (YO-07407-75)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-75
More Details
Tygon lab tubing, Non-DEHP, 1/8"ID x 1/4"OD, 50 ft/pack (YO-07407-76)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-76
More Details
TYGON LAB TUBING, Non-DEHP, 5/32"ID X 7/32"OD, 50 FT/PACK (YO-07407-77)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-77
More Details
Tygon lab tubing, Non-DEHP, 3/16"ID x 5/16"OD, 50 ft/pack (YO-07407-78)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-78
More Details
Tygon lab tubing, Non-DEHP, 3/16"ID x 3/8"OD, 50 ft/pack (YO-07407-79)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-79
More Details
TYGON LAB TUBING, Non-DEHP, 1/4"ID X 3/8"OD, 50 FT/PACK (YO-07407-80)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-80
More Details
TYGON LAB TUBING, Non-DEHP, 1/4"ID X 7/16"OD, 50 FT/PACK (YO-07407-81)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-81
More Details
TYGON LAB TUBING, Non-DEHP, 1/4"ID X 1/2"OD, 50 FT/PACK (YO-07407-82)
Category: TYGON Lab Tubing
Description: Tygon E - 3603 Lab Tubing
Brand:
Tygon
YO-07407-82
More Details
Tygon Tubing R 3603
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Tygon Lab Bulk Packs
|
Next: Tygon R 3603
>
31125329-s-content_2 31125329scontent2