Whitey 1 8 Needle Valves

Image
  Product Detail
SKU

Chemically Inert Panel-Mount Needle Valve, 1/8" NPT(M) (MV-13-163)

Category: Needle Valves
Description: PFA Needle Valves
Brand: Cole-Parmer


Chemically Inert Panel-Mount Needle Valve, 1/8" tube OD (MV-13-163)

Category: Needle Valves
Description: PFA Needle Valves
Brand: Cole-Parmer


Cole-Parmer Multi-Turn Aluminum Needle Valve, 1/8" NPT(F), 2800 mL/min water

Category: Needle Valves
Description: Cole-Parmer Multi-Turn Needle Valves
Brand: Cole-Parmer


Cole-Parmer Multi-Turn Aluminum Needle Valve, 1/8" NPT(F), 130 mL/min water

Category: Needle Valves
Description: Cole-Parmer Multi-Turn Needle Valves
Brand: Cole-Parmer


Cole-Parmer Multi-Turn 316 SS Needle Valve, 1/8" NPT(F), 130 mL/min water

Category: Needle Valves
Description: Cole-Parmer Multi-Turn Needle Valves
Brand: Cole-Parmer


Cole-Parmer Multi-Turn 316 SS Needle Valve, 1/8" NPT(F), 2800 mL/min water

Category: Needle Valves
Description: Cole-Parmer Multi-Turn Needle Valves
Brand: Cole-Parmer


Needle Valve, Hard Seat ss Female-Female 3/8 in.NPT Female-Female ss Valve

Category: Noshok Precision Needle Valves
Description: Noshok 400 Series Precision Needle Valves
Brand: Noshok


Needle Valve, Soft Seat ss Male-Female 3/8 in.NPT Female-Female ss Valve

Category: Noshok Precision Needle Valves
Description: Noshok 500 Series Precision Needle Valves
Brand: Noshok


Needle Valve, Hard Seat ss Female-Female 1 1/4 in.NPT Female-Female ss Valve

Category: Noshok Precision Needle Valves
Description: Noshok 400 Series Precision Needle Valves
Brand: Noshok


Needle Valve, Hard Seat ss Male-Female 1 1/4 in. NPT Male-Female

Category: Noshok Precision Needle Valves
Description: Noshok 400 Series Precision Needle Valves
Brand: Noshok


Needle Valve, Hard Seat ss Female-Female 1 1/2 in.NPT Female-Female ss Valve

Category: Noshok Precision Needle Valves
Description: Noshok 400 Series Precision Needle Valves
Brand: Noshok


Needle Valve, Hard Seat ss Male-Female 1 1/2 in. NPT Male-Female

Category: Noshok Precision Needle Valves
Description: Noshok 400 Series Precision Needle Valves
Brand: Noshok


Chemically Inert Panel-Mount Needle Valve, 3/8" NPT(M) (MV-13-163)

Category: Needle Valves
Description: PFA Needle Valves
Brand: Cole-Parmer


PVDF Needle Valve, 3/8" NPT(F) (155.522.554)

Category: Needle Valves
Description: Plastic Needle Valves
Brand: Cole-Parmer


PVC Needle Valve, 3/8" NPT(F) (155.522.554)

Category: Needle Valves
Description: Plastic Needle Valves
Brand: Cole-Parmer


PP Needle Valve, 3/8" NPT(F) (NV40025T)

Category: Needle Valves
Description: Plastic Needle Valves
Brand: Cole-Parmer


Chemically Inert Panel-Mount Needle Valve, 3/8" tube OD (MV-13-163)

Category: Needle Valves
Description: PFA Needle Valves
Brand: Cole-Parmer


Cole-Parmer Multi-Turn Needle Valve, Aluminum; 3/8" ID, 130mL/min (l)

Category: Needle Valves
Description: Cole-Parmer Multi-Turn Needle Valves
Brand: Cole-Parmer


Cole-Parmer Multi-Turn Needle Valve, Aluminum; 3/8" ID, 2800 mL/min(l)

Category: Needle Valves
Description: Cole-Parmer Multi-Turn Needle Valves
Brand: Cole-Parmer




Whitey 1 8 Needle Valves
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended |  Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended

Previous: Needle Valves  |  Next: Weatherford 1 8 Inch Needle Valve