Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
A D Ethernet Option for Phoenix Series GH Balances
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-11111-39
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances / Scales Accessories
Share
|
A&D Ethernet Option for Phoenix Series GH Balances
YO-11111-39
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Technical Resources
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Related Searches for A&D Ethernet Option for Phoenix Series GH Balances
Denver Instrument Summit Series Toploading Balances
Denver Instrument Maxx Series Portable Balances
Denver Instrument Summit Series Analytical Balances
Mettler Toledo Excellence Xs-Series Toploading Balances
Phoenix Balances & Scales
A&D Weighing Phoenix Gh Analytical Balances
Weighing Calibration
Series Summit
Scales Mettler
Mettler Toledo Products
Related Products for A&D Ethernet Option for Phoenix Series GH Balances
Image
Product Detail
SKU
A&D Phoenix Series Analytical Balance 120g x 0.1mgwith Internal Calibr
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Phoenix GH Analytical Balances
Brand:
A&D Weighing
Model Number:
GH-120
YO-11111-34
More Details
A&D Phoenix Series Analytical Balance 220g x 0.1mg with Internal Calib
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Phoenix GH Analytical Balances
Brand:
A&D Weighing
Model Number:
GH-200
YO-11111-35
More Details
A&D Phoenix Series Analytical Balance 320g x 0.1mg with Internal Calib
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Phoenix GH Analytical Balances
Brand:
A&D Weighing
Model Number:
GH-300
YO-11111-36
More Details
A&D Phoenix Series Analytical Balance 220/51g x 0.1/0.01mg with Intern
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Phoenix GH Analytical Balances
Brand:
A&D Weighing
Model Number:
GH-202
YO-11111-37
More Details
A&D Phoenix Series Analytical Balance 250/101g x0.1/0.01mg with Intern
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Phoenix GH Analytical Balances
Brand:
A&D Weighing
Model Number:
GH-252
YO-11111-38
More Details
Summit Series Toploading Balance 4000G X 0.01G 230 VAC - EXTERNAL CALI (S-4002.3)
Category: Denver Instrument® Toploading Balances
Description: Denver Instrument Summit Series Toploading Balances
Brand:
Denver Instrument
Model Number:
S-4002.3
YO-11421-68
More Details
Summit Series Toploading Balance 600G X 0.001G 230 VAC - EXTERNAL CALI (S-603.3)
Category: Denver Instrument® Toploading Balances
Description: Denver Instrument Summit Series Toploading Balances
Brand:
Denver Instrument
Model Number:
S-603.3
YO-11421-70
More Details
Summit Series Toploading Balance 200G X 0.001G 230 VAC - EXTERNAL CALI (S-203.3)
Category: Denver Instrument® Toploading Balances
Description: Denver Instrument Summit Series Toploading Balances
Brand:
Denver Instrument
Model Number:
S-203.3
YO-11421-72
More Details
Summit Series Toploading Balance, 400 g x 0.001 g, 230 VAC (S403.3)
Category: Denver Instrument® Toploading Balances
Description: Denver Instrument Summit Series Toploading Balances
Brand:
Denver Instrument
Model Number:
S403.3
YO-11421-86
More Details
Summit Series Toploading Balance, 2000 g x 0.01 g, 230 VAC (S2002.3)
Category: Denver Instrument® Toploading Balances
Description: Denver Instrument Summit Series Toploading Balances
Brand:
Denver Instrument
Model Number:
S2002.3
YO-11421-92
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
A&D Ethernet Option for Phoenix Series GH Balances
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: A&D Engineering Washdown Bench Scale;Capacity 10 kg x 0.001 g, 120 VAC
|
Next: A&D FC-i Counting Scale, 10,000g capacity
31125329-p-content_2 31125329pcontent2