Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-78226-72
Stainless steel cannula, 13 gauge; for use with Luer lock dispensing tip. Autoclavable.
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-78226-72
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Stainless steel cannula, 13 gauge; for use with Luer lock dispensing t
YO-78226-72
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Model
91-015-220
Related Searches for Stainless steel cannula, 13 gauge; for use with Luer lock dispensing t
Weksler Stainless Steel Case Contractor Gauges
Stainless Steel Funnel Rain Gauge
304 Stainless Steel Tubing 25 Gauge
Stainless Steel Funnels in Dallas Tx
Stainless Steel Funnels in Tampa Bay
Mini Stainless Steel Funnel
Stainless Steel Funnels Search
Stainless Steel Funnel with Detachable Strainer
Oval Stainless Steel Funnels
Durham Nc Stainless Steel Funnel
Related Products for Stainless steel cannula, 13 gauge; for use with Luer lock dispensing t
Image
Product Detail
SKU
Cole-parmer Stainless Steel Syringe, 20 mL, Viton O-ring, Luer Lock
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Stainless Steel Syringes
Brand:
Cole-Parmer
YO-74044-26
More Details
Cole-parmer Stainless Steel Syringe, 50 mL, Viton O-ring, Luer Lock
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Stainless Steel Syringes
Brand:
Cole-Parmer
YO-74044-36
More Details
Cole-parmer Stainless Steel Syringe, 100 mL, Viton O-ring, Luer Lock
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Stainless Steel Syringes
Brand:
Cole-Parmer
YO-74044-46
More Details
Cole-parmer Stainless Steel Syringe, 200 mL, Viton O-ring, Luer Lock
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Stainless Steel Syringes
Brand:
Cole-Parmer
YO-74044-56
More Details
Stainless steel funnel, 13 oz (YO-07265-00)
Category: Utility Funnels
Description: Stainless Steel Funnels
Brand:
Guaranteed In Stock Program
YO-07265-00
More Details
Protective Rubber Boot For Wika Stainless Steel Digital Pressure Gauge
Category: Digital Pressure Gauges and Meters
Description: WIKA Stainless Steel Digital Gauges
Brand:
Wika
Model Number:
DG-10
YO-68052-90
More Details
Push-to-connect male branch tee, 316 stainless steel, 1/4" x 1/4" NPTM (YO-31370-13)
Category: Stainless Steel Push-to-Connect Fittings
Description: Stainless steel Push-to-Connect Male Branch Tees
Brand:
Cole-Parmer
YO-31370-13
More Details
Receiving Pan for 8" Diameter Stainless Steel Full Height Sieves
Category: Sieve Covers and Receiving Pans for Stainless Steel Sieves
Description: Receiving Pans for 8" Stainless Steel Bottom Sieves
Brand:
Cole-Parmer
YO-59986-90
More Details
Receiving Pan for 8" Diameter Stainless Steel Half Height Sieves
Category: Sieve Covers and Receiving Pans for Stainless Steel Sieves
Description: Receiving Pans for 8" Stainless Steel Bottom Sieves
Brand:
Cole-Parmer
YO-59986-91
More Details
Receiving Pan with Fitted Rim for Nesting 8" Stainless Steel Sieves, F
Category: Sieve Covers and Receiving Pans for Stainless Steel Sieves
Description: Receiving Pans with Fitted Rim for Nesting 8" Stainless Steel Sieves
Brand:
Cole-Parmer
YO-59986-92
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Stainless steel cannula, 13 gauge; for use with Luer lock dispensing t
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Stainless Steel Bucket Stand with four 2" Swivel Casters.
|
Next: Stainless steel chopping blade (YO-04300-22)
31125329-p-content_2 31125329pcontent2