Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Flourescent FLT Yellow Green Dye Concentrate Gallon
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-00298-07
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Dye Tracers
Share
|
Flourescent FLT Yellow/Green Dye Concentrate, Gallon
YO-00298-07
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Determine water flow in closed circuits or open channels
Biodegradable and nontoxic
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
Specifications
Product Type
Dye Tracers
Gallons of water for 1 ppm
100,000
Applications
Metered systems
Manufacturer number
106501G
Technical Resources
00298-17MSDS.pdf
MSDS 1 (135 B)
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Flourescent FLT Yellow/Green Dye Concentrate, Gallon
Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Fluorescent Yellow Green Dye Tracers
Green Dye
Concentrate Gallon
Pint Concentrate
Dye Tablet Tracers
Fluorescent Dye Water Tracer
Tracer Dye
Rhodamine Wt Dye
Cole-Parmer Dye Tracers
Related Products for Flourescent FLT Yellow/Green Dye Concentrate, Gallon
Image
Product Detail
SKU
Flourescent FLT Yellow/Green Dye Concentrate, Pint
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Brand:
Cole-Parmer
YO-00298-17
More Details
Flourescent FLT Yellow/Green Dye Tablets, 200 tablets
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Brand:
Cole-Parmer
YO-00295-17
More Details
Flourescent FLT Yellow/Green Dye Powder, 1 Lb
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Brand:
Cole-Parmer
YO-00298-27
More Details
Flourescent FLT Orange Dye Concentrate, Gallon
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand:
Cole-Parmer
YO-00298-09
More Details
Flourescent FWT Red Dye Concentrate, Gallon
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Red Dye Tracers
Brand:
Cole-Parmer
YO-00298-06
More Details
Blue Dye Concentrate, Gallon
Category: Dye Tracers
Description: Cole-Parmer Blue Dye Tracers
Brand:
Cole-Parmer
YO-00298-08
More Details
Flourescent FLT Orange Dye Concentrate, Pint
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand:
Cole-Parmer
YO-00298-19
More Details
Flourescent FLT Orange Dye Tablets, 200 tablets
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand:
Cole-Parmer
YO-00295-19
More Details
Flourescent FWT Red Dye Concentrate, Pint
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Red Dye Tracers
Brand:
Cole-Parmer
YO-00298-16
More Details
Flourescent FLT Orange Dye Powder, 1 Lb
Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand:
Cole-Parmer
YO-00298-29
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Flourescent FLT Yellow/Green Dye Concentrate, Gallon
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Flourescent FLT Orange Dye Tablets, 200 tablets
|
Next: Flourescent FLT Yellow/Green Dye Concentrate, Pint
31125329-p-content_2 31125329pcontent2