Fluorescent FLT Yellow/Green Dye Concentrate, Gallon

YO-00298-07
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
Product Type Dye Tracers
Gallons of water for 1 ppm 100,000
Applications Metered systems
Manufacturer number 106501G





Technical Resources
 
Related Products for Fluorescent FLT Yellow/Green Dye Concentrate, Gallon
Image
  Product Detail
SKU

Fluorescent FLT Yellow/Green Dye Concentrate, Pint

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Brand: Cole-Parmer


Fluorescent FLT Yellow/Green Dye Tablets, 200 tablets

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Brand: Cole-Parmer


Fluorescent FLT Yellow/Green Dye Powder, 1 Lb

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Yellow/Green Dye Tracers
Brand: Cole-Parmer


Fluorescent FLT Orange Dye Concentrate, Gallon

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand: Cole-Parmer


Fluorescent FWT Red Dye Concentrate, Gallon

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Red Dye Tracers
Brand: Cole-Parmer


Blue Dye Concentrate, Gallon

Category: Dye Tracers
Description: Cole-Parmer Blue Dye Tracers
Brand: Cole-Parmer


Fluorescent FLT Orange Dye Concentrate, Pint

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand: Cole-Parmer


Fluorescent FLT Orange Dye Tablets, 200 tablets

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand: Cole-Parmer


Fluorescent FWT Red Dye Concentrate, Pint

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Red Dye Tracers
Brand: Cole-Parmer


Fluorescent FLT Orange Dye Powder, 1 Lb

Category: Dye Tracers
Description: Cole-Parmer Fluorescent Orange Dye Tracers
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

23 Questions | 24 Answers
Displaying questions 1-10Previous | Next »
  • Q:

    What concentration is this solution (weight per volume g/ml)?
    Asked on 1/29/2014 by nsmith

    1 answer

    • STAFF EXPERT

      A:

      The concentration of the active ingredient is 7%.

      Answered on 1/29/2014 by Jack from Technical Support
  • Q:

    What wavelength should be used for detection of the fluorescent red dye?
    Asked on 8/12/2013 by Lara

    1 answer

    • STAFF EXPERT

      A:

      This part number is for the yellow/green color, but any of these fluorescent dyes should be detectable using 365 nm UV light.

      Answered on 8/12/2013 by Jack from Technical Support
  • Q:

    Which dye would you recommend for SOIL water tracing of acid yellow (lissamine FF) rhodamine WT and fluorescein. I am concerned about adsorption on the particles.
    Asked on 6/3/2013 by none from UK

    1 answer

    • STAFF EXPERT

      A:

      The dyes will pass through soil, so it is recommended.

      Answered on 6/4/2013 by Dan from Technical Support
  • Q:

    can we use flouroscein with any biodiesel.i am really interested in testing the dye on waste vegetable oil biodiesel and a crude biodiesel.
    Asked on 11/18/2011 by sid from birmingham, united kingdom

    2 answers

    • A:

      Laser-induced fluorescence screening tools "see" the natural innate fluorescence of the biodiesel materials you are hoping to "label" with dye... so make sure you even need to label them first before you worry about dyes.

      Answered on 12/8/2011 by Randy from Fargo, ND USA
    • STAFF EXPERT

      A:

      No adverse reaction will occur using water based Item 00298-07, with any biodiesel, but it may not mix well with biodiesel products, therefore test results are not 100% accurate.

      Answered on 11/21/2011 by Rita from Customer Service
  • Q:

    I have my yellow/green dye. Now I need the right equipment to see it. The question of wavelength was already answered but what about intensity of the light source? Is the dye going to be perceptible by naked eye or is there eye wear needed?
    Asked on 10/25/2011 by neil from pennsylvania

    1 answer

    • Staff Reviewer

      A:

      The limit of quantification for this dye is 1 pint in 98 gallons of water (~0.1276%). The optimal wavelength is between 490 - 520 nm. 490-520 nm is not in the UV light range, it is in the white or visual light range and would be blue or green in color. So, a normal light bulb used in the home or office (or just outside) would cause this to fluoresce. That is why limit of detections are listed in ppm (parts per million and percentages).
      Please call for further assistance.

      Answered on 10/26/2011 by Jen from Technical Support
  • Q:

    I am trying to measure the depth of steam penetration in polystyrene molded forms and need a compound that can be carried by or vaporized with the steam. Will this dye work?
    Asked on 10/19/2011 by Neil from Pennsylvania

    1 answer

    • Staff Reviewer

      A:

      This dye will not stand up to that kind of heat, however the red dye will. Please see part number 00298-06 for the red dye concentrate and NOTE: the red dye is not NSF approved so it cannot come into contact with potable water (water intended to be used for human consumption). So, as long as you are using it in a closed loop system and then disposing of it like waste, this is fine.

      Answered on 10/21/2011 by Jen from Technical Support
  • Q:

    what is the Limit of quantitation for this dye under UV light?
    and what wavelength light should be used?
    Asked on 9/14/2011 by Mitesh from Rensselaer, NY

    1 answer

    • STAFF EXPERT

      A:

      The limit of quantification for this dye is 1 pint in 98 gallons of water (~0.1276%). The optimal wavelength is between 490 - 520 nm.

      Answered on 9/15/2011 by James from Technical Support
  • Q:

    How many ppm would be necessary for a mid-strong visual detection in silty river water?
    Asked on 7/26/2011 by Zack from Kansas City, MO

    1 answer

    • STAFF EXPERT

      A:

      The cutoff for strong visual detection in relatively clear water is 10 ppm, so the mid-strong detection you desire will be somewhere near 10ppm in river water.

      Answered on 7/28/2011 by Jack from Technical Support
  • Q:

    Is the dye neutrally buoyant? What is the refractive index? Thank you.
    Asked on 7/5/2011 by TN from Seattle, wa

    1 answer

    • STAFF EXPERT

      A:

      Yes, the dye has a neutral bouyancy. The refractive index has not been tested as it will be different in every application based on the volume of your sample and the amount of dye used.

      Answered on 7/7/2011 by Mike from Tech Support
  • Q:

    If we immerse a porous rock into water containing tracer for few hours, pull it out and cut it in half, can we use a UV light to figure if the dye would have enter the pore structure and reach the center of the rock
    Asked on 6/30/2011 by Pete from UK

    1 answer

    • Staff Reviewer

      A:

      I see no reason why this would not work.

      Answered on 7/5/2011 by Jen from Technical Support
Displaying questions 1-10Previous | Next »

Do you have a question about this product? 

Product Reviews
Customers Who Bought This Item Also Bought

Fluorescent FLT Yellow/Green Dye Concentrate, Gallon
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Fluorescent FLT Orange Dye Tablets, 200 tablets  |  Next: Fluorescent FLT Yellow/Green Dye Concentrate, Pint