3M KCL Fill Solution, 250 mL

YO-58905-82
Loading price...

Usually ships in 14 days or sooner.
Qty





Technical Resources
 
Related Products for 3M KCL Fill Solution, 250 mL
Image
  Product Detail
SKU

Oakton reference fill solution, 3.8 M KCl with AgCl, two 125-mL bottles/pk

Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton Reference Fill Solutions
Brand: Oakton


Oakton reference fill solution, 4 M KCl, two 125-mL bottles/pk

Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton Reference Fill Solutions
Brand: Oakton


Electrode fill solution, 4 M KCl with Ag+. Case of five 60-mL bottles (900011)

Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion Reference Fill Solutions
Brand: Thermo Scientific Orion


ROSS electrode fill solution, 3 M KCl. Case of five 60-mL bottles (810007)

Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion Reference Fill Solutions
Brand: Thermo Scientific Orion


Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bottles

Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion Reference Fill Solutions
Brand: Thermo Scientific Orion


Azlon solvent venting wash bottle for acetone, 250 mL

Category: Solvent Venting Wash Bottles
Description: Azlon Solvent Venting Wash Bottles, 250-mL
Brand: Dynalon


Azlon solvent venting wash bottle for ethanol, 250 mL

Category: Solvent Venting Wash Bottles
Description: Azlon Solvent Venting Wash Bottles, 250-mL
Brand: Dynalon


Azlon solvent venting wash bottle for isopropanol, 250 mL

Category: Solvent Venting Wash Bottles
Description: Azlon Solvent Venting Wash Bottles, 250-mL
Brand: Dynalon


Azlon solvent venting wash bottle for methanol, 250 mL

Category: Solvent Venting Wash Bottles
Description: Azlon Solvent Venting Wash Bottles, 250-mL
Brand: Dynalon


Azlon solvent venting wash bottle assortment pack, 250 mL

Category: Solvent Venting Wash Bottles
Description: Azlon Solvent Venting Wash Bottles, 250 mL Assortment Pack
Brand: Dynalon


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

2 Questions | 2 Answers
Displaying questions 1-2
  • Q:

    Is this product the same as Corning 477006?
    Asked on 1/2/2014 by Student

    1 answer

    • STAFF EXPERT

      A:

      The 477006 is discontinued. It is replaced with single 250 ml bottles. New part number
      58905-82, only change is in packaging size.

      Answered on 1/3/2014 by Dan from Technical Support
  • Q:

    Dear,

    I would like to know the compounds (substance) inside the corning 3M KCl nr 477006. Let me know.
    Thank you
    Asked on 9/26/2013 by Aquidauana from Brasil

    1 answer

    • STAFF EXPERT

      A:

      The electrolyte solution is 3M KCl (22% KCl salt dissolved in DI water).

      Answered on 9/27/2013 by James from Technical Support
Displaying questions 1-2

Do you have a question about this product? 

Product Reviews