8-in-1 Lighted Screwdriver

YO-97100-09
Loading price...

Call for availability.
Qty

  • Ideal for industrial, laboratory, electronics, and residential applications
  • Features two drivers in one handle: full-size and miniature
  • Two bright LED lights provide 360° tip illumination
  • Transparent collar maximizes illumination
  • Ergonomic cushioned grip
  • Bit storage in handle

What's Included:
Four double-ended bits (No. 0, No. 000, 1/8”, 5/64”, T5, T8, T6, T10) and three LR41 batteries.
Screwdriver style 8 in 1
Product Type Screwdriver
Bits per Set 4
Brand General
Model 75108
Manufacturer number 75108





 
Related Products for 8-in-1 Lighted Screwdriver
Image
  Product Detail
SKU

Cordless Lighted Screwdriver

Category: Screwdrivers
Description: General Tools Cordless Lighted Screwdriver
Brand: General


1',3',3'-Trimethylspiro-8-nitro(2H-1-benzopyran-2',2'-indoline) 99% (1g)

Category: Trimeth
Description: 1',3',3'-Trimethylspiro-8-nitro(2H-1-benzopyran-2',2'-indoline)
Brand: Cole-Parmer


Disposable filler nozzle, SS needle and polycarbonate base, 1/8 (YO-30619-07)

Category: Masterflex Tubing and Pump Accessories
Description: Overlook Industries OneShot Single-Use Filler Nozzles
Brand: Overlook Industries


Cole-Parmer Manifold Mixing Solenoid Valve, PTFE; 8 In/1 Out; 12 VDC

Category: Solenoid Valves
Description: Cole-Parmer Manifold Mixing Solenoid Valves
Brand: Cole-Parmer


Cole-Parmer Manifold Mixing Solenoid Valve, PEEK; 8 In/1 Out; 12 VDC

Category: Cole-Parmer Manifold Mixing Solenoid Valves
Description: Cole-Parmer Manifold Mixing Solenoid Valves with PEEK body
Brand: Cole-Parmer


Cole-Parmer Manifold Mixing Solenoid Valve, PTFE; 8 In/1 Out; 24 VDC

Category: Solenoid Valves
Description: Cole-Parmer Manifold Mixing Solenoid Valves
Brand: Cole-Parmer


Cole-Parmer Manifold Mixing Solenoid Valve, PEEK; 8 In/1 Out; 24 VDC

Category: Cole-Parmer Manifold Mixing Solenoid Valves
Description: Cole-Parmer Manifold Mixing Solenoid Valves with PEEK body
Brand: Cole-Parmer


Silicone rubber insulated heating tape, 8' x 1", 416 watts, 120 VAC (TBSO-101-080)

Category: Silicone Rubber Heating Tapes
Description: Silicone Rubber Heating Tapes - 120 VAC
Brand: BriskHeat


Silicone rubber insulated heating tape, 8' x 1", 416 watts, 240 VAC (TBSO-102-080)

Category: Silicone Rubber Heating Tapes
Description: Silicone Rubber Heating Tapes - 240 VAC
Brand: BriskHeat


Grounded fiberglass insulated heat tape, 8' x 1", 400 watts, 115 VAC

Category: Grounded Heating Tapes
Description: Grounded Heating Tapes - 115 VAC
Brand: BriskHeat


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews