Home  >  Colorimeters >
  

Hach DR 900 COLORIMETER

YO-99573-10
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Select from 90 preprogrammed tests for the most common parameters
  • "Favorites" list for easy method selection
  • Intuitive user interface allows quick selection and easy testing in 22 languages
  • Easy USB data transfer for up to 500 data sets
  • Rugged, waterproof housing is IP67 rated and permits on-site testing
The handheld DR 900 colorimeter allows quick and easy access to the most common water testing methods. Intuitive userinterface provides helpfulprompts in user-selectable languages. "Favorite" list saves your most commonly used methods for quick access.

Colorimeter features automatic wavelength selection, advanced reagent blank correction, and calibration curve adjustment. Analysis can be performed with user-set or preset reaction times with built-in timer. Large, backlit display reads results as concentration, absorbance, or percent transmittance.

Store up to 500 data sets with sample ID and user ID. Easy USB data transfer with no software or accessories needed; data simply transfers as .cvs file.

Durable over-molded housing is waterproof, dustproof, and shockproof; ideal for demanding field environments.
What's Included:
two 25-mL glass vials, two 10-mL plastic vials, COD vial adapter, USB cable, and four AA batteries.
Product Type Colorimeter
Test parameters 90 preprogrammed methods
Display LED
CE Compliance Yes
Wavelengths 420, 520, 560, 610 nm
Output USB
Waterproof Yes
Datalogging 500
Dimensions 9 1/8" x 3 13/16" x 1 7/8"
Range (Absorbance) 0 to 2 A
Power four AA batteries included

Accessories
Qty/Price
Availability

PACKAGE of 4

Usually ships in 14 days or sooner.

EACH

Call for availability.
Hard-sided case
YO-99573-53

EACH

Call for availability.
STD USB Cable
YO-99573-58

EACH

Call for availability.

EACH

Call for availability.
Replacement Cap
YO-99573-62

EACH

Call for availability.




Technical Resources
 
Related Products for Hach DR 900 COLORIMETER
Image
  Product Detail
SKU

Hach DR/870 Colorimeter

Category: Colorimeters
Description: Hach DR/870 Colorimeter
Brand: Hach


STD USB Cable

Category: Colorimeters
Description: Hach DR 900 Colorimeter Accessories
Brand: HACH


Replacement Battery Cover

Category: Colorimeters
Description: Hach DR 900 Colorimeter Accessories
Brand: HACH


Replacement Cap

Category: Colorimeters
Description: Hach DR 900 Colorimeter Accessories
Brand: HACH


Hach Pocket Colorimeter II, TPH in Soil Kit

Category: Colorimeters
Description: Hach Pocket Colorimeter II Soil Kits
Brand: HACH


Hach Pocket Colorimeter II, PCB in Soil Kit

Category: Colorimeters
Description: Hach Pocket Colorimeter II Soil Kits
Brand: HACH


Hach Pocket Colorimeter II, Aluminum

Category: Colorimeters
Description: Hach Pocket Colorimeter II Kits
Brand: HACH


Hach Pocket Colorimeter II, Ammonia

Category: Colorimeters
Description: Hach Pocket Colorimeter II Kits
Brand: HACH


Hach Pocket Colorimeter II, Ammonia, Mono

Category: Colorimeters
Description: Hach Pocket Colorimeter II Kits
Brand: HACH


Hach Pocket Colorimeter II, Bromine

Category: Colorimeters
Description: Hach Pocket Colorimeter II Kits
Brand: HACH


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

2 Questions | 2 Answers
Displaying questions 1-2
  • Q:

    what is the range of mg/L to detect chromium hexavalent and total chromium. I am looking for one has wide range. I have one has range from 0.0 - 0.6 mg/L.
    Asked on 2/19/2014 by Maged from Everett, WA

    1 answer

    • STAFF EXPERT

      A:

      The measurement range for hexavalent Chromium would be 0.01-0.60 mg/L; the range for total Chromium would also be 0.01-0.60 mg/L. Please contact our technical support department for other options which may have the desired measurement range.

      Answered on 2/19/2014 by James from Technical Support
  • Q:

    Are there test kits for this instrument to test for Silicate, Chloride, Nitrate, Phosphate, and Sulfate? Low Range kits would be preferred.
    Asked on 1/10/2014 by Amy

    1 answer

    • STAFF EXPERT

      A:

      We would have test kits available for measurement of many of these parameters. Included are Cole-Parmer part numbers and ranges for these kits: 1) silica 0.01-1.60mg/L SiO₂ [00214VC]; 2) total chlorine 0.02-2.00mg/L Cl₂ [99574-05]; 3) nitrate 0.2-5.0 mg/L NO₃⁻­N [99574-11]; 4) phosphonates 0.02-2.50mg/L PO42- [00240BN], and 5) sulfate 2-7000mg/L SO₄ ²- [99574-21]

      Answered on 1/10/2014 by James from Technical Support
Displaying questions 1-2

Do you have a question about this product? 

Product Reviews

Hach DR 900 COLORIMETER
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Hach Dissolved Oxygen Test Kit (25150-25)  |  Next: Hach DR/870 Colorimeter