Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Solvent extractant S 316 1 5 kg for Horiba Oil Content Analyzer
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-82900-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Specialty spectrophotometers
Share
|
Solvent extractant S-316, 1.5 kg, for Horiba Oil Content Analyzer
YO-82900-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Petroleum Testing Spectroscopes
Model
100690
Brand
Horiba
Technical Resources
MSDS 1 (103 KB)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show Less...
Related Searches for Solvent extractant S-316, 1.5 kg, for Horiba Oil Content Analyzer
Horiba Oil Content Analyzer Accessories
Horiba Oil Content Analyzer
Horiba Spectroscopy and Spectrophotometers
Horiba Material Testing & Physical Testing
Petroleum Analyzer
Recycling Cans
Specialty Spectrophotometers
Balance 15
Sartorius Products
Torque in Lb
Related Products for Solvent extractant S-316, 1.5 kg, for Horiba Oil Content Analyzer
Image
Product Detail
SKU
Solvent reclaimer SR-300, for recycling Horiba Oil Content Analyzer ex
Category: Specialty spectrophotometers
Description: Horiba Oil Content Analyzer Accessories
Brand:
Horiba
Model Number:
SR-305
YO-82900-20
More Details
Disposable bailer, PTFE, 1-1/5" OD x 3', 10/cs (BTEFS1.5X36X)
Category: Disposable Bailers
Description: FEP 1-1/5" OD Bailers
Brand:
PTFE
Model Number:
BTEFS1.5X36X
YO-99155-02
More Details
Scienceware SPINBAR PTFE ROUND 1-5/8" X 3/8"
Category: Scienceware Stirbars
Description: Scienceware SPINBAR PTFE STIRRING BARS
Brand:
Scienceware
YO-04775-16
More Details
Calibration Service For mass Set With Maximum Value Between 3 To 5 Kg (YO-17109-28)
Category: Mass Sets
Description: ASTM 1-3; OIML F1, F2 (mass sets)
Brand:
Cole-Parmer
YO-17109-28
More Details
Compression Fitting, Reducing Union, 316 SS, 5/16" x 1/4" OD (YO-31406-24)
Category: Reducing Union Connectors
Description: Stainless Steel Compression Reducing Unions
Brand:
Cole-Parmer
YO-31406-24
More Details
Chempette digital repetitive dispenser, 1-5 mL (A3-21DG-5)
Category: Repetitive Dispensers/Pipettors
Description: Cole-Parmer Chempette Digital Repetitive Dispensers
Brand:
Chempette
Model Number:
A3-21DG-5
YO-07848-30
More Details
Brinkmann ChemSaver Dispenser 1-5 mL
Category: Bottletop Dispensers
Description: Brinkmann ChemSaver Dispensers
Brand:
Brinkmann
Model Number:
22 22 100-1
YO-07932-01
More Details
Positive Displacement Microdispensers, 1-5 µ, 250/pk (5-000-2005)
Category: Glass Pipettes
Description: Positive Displacement Microdispensers
Brand:
Cole-Parmer
Model Number:
5-000-2005
YO-07951-00
More Details
Brand Positive Displacement Pipettor Tips, PP, 1-5 Ml, 10/pk
Category: Pipette Tips
Description: Brand Positive Displacement Pipettor Tips
Brand:
Cole-Parmer
Model Number:
702858
YO-24550-77
More Details
Flowmeter, Vortex, Heavy-Duty Brass, 1", 5 to 50 gpm (CP-1)
Category: Heavy-Duty Process Vortex Flowmeters/Transmitters
Description: UFM Vortex Flowmeters/Transmitters
Brand:
Cole-Parmer
Model Number:
CP-1
YO-32997-36
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Solvent extractant S-316, 1.5 kg, for Horiba Oil Content Analyzer
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Solution, 140Na/5K/1.5Li, for Flame Photometers, 100-mL
|
Next: Solvent insert, for 13040-50 chemical trap (YO-13040-54)
31125329-p-content_2 31125329pcontent2