Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-83060-25
Thermo Scientific NanoDrop ND-8000 8-position Spectrophotometer
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-83060-25
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Thermo Scientific NanoDrop Spectrophotometers
Share
|
Thermo Scientific NanoDrop ND-8000 8-position Spectrophotometer
YO-83060-25
Loading price...
Qty
:
Save to shopping list
Saves time and space while utilizing just 1 microliter of sample
Pipette sample directly onto optical surfaceno need for separate cuvettes
Readings in as little as 10 seconds
Small footprint for even the most space-limited lab
These revolutionary Thermo Scientific NanoDrop spectrophotometers have been designed for ease of use and minimal sample utilization. The unit uses the surface tension of the sample as the pathlength and returns results in 10 to 30 seconds.
The small sample size allows you to minimize loss, making units ideal for precious samples. The 1-µL sample can be pipetted back into the container if desired, or simply wipe off and the unit is ready for the next measurement. Eliminating the need for cuvettes helps reduce error in results, giving you confidence that the results obtained are accurate.
The eight-position spectrophotometer has a footprint of just 9-1⁄2" x 12-5⁄8" and includes software. Simply plug the instrument into your computer and you're ready to go.
Note:
Unit is available for sale and use in USA only.Eight-Position Spectrophotometer provides high throughput capabilities by reading eight 1-µL samples simultaneously. Unit has the same features as the single position unit but reads up to 96 samples in just 5 minutes. Easy to use; simply pipette 1 µL of sample onto each optical reader using an 8-channel pipettor.
Specifications
Product Type
UV/Visible Spectroscopes
Wavelength range
220 to 750 nm
Wavelength accuracy
±1 nm
Photometric range
0.02 to 75
Photometric accuracy
±2%
Detector
2048-element linear silicon CCD array
Sample compartments
8
Pathlength
1 mm (with auto range to 0.2 mm)
Source lamp
Xenon flash lamp
Scan time
20 seconds
Weight
7.5 lbs (3.4 kg)
Model
ND-8000
Brand
Thermo Scientific NanoDrop
Accessories
Qty
/
Price
Availability
Calibration check kit for Thermo Scientific NanoDrop ND-8000
YO-83060-92
at
USA only
Call for availability.
Add to Cart
Reconditioning compound PR1 for Thermo Scientific NanoDrop
YO-83060-94
at
USA only
Call for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show Less...
Related Searches for Thermo Scientific NanoDrop ND-8000 8-position Spectrophotometer
Thermo Scientific Nanodrop Nd 8000 Spectrophotometer
Thermo Scientific Nanodrop Spectroscopy and Spectrophotometers
Thermo Scientific Nanodrop 2000
Thermo Scientific Nanodrop Visible/Uv-Visible Spectroscopy
Thermo Scientific Nanodrop Spectrometer Accessories
Thermo Scientific Nanodrop Lite Spectrophotometer
Thermo Scientific Nanodrop Fluorometers
Thermo Scientific Nanodrop Spectrophotometry
Thermo Scientific Nanodrop Accessories for Spectroscopy
Thermo Scientific Nanodrop Fluorospectrometers
Related Products for Thermo Scientific NanoDrop ND-8000 8-position Spectrophotometer
Image
Product Detail
SKU
Thermo Scientific NanoDrop™ ND-8000 UV-VIS Spec with Laptop
Category: Thermo Scientific NanoDrop Spectrophotometers
Description: Thermo Scientific NanoDrop ND 8000 Spectrophotometer
Brand:
Thermo Scientific NanoDrop
Model Number:
ND-2000
YO-83061-54
More Details
Thermo Scientific NanoDrop™ ND-2000 1-position Spectrophotometer
Category: Thermo Scientific NanoDrop Spectrophotometers
Description: Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Brand:
Thermo Scientific NanoDrop
Model Number:
ND-2000
YO-83061-00
More Details
Thermo Scientific NanoDrop™ ND-2000 1-position Spectrophotometer wi
Category: Thermo Scientific NanoDrop Spectrophotometers
Description: Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Brand:
Thermo Scientific NanoDrop
Model Number:
ND-2000
YO-83061-50
More Details
Calibration check kit for Thermo Scientific NanoDrop ND-8000
Category: Calibration Products and Standards
Description: Thermo Scientific NanoDrop Spectrophotometer and Fluorometer Calibration Accessories
Brand:
Thermo Scientific NanoDrop
Model Number:
CHEM-CF-8 (1)
YO-83060-92
More Details
Thermo Scientific NanoDrop™ ND-2000c Spectrophotometer with Cuvett
Category: Thermo Scientific NanoDrop Spectrophotometers
Description: Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Brand:
Thermo Scientific NanoDrop
Model Number:
ND-2000c
YO-83061-12
More Details
Thermo Scientific NanoDrop™ ND-2000 Dual-mode UV-Vis Spectrophotome
Category: Thermo Scientific NanoDrop Spectrophotometers
Description: Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Brand:
Thermo Scientific NanoDrop
Model Number:
ND-2000
YO-83061-52
More Details
Calibration fluid for Thermo Scientific NanoDrop
Category: Calibration Products and Standards
Description: Thermo Scientific NanoDrop Spectrophotometer and Fluorometer Calibration Accessories
Brand:
Thermo Scientific NanoDrop
Model Number:
CHEM-CF1
YO-83060-90
More Details
Thermo Scientific NanoDrop™ Fluorospectrometer, ND-3300 1-position
Category: Fluorometers
Description: Thermo Scientific NanoDrop Fluorospectrometers
Brand:
Thermo Scientific
YO-83060-50
More Details
Reconditioning compound PR1 for Thermo Scientific NanoDrop
Category: Calibration Products and Standards
Description: Thermo Scientific NanoDrop Spectrophotometer and Fluorometer Calibration Accessories
Brand:
Thermo Scientific
Model Number:
CHEM-PR1-KIT
YO-83060-94
More Details
Thermo Scientific NanoDrop Lite Printer Paper Refill 2/pk
Category: Thermo Scientific
Nanodrop Lite Spectrophotometer
Description: Printer Accessories for Thermo Scientific Nanodrop Lite Spectrophotometer
Brand:
Thermo Scientific
Model Number:
714-071400
YO-83060-52
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific NanoDrop ND-8000 8-position Spectrophotometer
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific NanoDrop Lite Spectrophotometer with Printer, 120V
|
Next: Thermo Scientific NanoDrop™ Fluorospectrometer, 120V with Laptop
31125329-p-content_2 31125329pcontent2