Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-84551-70
Hand Scrub Brush, Plastic Block, 24/pk
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-84551-70
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Brushes
Share
|
Hand Scrub Brush, Plastic Block, 24/pk
YO-84551-70
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
These brushes are ideal for increasing personal cleanliness, reducing cross-contamination, and decreasing potential exposure to dangerous chemicals. Choose from three models; plastic-block handle with bristles on both sides, wood-block handle, or the workstation kit. Workstation kit includes brush, security cord, hook, and adhesive-backed washing instructions; instructions in English and Spanish.
Specifications
Product Type
Brushes
Description
Plastic Block
Material
Polypropylene Bristles, Plastic Block
Color
White
Quantity
24
Dimensions
3.5 in L x 1.5 in W
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
Related Searches for Hand Scrub Brush, Plastic Block, 24/pk
Cole-Parmer Hand Scrub Brushes
Hand Scrub Brushes
Plastic Block
Brush Scrubbing
Plastic Brush
Hand Cleaning Products
Cole-Parmer Utility Block Brushes
Cole-Parmer Tap Head Brushes
Cole-Parmer Carboy Brushes
Cole-Parmer Utility & Counter Brushes
Related Products for Hand Scrub Brush, Plastic Block, 24/pk
Image
Product Detail
SKU
Hand Scrub Brush, Wood Block, 12/pk
Category: Laboratory Brushes
Description: Hand Scrub Brushes
Brand:
Cole-Parmer
Model Number:
4550042
YO-84551-71
More Details
Hand Scrub Brush, Plastic Block; Each
Category: Laboratory Brushes
Description: Hand Scrub Brushes
Brand:
Cole-Parmer
Model Number:
3623900
YO-84554-28
More Details
Hand Scrub Brush, Wood Block; Each
Category: Laboratory Brushes
Description: Hand Scrub Brushes
Brand:
Cole-Parmer
Model Number:
4550042
YO-84554-30
More Details
Hand Scrub Brush Workstation Kit (4002100)
Category: Brushes
Description: Hand Scrub Brushes
Model Number:
4002100
YO-84551-72
More Details
Counter scrub brush, blue
Category: Clean Room
Description: Sanitary Scrub Brushes
Brand:
Cole-Parmer
Model Number:
3050-BLUE
YO-86517-32
More Details
Tap Head Brush, 6-1/2", 24/pk
Category: Laboratory Brushes
Description: Tap Head Brushes
Brand:
Cole-Parmer
Model Number:
4015800
YO-84550-57
More Details
Xl Bottle Brush Tufted End 24", 12/pk
Category: Laboratory Brushes
Description: Beaker/Bottle/Flask Brushes
Brand:
Cole-Parmer
Model Number:
620366
YO-84550-88
More Details
Hand tool for plastic hose clamps (YO-06832-97)
Category: Tubing Clamps, Tubing Cutters, and Accessories
Description: Cole-Parmer Plastic Hose Clamp Hand Tool
Brand:
Cole-Parmer
YO-06832-97
More Details
Carboy Brush, 22", Nylon, 12/pk
Category: Laboratory Brushes
Description: Carboy Brushes
Brand:
Cole-Parmer
Model Number:
720354
YO-84550-05
More Details
Carboy Brush, 34", Nylon, 12/pk
Category: Laboratory Brushes
Description: Carboy Brushes
Brand:
Cole-Parmer
Model Number:
720356
YO-84550-06
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hand Scrub Brush, Plastic Block, 24/pk
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hand Scrub Brush Workstation Kit (4002100)
|
Next: Hand Scrub Brush, Plastic Block; Each
31125329-p-content_2 31125329pcontent2