Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-86105-52
Absorbent Zorb Sheet
, 100/cs
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-86105-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Spill Cleanup
Share
|
Absorbent Zorb Sheet™, 100/cs
YO-86105-52
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Pack with liquid shipments to meet federal shipping regulations.
Disposable towel absorbs and encapsulates liquid spills
Specifications
Qty/cs
100
Dimensions
12" x 12"
Brand
Zorb Sheet
Technical Resources
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Isolating Genomic DNA from Whole Blood
Show Less...
Related Searches for Absorbent Zorb Sheet™, 100/cs
Advantec Petri Dishes and Contact Plates
Lab-Tek Plastic Containers
Centrifugation Equipment
Costar Filtration
Aluminum Seal
Chromatography Consumables
Centrifuge Tubes
Wheaton Headspace Vials
Amber Vials
Cole-Parmer Disposable Griffin Low-Form Beakers
Related Products for Absorbent Zorb Sheet™, 100/cs
Image
Product Detail
SKU
Petri dish with pads, 50x11 mm, 100/cs (YO-14005-20)
Category: Petri Dishes and Contact Plates
Description: Advantec Disposable 50-mm Sterile Petri Dishes with Absorbent Pads
Brand:
Advantec
YO-14005-20
More Details
Petri dish without pads, 50x11 mm, 100/cs
Category: Petri Dishes and Contact Plates
Description: Advantec Disposable 50-mm Sterile Petri Dishes with Absorbent Pads
Brand:
Advantec
YO-14005-24
More Details
Disposable polypropylene Griffin low-form beakers, 100mL 100/cs (BMP-BK-100)
Category: Plastic Beakers
Description: Disposable Griffin Low-Form Beakers, PP
Brand:
Guaranteed In Stock Program
Model Number:
BMP-BK-100
YO-06006-42
More Details
Tyvek apron, 36"L x 28"W, 100/cs
Category: Aprons
Description: Dupont Tyvek Apron
Brand:
Tyvek
Model Number:
TY273BWH00010000
YO-86225-80
More Details
Polystyrene lids, Corner-notch, 100/cs (YO-01730-01)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-01730-01
More Details
Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.22um 100/cs
Category: Centrifugal Devices and Filters
Description: Costar Spin-X Centrifuge Tube Filters
Brand:
Costar Spin-X
YO-01937-32
More Details
Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.45µm 100/cs
Category: Centrifugal Devices and Filters
Description: Costar Spin-X Centrifuge Tube Filters
Brand:
Costar Spin-X
YO-01937-36
More Details
Disposable polypropylene Griffin low-form beakers, 15mL 100/cs (BMP-BK-15)
Category: Plastic Beakers
Description: Disposable Griffin Low-Form Beakers, PP
Brand:
Cole-Parmer
Model Number:
BMP-BK-15
YO-06006-12
More Details
Disposable polypropylene Griffin low-form beakers, 30mL 100/cs (BMP-BK-30)
Category: Plastic Beakers
Description: Disposable Griffin Low-Form Beakers, PP
Brand:
Guaranteed In Stock Program
Model Number:
BMP-BK-30
YO-06006-22
More Details
Disposable polypropylene Griffin low-form beakers, 50mL 100/cs (BMP-BK-50)
Category: Plastic Beakers
Description: Disposable Griffin Low-Form Beakers, PP
Brand:
Guaranteed In Stock Program
Model Number:
BMP-BK-50
YO-06006-32
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Absorbent Zorb Sheet™, 100/cs
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Absolute pressure monitor, 0 to 50 psia
|
Next: Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM
31125329-p-content_2 31125329pcontent2