AURELIA Robust ™ Nitrile Gloves, Large, 100/Box

YO-86246-22
Loading price...

  • Micro-textured finish offers a sure grip in wet environments
  • Nonlatex, protein-free, powder-free
Made from a special nitrile compound, these blue gloves provide comfortable fit, durable wear, and tactile sensitivity. Micro-textured finish for superior grip in wet or dry conditions. Beaded cuffs for easy donning and added protection. For single use only. Ambidextrous.
Product Type Gloves
Material Nitrile
Size Large
Length 9.5 in (24 cm)
Thickness (mil) 5
Color Light Blue
Brand AURELIA
Model 9389-L
Manufacturer number 9389-L





Technical Resources
 
Related Products for AURELIA Robust ™ Nitrile Gloves, Large, 100/Box
Image
  Product Detail
SKU

AURELIA Robust ™ Nitrile Gloves, Small, 100/Box

Category: Disposable Nitrile Gloves
Description: Aurelia Robust Nitrile Gloves
Brand: AURELIA


AURELIA Robust ™ Nitrile Gloves, Medium, 100/Box

Category: Disposable Nitrile Gloves
Description: Aurelia Robust Nitrile Gloves
Brand: AURELIA


AURELIA Protege ™ Nitrile Gloves, Large, 100/Box

Category: Disposable Nitrile Gloves
Description: Aurelia Protege Nitrile Gloves
Brand: AURELIA


AURELIA Protege ™ Nitrile Gloves, Small, 100/Box

Category: Disposable Nitrile Gloves
Description: Aurelia Protege Nitrile Gloves
Brand: AURELIA


AURELIA Protege ™ Nitrile Gloves, Medium, 100/Box

Category: Disposable Nitrile Gloves
Description: Aurelia Protege Nitrile Gloves
Brand: AURELIA


AURELIA Robust ™ Nitrile Gloves, X-Large, 100/Box

Category: Disposable Nitrile Gloves
Description: Aurelia Robust Nitrile Gloves
Brand: AURELIA


Microflex Tranquility ™ White Nitrile Gloves, large, 100/box

Category: Disposable Nitrile Gloves
Description: Microflex Tranquility White Nitrile Gloves
Brand: Microflex


High Five Industrial Nitrile Gloves, Large, 100/box

Category: Disposable Nitrile Gloves
Description: High Five Industrial-Grade Nitrile Gloves
Brand: High Five


High Five General-Purpose Cobalt Blue Nitrile Gloves, Large, 100/box

Category: Disposable Nitrile Gloves
Description: High Five General-Purpose Cobalt® Nitrile Gloves
Brand: High Five


Microflex MidKnight ™ Black Nitrile Glove, large, 100/box

Category: Disposable Nitrile Gloves
Description: Microflex MidKnight Black Nitrile Glove
Brand: Microflex


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought

AURELIA Robust ™ Nitrile Gloves, Large, 100/Box
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: AURELIA Protege ™ Nitrile Gloves, X-Large, 100/Box  |  Next: AURELIA Robust ™ Nitrile Gloves, Medium, 100/Box