Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-86364-10
ZetexPlus
®
High-Temperature Gloves, up to 2000°F, one size fits all, 14"L, one pair
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-86364-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature-Resistant Gloves
Share
|
ZetexPlus High-Temperature Gloves, up to 2000°F, one size fits all
YO-86364-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Protect your hands up to 2000°F (1093°C)
A safe alternative to asbestos, these gloves are made of a durable cotton-backed, 11-ounce wool liner. Liner is sewn at the fingertips and double stitched with Kevlar
®
thread throughout for strength and durability. One size fits all. Made in the USA.
Specifications
Product Type
Gloves
Material
Cotton
Size
One size fits all
Length
14 in
Color
White
Lining
11-ounce wool liner
Compliance
European EN407 industrial standard
Quantity
One pair
Brand
ZetexPlus
Manufacturer number
2100012
Model
16000-14-50-00
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
If the Glove Fits: The Right Safety Glove for your Application
Safety Gloves Sizing Guide
Safety Gloves Selection Guide
Show All...
Chemical Compatibility Chart for Safety Apparel
Safety Apparel Material Selection Guide
Cleanroom Garments
Show Less...
Related Searches for ZetexPlus High-Temperature Gloves, up to 2000°F, one size fits all
Zetexplus High Temperature Gloves
Kevlar High Temperature Gloves
Water High Temperature Gloves
18 High Temperature Gloves
Cleanroom High Temperature Gloves
Zetex High-Temperature Gloves and Finger Cots
Fitting High Temperature
High Temperature Plastic Fittings
High Temperature Plastic Tube Fittings
High Temperature Data Loggers
Related Products for ZetexPlus High-Temperature Gloves, up to 2000°F, one size fits all
Image
Product Detail
SKU
Zetex High-Temperature Gloves, up to 1100°F, one size fits all, 14
Category: Temperature-Resistant Gloves
Description: Zetex High-Temperature Gloves and Finger Cots
Brand:
Zetex
Model Number:
ZG1
YO-86364-00
More Details
Zetex High-Temperature Finger Cots, up to 1100°F, one size fits al
Category: Temperature-Resistant Gloves
Description: Zetex High-Temperature Gloves and Finger Cots
Brand:
Zetex
Model Number:
ZFC
YO-86364-02
More Details
Zetex High-Temperature Gloves with Aluminized Back, up to 1100°F,
Category: Temperature-Resistant Gloves
Description: Zetex High-Temperature Gloves and Finger Cots
Brand:
Zetex
Model Number:
160001-14-00-00
YO-86364-20
More Details
Type J High Temperature 12"L, .020" diameter thermocouple probe with m (073098A-J-E-12)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 12L with Miniconnector
Brand:
Oakton
Model Number:
073098A-J-E-12
YO-93630-00
More Details
Type J High Temperature 12"L, .020" diameter thermocouple probe with (073098A-J-G-12)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 12L with Miniconnector
Brand:
Oakton
Model Number:
073098A-J-G-12
YO-93630-01
More Details
Type J High Temperature 12"L, .020" diameter thermocouple probe with m (073098A-J-U-12)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 12L with Miniconnector
Brand:
Oakton
Model Number:
073098A-J-U-12
YO-93630-02
More Details
Type J High Temperature 12"L, .040" diameter thermocouple probe with m (073098B-J-E-12)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 12L with Miniconnector
Brand:
Oakton
Model Number:
073098B-J-E-12
YO-93630-10
More Details
Type J High Temperature 12"L, .040" diameter thermocouple probe with m (073098B-J-G-12)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 12L with Miniconnector
Brand:
Oakton
Model Number:
073098B-J-G-12
YO-93630-11
More Details
Type J High Temperature 12"L, .040" diameter thermocouple probe with m (073098B-J-U-12)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 12L with Miniconnector
Brand:
Oakton
Model Number:
073098B-J-U-12
YO-93630-12
More Details
Type J High Temperature 25"L, .063" diameter thermocouple probe with m (073098C-J-E-25)
Category: Oakton High-Temperature - Flexible Insulated-Wire Probes
Description: High-Temperature Insulated-Wire Probes - 25L with Miniconnector
Brand:
Oakton
Model Number:
073098C-J-E-25
YO-93630-20
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
ZetexPlus High-Temperature Gloves, up to 2000°F, one size fits all
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Zetex High-Temperature Gloves, up to 1100°F, one size fits all, 14
|
Next: ZINC ACETATE DIHYDRATE R 500G (YO-88289-38)
31125329-p-content_2 31125329pcontent2