Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-86364-32
Zetex gloves with leather palm, 1 pr
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-86364-32
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature-Resistant Gloves
Share
|
Zetex gloves with leather palm, 1 pr
YO-86364-32
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Reinforced palms provide excellent abrasion and cut protection
Reinforced gloves offer heat protection up to 800°F (427°C). Reinforced palms are sewn-in and double-stitched for maximum durability. Select kevlar palm gloves for additional cut protection. Leather palm gloves feature superior abrasion resistance. One size fits all. Made in the USA.
Approval:
UL listed
Compliance:
European EN 407 industrial standard
Specifications
Product Type
Gloves
Material
Leather
Size
One size fits all
Color
White
Brand
Zetex
Manufacturer number
2100009
Model
160000-14-00-10
Technical Resources
Related technical articles (Roll over for summary)
If the Glove Fits: The Right Safety Glove for your Application
Safety Gloves Sizing Guide
Safety Gloves Selection Guide
Show All...
Chemical Compatibility Chart for Safety Apparel
Safety Apparel Material Selection Guide
Cleanroom Garments
Show Less...
Related Searches for Zetex gloves with leather palm, 1 pr
Zetex Kevlar and Leather Palm Gloves
Leather Palm
Cole-Parmer Leather-Palm Work Gloves
Gloves Insulated Leather
Cole-Parmer Leather Gloves
Gauntlet Leather Gloves
Leather Working Gloves
Quality Leather Gloves
Zetex Temperature-Resistant Gloves
Nitrile Coated Palm Work Gloves
Related Products for Zetex gloves with leather palm, 1 pr
Image
Product Detail
SKU
Zetex gloves with kevlar palm, 1 pr
Category: Temperature-Resistant Gloves
Description: Zetex Kevlar and Leather Palm Gloves
Brand:
Zetex
Model Number:
160000-14-00-20
YO-86364-30
More Details
Quality Leather-Palm Work Gloves, safety cuffs; size, large; pack of 1
Category: Leather Gloves
Description: Leather-Palm Work Gloves
Brand:
Cole-Parmer
Model Number:
6563-L
YO-86370-12
More Details
Economical Leather-Palm Work Gloves, gauntlet cuffs; one size fits all (7612)
Category: Leather Gloves
Description: Leather-Palm Work Gloves
Brand:
Cole-Parmer
Model Number:
7612
YO-86370-02
More Details
Quality Leather-Palm Work Gloves, gauntlet cuffs; size, extra-large; p
Category: Leather Gloves
Description: Leather-Palm Work Gloves
Brand:
Cole-Parmer
Model Number:
6663-XL
YO-86370-24
More Details
Water-resistant Cryo-Glove gloves, 12"L, medium, 1 pair
Category: Cryogenic Apparel
Description: Cryo-Glove Cryogenic Gloves
Brand:
Cryo-Glove
Model Number:
WRM
YO-09113-02
More Details
Water-resistant Cryo-Glove gloves, 12"L, large, 1 pair
Category: Cryogenic Apparel
Description: Cryo-Glove Cryogenic Gloves
Brand:
Cryo-Glove
Model Number:
WRL
YO-09113-04
More Details
Water-resistant Cryo-Glove gloves, 15"L, medium, 1 pair
Category: Cryogenic Apparel
Description: Cryo-Glove Cryogenic Gloves
Brand:
Cryo-Glove
Model Number:
MAM
YO-09113-12
More Details
Water-resistant Cryo-Glove gloves, 15"L, Large, 1 pair
Category: Cryogenic Apparel
Description: Cryo-Glove Cryogenic Gloves
Brand:
Cryo-Glove
Model Number:
MAL
YO-09113-14
More Details
Neoprene Gloves for Glove Boxes; size 8 1/2; 30"L (5005500)
Category: Accessories For All Glove Boxes
Description: Glove Box Replacement Gloves
Brand:
Cole-Parmer
Model Number:
5005500
YO-09112-12
More Details
Replacement latex gloves for Mini Glove Box; size 9 1/2 (800-GLL)
Category: Glove Boxes
Description: Plas-Labs La Petite Glove Box
Brand:
Plas-Labs
Model Number:
800-GLL
YO-34750-21
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Zetex gloves with leather palm, 1 pr
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Zetex gloves with kevlar palm, 1 pr
|
Next: Zetex High-Temperature Finger Cots, up to 1100°F, one size fits al
31125329-p-content_2 31125329pcontent2