Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-86396-03
NFPA Label, 2" X 2"
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-86396-03
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Warning Labels and Stickers
Share
|
NFPA Label, 2" X 2"
YO-86396-03
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
National Fire Protection Association (NFPA) labels convey general information to emergency personnel regarding the handling of hazardous chemicals. Easily identify the severity of the health hazard, flammability, instability, and the necessary personal protective equipment. Write in the information needed for your particular use. Labels are self-adhesive and made of pressure sensitive paper.
Compliance:
OSHA 29 CFR1910.1200
Specifications
Product Type
Right-to-Know
Compliance
OSHA 29 CFR1910.1200
Material
Pressure sensitive paper
Quantity
250 per roll
Dimensions
2 in x 2 in
Width (in.)
2
Height (in.)
2
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related Searches for NFPA Label, 2" X 2"
Cleanroom Labels Removable 2 X 2
Nfpa Labels
Headspace O 2 /Co 2 Analyzers
Benchtop O 2 /Co 2 Analyzers
1 X 2 Labels
Cole-Parmer Nfpa Right-to-Know Hazard Communication Labels
2 Fuel Flow Meter
2 in Mild Steel Tubing
Cole 2-Propanol
2 Grips
Related Products for NFPA Label, 2" X 2"
Image
Product Detail
SKU
NFPA Label, 2-7/8" X 7/8"
Category: Warning Labels and Stickers
Description: NFPA Right-To-Know Hazard Communication Labels
Brand:
Cole-Parmer
Model Number:
WOL3
YO-86396-13
More Details
NFPA Label, 4" X 4"
Category: Warning Labels and Stickers
Description: NFPA Right-To-Know Hazard Communication Labels
Brand:
Cole-Parmer
Model Number:
HMS14L
YO-86396-04
More Details
NFPA Label, 7/8" X 7/8"
Category: Warning Labels and Stickers
Description: NFPA Right-To-Know Hazard Communication Labels
Brand:
Cole-Parmer
Model Number:
WML1
YO-86396-11
More Details
Waterproof label, 2"W x 1"H
Category: Labels
Description: Waterproof Labels
Brand:
Cole-Parmer
Model Number:
62204-00
YO-62204-00
More Details
Rite in the Rain vinyl labels, 2" x 1-1/8" 3200/pk
Category: Labels
Description: Rite in the Rain Vinyl Labels
Brand:
Cole-Parmer
Model Number:
6506
YO-06570-38
More Details
Cole-Parmer vinyl labels, 2" x 1-1/8" 3200/pk
Category: Labels
Description: Cole-Parmer Vinyl Labels
Brand:
Cole-Parmer
Model Number:
LVL-1182
YO-06530-02
More Details
Hazardous Material Identification Label, 2" X 2"
Category: Warning Labels and Stickers
Description: Right-To-Know Hazard Communication Labels
Brand:
Cole-Parmer
Model Number:
HM29
YO-86396-00
More Details
Hazardous Material Identification Label, 2-7/8"W X 7/8"H
Category: Warning Labels and Stickers
Description: Right-To-Know Hazard Communication Labels
Brand:
Cole-Parmer
Model Number:
WOL4
YO-86396-12
More Details
Waterproof label, 4"W x 2"H
Category: Labels
Description: Waterproof Labels
Brand:
Cole-Parmer
Model Number:
62204-20
YO-62204-20
More Details
Cryogenic label for laser printers, 1 9/32"W x 1/2"H
Category: Cryogenic Labels
Description: Cryogenic Labels for Laser Printers
Brand:
Cole-Parmer
Model Number:
9187-1700
YO-62206-70
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
NFPA Label, 2" X 2"
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: NFPA Label, 2-7/8" X 7/8"
|
Next: NFPA Label, 4" X 4"
31125329-p-content_2 31125329pcontent2