3M Virtua ™ 2.0 Magnifier Safety Eyewear, Clear Lens

YO-86472-06
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Safety glasses for those who need the assistance of a reading lens
Large lens size offers maximum protection while allowing for reading magnification at close range. Placement of magnifying lens, like a bifocal, reduces headaches and eyestrain. Wraparound polycarbonate construction is designed to resist impacts and block 99.9% of UV rays. Flexible temples and universal-fit nosebridge.

Compliance: ANSI Z87.1-2003, CSA Z94.3
Product Type Glasses
Color Black
Lens Clear
Compliance ANSI Z87.1-2003, CSA Z94.3
Brand 3M
Model 11514-00000
Manufacturer number 11514-00000-20





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for 3M Virtua ™ 2.0 Magnifier Safety Eyewear, Clear Lens
    Image
      Product Detail
    SKU

    3M Virtua ™ 1.5 Magnifier Safety Eyewear, Clear Lens

    Category: AOSafety Safety Glasses
    Description: 3M Virtua Readers Safety Eyewear
    Brand: 3M


    3M Virtua ™ 2.5 Magnifier Safety Eyewear, Clear Lens

    Category: AOSafety Safety Glasses
    Description: 3M Virtua Readers Safety Eyewear
    Brand: 3M


    3M Virtua V6 ™ Safety Eyewear, Clear Lens

    Category: AOSafety Safety Glasses
    Description: 3M Virtua V6 Safety Eyewear
    Brand: 3M


    3M QX ™ Safety Eyewear, Clear lens

    Category: AOSafety Safety Glasses
    Description: 3M QX Safety Eyewear
    Brand: 3M


    3M Metaliks ™ Safety Eyewear, Clear lens

    Category: AOSafety Safety Glasses
    Description: 3M Metaliks Sport Safety Eyewear
    Brand: 3M


    3M, Refine ™ , Women's Safety Eyewear, Clear Lens Anti-Fog, Mocha Frame

    Category: AOSafety Safety Glasses
    Description: 3M Refine Women's Safety Eyewear
    Brand: 3M


    Uvex Seismic ™ Safety Eyewear, Clear lens

    Category: Uvex Safety Glasses
    Description: Uvex Seismic Safety Eyewear
    Brand: Uvex


    Uvex Seismic ™ Safety Eyewear, Clear lens, Antifog coating

    Category: Uvex Safety Glasses
    Description: Uvex Seismic Safety Eyewear
    Brand: Uvex


    Uvex Seismic ™ Safety Eyewear, Clear lens, Dual antifog/antiscratch coating

    Category: Uvex Safety Glasses
    Description: Uvex Seismic Safety Eyewear
    Brand: Uvex


    3M Virtua Sport ™ Safety Eyewear, Clear frame, Clear lens

    Category: AOSafety Safety Glasses
    Description: 3M Virtua Sport Safety Eyewear
    Brand: 3M


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought

    3M Virtua ™ 2.0 Magnifier Safety Eyewear, Clear Lens
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: 3M Virtua ™ 1.5 Magnifier Safety Eyewear, Clear Lens  |  Next: 3M Virtua ™ 2.5 Magnifier Safety Eyewear, Clear Lens