Home  >  General Accessories   





Technical Resources
 
Related Products for Pulse input cable, 1/8" stereo plug to BNC connector, 6 ft
Image
  Product Detail
SKU

Pulse output cable, 1/8” mono plug to BNC connector, 6 ft

Category: General Accessories
Description: Replacement Accessories
Brand: Lab Products


Eutech Instruments pH 200 1/8-DIN pH/ORP controller

Category: 1/8-DIN pH/ORP Control
Description: Eutech Instruments pH 200 1/8-DIN pH/ORP Controller/Transmitter
Brand: Eutech Instruments


Eutech Instruments pH 190 1/8-DIN pH/ORP controller

Category: 1/8-DIN pH/ORP Control
Description: Eutech Instruments pH 190 1/8-DIN pH/ORP Controller/Transmitter
Brand: Eutech Instruments


Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Transmitter

Category: 1/8-DIN pH/ORP Control
Description: Cole-Parmer pH/ORP/Temperature 1/8-DIN Controller and Transmitter
Brand: Cole-Parmer


Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Controller

Category: 1/8-DIN pH/ORP Control
Description: Cole-Parmer pH/ORP/Temperature 1/8-DIN Controller and Transmitter
Brand: Cole-Parmer


Cole-Parmer 300 Conductivity/Resistivity/Temperature 1/8-DIN Transmitter

Category: 1/8-DIN Conductivity Control
Description: Cole-Parmer Conductivity/Resistivity/Temperature 1/8-DIN Transmitter and Controller
Brand: Cole-Parmer


Cole-Parmer 350 Conductivity/Resistivity/Temperature 1/8- DIN Controller

Category: 1/8-DIN Conductivity Control
Description: Cole-Parmer Conductivity/Resistivity/Temperature 1/8-DIN Transmitter and Controller
Brand: Cole-Parmer


Tygon Silicone Tubing, 1/8"ID x 3/16"OD, 50 ft/pack

Category: Tygon Tubing
Description: Tygon Silicone Tubing
Brand: Tygon


FEP Tubing, 1/8"ID x 3/16"OD, 25 ft/pack (50694)

Category: Rigid Tubing
Description: Cole-Parmer FEP Tubing
Brand: Cole-Parmer


Peroxide-Cured Silicone Tubing, 1/8"ID x 3/16"OD, 25 ft/pack

Category: Silicone Tubing
Description: Peroxide-Cured Silicone Tubing, 25 ft/pack
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews