Home  >  Chlorof to Chloroi >
  

Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 liter)

YO-88005-34
Loading price...

Usually ships in 14 days.
Qty

  • Assay (including preservative)—≥99.8%
  • Color (APHA)—≤10
  • Preservative (Pentene)—50ppm
  • Optical Absorbance at 245nm—≤1.00
  • at 250nm—≤0.32
  • at 254nm—≤0.15
  • at 280nm—≤0.01
  • Fluorescence Background (as Quinine Sulfate)—To pass test
  • Refractive Index at 25°C—1.4430-1.4460
  • Residue after Evaporation—≤2ppm
  • Water—≤0.02%
  • Acid and Chloride—To pass test
  • Free Chlorine—To pass test
  • Lead—≤0.05ppm
  • Suitability for Use in Dithizone Tests—To pass test
  • Acetone and Aldehyde [as (CH3)2CO]—To pass test (about 0.005%)
  • Submicron Filtered.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagents
Purity grade HPLC
CAS Number 67-66-3
Form Liquid
Formula weight 119.38
Empirical formula CHCl3
Synonyms Trichloromethane
Packaging Amber Glass
Storage symbol Blue
UN number UN 1888
Hazard symbol Harmful
Quantity (mL) 4000
DOT information DOT Class 6.1
DOT description Poison
Quantity 4 liter





Technical Resources
 
Related Products for Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 liter)
Image
  Product Detail
SKU

Methylene Chloride (with Cyclohexene Preservative/HPLC) (4 liter)

Category: Methyle to Methylh
Description: Methylene Chloride
Brand: Cole-Parmer


Chloroform (Approx. 0.75% Ethanol as Preservative/Technical) (20 liter)

Category: Chlorof to Chloroi
Description: Chloroform
Brand: Cole-Parmer


Chloroform (Approx. 0.75% Ethanol as Preservative/Certified ACS) (4 liter)

Category: Chlorof to Chloroi
Description: Chloroform
Brand: Cole-Parmer


Methylene Chloride (Not Stabilized/HPLC) (4 liter)

Category: Methyle to Methylh
Description: Methylene Chloride
Brand: Cole-Parmer


1,2,4-Trichlorobenzene (HPLC) (4 liter)

Category: Trichlo to Triethyl
Description: 1,2,4-Trichlorobenzene
Brand: Cole-Parmer


n-Butyl Chloride (HPLC) (4 liter) n-butyl chloride n-butyl chloride hplc 4l

Category: Bug to Butyll
Description: n-Butyl Chloride
Brand: Cole-Parmer


n-Butyl Chloride (HPLC) (4 liter) n-butyl chloride n-butyl chloride hplc 4l sk

Category: Bug to Butyll
Description: n-Butyl Chloride
Brand: Cole-Parmer


Chloroform (Approx. 0.75% Ethanol as Preservative/Certified ACS) (200 liter)

Category: Chlorof to Chloroi
Description: Chloroform
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 liter)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Chloroform (Approx. 0.75% Ethanol as Preservative/Technical) (20 liter)  |  Next: Chloroform 99.9+% Hplc 4lt