Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-55200-02
Buffer tablets
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-55200-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Buffers and Solutions for pH/Ion Measurement
Share
|
Buffer tablets, pH 7 (bag of 50)
YO-55200-02
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Ideal when small amounts of buffer are needed
Quickly calibrate your pH meter in the lab or field. To make your own fresh buffer solutions, simply stir a tablet into 20 mL of distilled water. Bag of 50.
Specifications
Product Type
pH Buffers
Description
Buffer tablets, pH 7
Quantity
bag of 50
Qty/pk
50
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Tips for pH Buffer Use
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Related Searches for Buffer tablets, pH 7 (bag of 50)
Buffer Solution Ph 7 Yellow
Ph Buffer Tablets
Buffer Sol Ph 4.01
Cole-Parmer Buffer Solution Ph 10
Cole-Parmer Buffer Solution
Buffer Solution Ph 4.01 Red
Buffer Sol Ph 10.00
Buffer Sol Ph 4.01 Red
Buffer Solution Ph 10 Blue
Buffer Sol Ph 10.00 Blue
Related Products for Buffer tablets, pH 7 (bag of 50)
Image
Product Detail
SKU
Buffer tablets, pH 4, (bag of 50)
Category: Buffers and Solutions for pH/Ion Measurement
Description: pH Buffer Tablets
Brand:
Guaranteed In Stock Program
Model Number:
MAKE-UP
YO-55200-00
More Details
Cole-Parmer Buffer, pH 7 One Shot™ Vials
Category: pH/Ion Buffers, and Storage Solutions
Description: Cole-Parmer Buffers
Brand:
Cole-Parmer
Model Number:
PH 7
YO-00655-32
More Details
Cole-Parmer Buffer, pH 7, 473-mL
Category: pH/Ion Buffers, and Storage Solutions
Description: Cole-Parmer Buffers
Brand:
Cole-Parmer
Model Number:
PH 7
YO-00655-42
More Details
Cole-Parmer pH 7 buffer solution, 500 mL bottle, NIST-traceable refere
Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00651-02
More Details
Cole-Parmer pH 7 buffer solution, 1 L bottle, NIST-traceable reference
Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00651-32
More Details
Cole-Parmer pH 7 buffer solution, 4 L bottle, NIST-traceable reference
Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00651-72
More Details
Cole-Parmer pH 7 buffer, yellow, 500mL, NIST-traceable
Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00651-08
More Details
Cole-Parmer pH 7 buffer, yellow, 1L, NIST-traceable
Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00651-38
More Details
Cole-Parmer pH 7 buffer, yellow, 4L, NIST-traceable
Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00651-78
More Details
Buffer capsules, pH 4.0, orange (272,PH4)
Category: Buffers and Solutions for pH/Ion Measurement
Description: pH Buffer Capsules
Brand:
Metrepak
Model Number:
272,PH4
YO-05977-12
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Buffer tablets, pH 7 (bag of 50)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Buffer tablets, pH 4, (bag of 50)
|
Next: Buffered beef extract broth, prepared; 1 liter, 10/pk (YO-14204-52)
31125329-p-content_2 31125329pcontent2