Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-55500-04
ACCUMET
PH ELECTRODE, COMBINATION, MICROPROBE, REFILLABLE, GLASS BODY, BNC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-55500-04
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
accumet Combination pH Electrodes from Cole-Parmer
Share
|
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
YO-55500-04
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
ACCUMET combination pH electrodes deliver superior reproducibility in aqueous and titration samples. Glass-body models resist chemical attack. Epoxy-body models are impact-resistant and are ideal for rough handling. Electrodes with BNC or U.S. Standard connectors are available; both include a 30-mL bottle of fill solution and 76-cm lead.
Specifications
Product Type
pH Electrodes
Description
Refillable, microprobe
Body type
Glass
Reference junction
Single
Temperature range
230°F (110°C)
Reference cell type
Ag/AgCl
Connector
BNC
Suggested uses
Small vessels
Dimensions
127 mm L x 6.4 mm Dia
Brand
Accumet
Accessories
Qty
/
Price
Availability
Oakton reference fill solution, 3.8 M KCl with AgCl, two 125-mL bottles/pk
YO-05992-47
PACKAGE
Call for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
pH FAQs
Spectroscopy Applications Selection Guide
Testing the pH Value of Pure Water
Show All...
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Part 2: Troubleshooting pH Electrodes
Series Part 1: How to Store, Clean, and Recondition pH Electrodes
pH Electrode Selection Guide
pH Electrode Theory
Measuring the pH of Pure Water
pH Systems
pH Electrode Care
pH Measurement FAQs
Laboratory pH Electrodes
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Accumet Ph Electrodes Refilling
Accumet Ph Electrode
Accumet Ph Electrode Solution
Combination Electrodes Glass Body Accumet
Accumet Microprobe Combination Ph Electrodes
Ap115 Accumet Ph Meter
Accumet Ph Meter 910
Serial Number of Accumet Ph Meter
Accumet Ph Meter Parts
Accumet Ph Meter User Manual
Related Products for Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Image
Product Detail
SKU
Accumet pH Electrode, Combination, Flat, Refillable, Epoxy Body, BNC
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand:
Accumet
YO-55500-14
More Details
Accumet pH Electrode, Combination, Microprobe, Long-neck, Refillable,
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Glass-body ACCUMET Compatible Combination pH Electrodes
Brand:
Accumet
YO-55500-06
More Details
Accumet Accuphast pH Electrode, Combination, Microprobe, Refillable, G
Category: pH/Ion Laboratory Electrodes
Description: Accumet accupHast high-performance, double-junction combination pH electrodes
Brand:
Accumet
YO-55500-24
More Details
pH electrode, combination, spear tip, glass body, interchangeable conn (YO-05658-60)
Category: pH/Ion Laboratory Electrodes
Description: Cole-Parmer pH Electrodes with Interchangeable Cables
Brand:
Cole-Parmer
YO-05658-60
More Details
Accumet pH Electrode, Combination, General Purpose, Refillable, Glass
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Glass-body ACCUMET Compatible Combination pH Electrodes
Brand:
Accumet
YO-55500-00
More Details
Accumet pH Electrode, Combination, General Purpose, Refillable, Epoxy
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand:
Accumet
YO-55500-10
More Details
Accumet pH Electrode, Combination, General Purpose, Gel-filled, Epoxy
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand:
Accumet
YO-55500-16
More Details
Accumet pH Electrode, Combination, General Purpose, Long-neck, Gel-fil
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand:
Accumet
YO-55500-18
More Details
Accumet pH Electrode, Combination, Double-junction, Gel-filled, Epoxy
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand:
Accumet
YO-55500-20
More Details
pH electrode, combination, long-neck, epoxy body, interchangeable conn (YO-05658-65)
Category: pH/Ion Laboratory Electrodes
Description: Cole-Parmer pH Electrodes with Interchangeable Cables
Brand:
Cole-Parmer
YO-05658-65
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Accumet pH Electrode, Combination, Microprobe, Long-neck, Refillable,
|
Next: Accumet XL150 pH Benchtop Meter Only
31125329-p-content_2 31125329pcontent2