Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-55500-20
ACCUMET
PH ELECTRODE, COMBINATION, DOUBLE-JUNCTION, GEL-FILLED, EPOXY BODY, BNC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-55500-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
accumet Combination pH Electrodes from Cole-Parmer
Share
|
Accumet pH Electrode, Combination, Double-junction, Gel-filled, Epoxy
YO-55500-20
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
ACCUMET combination pH electrodes deliver superior reproducibility in aqueous and titration samples. Glass-body models withstand high temperatures and resist chemical attack. Epoxy-body models are impact-resistant and are ideal for rough handling. Electrodes with BNC or U.S. Standard connectors are available; both include a 30-mL bottle of fill solution and 76-cm lead. Models 055500-20 and -21 have a one-meter lead.
Specifications
Product Type
pH Electrode
Body type
Epoxy
Reference junction
Double-junction
Temperature range
176°F (80°C)
Reference cell type
Ag/AgCl
Connector
BNC
Suggested uses
Paper pulp testing
Dimensions
140 mm L x 10 mm Dia
Brand
Accumet
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
pH FAQs
Spectroscopy Applications Selection Guide
Testing the pH Value of Pure Water
Show All...
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Part 2: Troubleshooting pH Electrodes
Series Part 1: How to Store, Clean, and Recondition pH Electrodes
pH Electrode Selection Guide
pH Electrode Theory
Measuring the pH of Pure Water
pH Systems
pH Electrode Care
pH Measurement FAQs
Laboratory pH Electrodes
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Accumet pH Electrode, Combination, Double-junction, Gel-filled, Epoxy
Ph Electrode Double Junction Gel Filled
Ph Tester 20 Double Junction
Accumet Ph Electrode
Accumet Ph Electrode Solution
Accumet Ph Electrodes Refilling
Accumet Microprobe Combination Ph Electrodes
Oakton Ph Electrode Double Junction
Accumet Combination Ph Electrodes from Cole-Parmer
Accumet Laboratory Ph Electrodes
Wd 358005 14 Ph Electrode
Related Products for Accumet pH Electrode, Combination, Double-junction, Gel-filled, Epoxy
Image
Product Detail
SKU
Accumet pH Electrode, Combination, General Purpose, Gel-filled, Epoxy
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand:
Accumet
YO-55500-16
More Details
Accumet Accuphastreg pH Electrode, Combination, Double-junction, Refi
Category: pH/Ion Laboratory Electrodes
Description: Accumet accupHast high-performance, double-junction combination pH electrodes
Brand:
Accumet
YO-55500-22
More Details
Accumet Accuphast pH Electrode, Combination, Double-junction, Refillab
Category: pH/Ion Laboratory Electrodes
Description: Accumet accupHast high-performance, double-junction combination pH electrodes
Brand:
Accumet
YO-55500-26
More Details
pH electrode, combination, double-junction, gel-filled, BNC connector
Category: pH/Ion Laboratory Electrodes
Description: Cole-Parmer Epoxy-Body General Purpose pH and ORP Electrodes
Brand:
Cole-Parmer
YO-59001-70
More Details
accumet accuTupH pH Electrode, Combination, Double-junction, Refillabl
Category: accumet Combination pH Electrodes from Cole-Parmer
Description: accumet accuTupH rugged, combination glass-body pH electrodes
Brand:
Accumet
YO-55502-02
More Details
pH electrode, combination, single-junction, gel-filled, BNC connector
Category: pH/Ion Laboratory Electrodes
Description: Cole-Parmer Epoxy-Body General Purpose pH and ORP Electrodes
Brand:
Cole-Parmer
YO-59001-65
More Details
PH ELECTRODE, COMBINATION, DOUBLE-JUNCTION, REFILLABLE, BNC (YO-05991-12)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Cole-Parmer
YO-05991-12
More Details
Cole-Parmer 3-ft pH electrode, submersible, double-junction, 100 ohm R (YO-27001-83)
Category: Cole-Parmer 3-ft Submersible pH and ORP Electrodes
Description: Cole-Parmer Submersible 3 Foot pH electrodes
Brand:
Cole-Parmer
YO-27001-83
More Details
Cole-Parmer Tuff-Tip pH Electrode with PTFE Junction, CPVC housing, no (YO-27003-01)
Category: Cole-Parmer In-Line or Submersible, Tuff-Tip, Semi-Domed, pH and ORP Electrodes with PTFE Junctions
Description: Cole-Parmer Tuff-Tip pH Electrodes with PTFE Junctions
Brand:
PTFE
YO-27003-01
More Details
Cole-Parmer Tuff-Tip pH Electrode With PTFE Junction, CPVC housing, 10 (YO-27003-03)
Category: Cole-Parmer In-Line or Submersible, Tuff-Tip, Semi-Domed, pH and ORP Electrodes with PTFE Junctions
Description: Cole-Parmer Tuff-Tip pH Electrodes with PTFE Junctions
Brand:
PTFE
YO-27003-03
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Accumet pH Electrode, Combination, Double-junction, Gel-filled, Epoxy
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Accumet MicroProbe™ combination electrode, rigid stem 1-1/2" L
|
Next: Accumet pH Electrode, Combination, Flat, Refillable, Epoxy Body, BNC
31125329-p-content_2 31125329pcontent2