Iodine Solution (Wijs) (4 liter) (SI1064)

YO-88084-75
Loading price...

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 7553-56-2
    Form Liquid
    Packaging Amber Glass
    Storage symbol White
    UN number UN 2920
    Hazard symbol Corrosive
    Quantity (mL) 4000
    DOT information DOT Class 8
    DOT description : Corrosive
    Manufacturer number SI1064
    Quantity 4 liter





    Technical Resources
     
    Related Products for Iodine Solution (Wijs) (4 liter) (SI1064)
    Image
      Product Detail
    SKU

    Starch Solution (Stabilized) (4 liter)

    Category: Solketa to Sulfom
    Description: Starch Solution
    Brand: Cole-Parmer


    Tris-Acetate-EDTA, 50X Solution (Electrophoresis) (4 liter)

    Category: Tris
    Description: Tris–Acetate–EDTA, 50X Solution
    Brand: Cole-Parmer


    Tris-Acetate-EDTA, 10X Solution (Electrophoresis) (4 liter)

    Category: Tris
    Description: Tris–Acetate–EDTA, 10X Solution
    Brand: Cole-Parmer


    Tris-Borate-EDTA, 1X Solution (Electrophoresis) (4 liter)

    Category: Tris
    Description: Tris–Borate–EDTA, 1X Solution
    Brand: Cole-Parmer


    Tris-Acetate-EDTA, 1X Solution (Electrophoresis) (4 liter)

    Category: Tris
    Description: Tris–Acetate–EDTA, 1X Solution
    Brand: Cole-Parmer


    Calcium Hydroxide Topical Solution (USP) (4 liter) (YO-88084-86)

    Category: Ca to Calycot
    Description: Calcium Hydroxide Topical Solution
    Brand: Hydroxide


    Silver Nitrate Solution (0.25N) (4 liter) (SS8164)

    Category: Shikimi to Soda
    Description: Silver Nitrate Solution, 0.25N
    Brand: Cole-Parmer


    Sodium Thiosulfate Solution (0.2N) (4 liter) (SS8174)

    Category: Sodiump to Sodiumz
    Description: Sodium Thiosulfate Solution
    Brand: Cole-Parmer


    Iodine solution according to Wijs 0.1 mol ICl/l (1l) (AC270570010)

    Category: Ia to Iodine
    Description: Iodine solution according to Wijs
    Brand: Cole-Parmer


    Ethylenediamine Tetraacetic Acid, Disodium Salt, Standard Solution (1mL=1mg CaCO 3 /Certified) (4 liter)

    Category: Ethyle to Ethyli
    Description: Ethylenediamine Tetraacetic Acid, Disodium Salt, Standard Solution
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews