2,5-Dimethylthiophene 98.5% (100ml) (AC116881000)

YO-88111-34
Loading price...

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 638-02-8
    Form Liquid
    Formula weight 112.19
    Empirical formula C6H8S
    Quantity (mL) 100
    Manufacturer number AC116881000
    Quantity 100ml





    Technical Resources
     
    Related Products for 2,5-Dimethylthiophene 98.5% (100ml) (AC116881000)
    Image
      Product Detail
    SKU

    -N-FMOC-2-(5-Bromothienyl)alanine 95%, (98% e.e.) (1g)

    Category: Fluoroo to FMOCB
    Description: ® -N-FMOC-2-(5-Bromothienyl)alanine
    Brand: Cole-Parmer
    Model Number: AC362480010


    -N-FMOC-2-(5-Bromothienyl)alanine 95%, (98% e.e.) (500mg)

    Category: Fluoroo to FMOCB
    Description: ® -N-FMOC-2-(5-Bromothienyl)alanine
    Brand: Cole-Parmer
    Model Number: AC362485000


    2-Furoyl chloride 98+% (100ml) (AC165381000)

    Category: Forskol to Fusaric
    Description: 2-Furoyl chloride
    Brand: Cole-Parmer
    Model Number: AC165381000


    5-Nonanone 98% (100ml) (AC159761000)

    Category: Nitropi to Nylon
    Description: 5-Nonanone
    Brand: Cole-Parmer
    Model Number: AC159761000


    4-Methyl-5-thiazoleethanol 98% (100ml) (AC184061000)

    Category: Methylr to Methylz
    Description: 4-Methyl-5-thiazoleethanol
    Brand: Cole-Parmer
    Model Number: AC184061000


    2-Thiophenecarbonyl chloride 98% (100ml) (AC187771000)

    Category: Tetrar to Thymine
    Description: 2-Thiophenecarbonyl chloride
    Brand: Cole-Parmer
    Model Number: AC187771000


    2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 99+% (1g) (AC403220010)

    Category: Bromon to Bromop
    Description: 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
    Brand: Cole-Parmer
    Model Number: AC403220010


    2 4-lutidine 98+% 100ml

    Category: Chemicals, Reagents, & Lubricants
    Description: Specialty Chemicals
    Brand: Cole-Parmer
    Model Number: AC125181000


    2 5-dichloropyridine 98% 25g

    Category: Chemicals, Reagents, & Lubricants
    Description: Specialty Chemicals
    Brand: Cole-Parmer
    Model Number: AC186140250


    2 3-lutidine 98+% 100ml (AC268761000)

    Category: Chemicals, Reagents, & Lubricants
    Description: Specialty Chemicals
    Brand: Cole-Parmer
    Model Number: AC268761000


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    2,5-Dimethylthiophene 98.5% (100ml) (AC116881000)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: 2,5-Dimethylpyrazine 99% (5g) (AC174520050)  |  Next: 2,5-Dimethylthiophene 98.5% (25ml) (AC116880250)