WTW ProfiLine pH 3110 meter with Sentix 21 probe

YO-58890-02
Loading price...

Call for availability.
Qty

  • A rugged new line of electrochemistry meters with a range of features, benefits, and price points to meet every application and budget!
Durable meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timer ensures more reliable results.

Meters include: four AA batteries.

Meter kits add: specified probe, calibration solutions, and hard carrrying case.

Meters with drinking water kit further add: Sentix® 81 probe, stand, beaker, measuring vessel, and CD-ROM.
Product Type pH Meter
Meter style Portable
Dimensions (" H) 3.1
Dimensions (" D) 2.2
Dimensions (" L) 7.0
Range (pH) 2.000 to 20.000
Range (mV) ± 1200, ±2000
Range (Temperature °C) 5 to 105
Range (Temperature °F) 23 to 221
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Accuracy (pH) ±0.005 pH
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ± 0.3 mV, ± 1 digit
Temperature compensation Automatic or manual
Display LCD
Calibration Up to 3 points
Waterproof Yes
Buffer recognition USA, NIST, DIN
Power Four AA batteries (included)
CE Compliance Yes
Brand WTW
Model pH 3110

Accessories
Qty/Price
Availability

EACH

Call for availability.

EACH

Usually ships in 14 days or sooner.

EACH

Call for availability.
Calibrations
Qty/Price
Availability

EACH
Usually ships in 21 days.




Technical Resources
 
Related Products for WTW ProfiLine pH 3110 meter with Sentix 21 probe
Image
  Product Detail
SKU

WTW ProfiLine pH 3110 meter kit with SenTix 41 probe

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3110 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3210 meter with Sentix 51 probe

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3210 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3210 meter with Sentix 41 probe

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3210 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3110 Handheld Meter Only

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3110 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3110 drinking water set

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3110 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3310 meter with Sentix 51 electrode

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3310 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3310 meter kit with SenTix 41 pH electrode

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3310 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3210 Handheld Meter Only

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3210 Handheld pH Meters
Brand: WTW


WTW ProfiLine pH 3310 Handheld Meter Only

Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3310 Handheld pH Meters
Brand: WTW


WTW Cond 3110 meter kit and TetraCon probe, 1-m cable

Category: Conductivity/Resistivity/TDS Handheld Meters
Description: WTW ProfiLine Cond 3110 Handheld Conductivity Meters
Brand: WTW


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

WTW ProfiLine pH 3110 meter with Sentix 21 probe
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: WTW ProfiLine pH 3110 meter kit with SenTix 41 probe  |  Next: WTW ProfiLine pH 3210 drinking water set