Home  >  Chemicals, Reagents, & Lubricants >
  

Gramine 99% 10g

YO-88117-05
Loading price...

Usually ships in 14 days.
Qty
    A lab reagent is a staple in chemical applications. It is a great way to test chemical reactions or to see if a reaction will occur. You can find one just about anywhere including Cole-Parmer’s catalog which features a broad range of laboratory reagents, cleaning chemicals and buffers commonly found in labs. But occasionally workers run across an application that requires a specialty lab reagent. You might need Malachite Green, a high purity biological stain. How do you go about selecting a lab reagent especially if there are new and more advanced compounds out there that you might want to try? For the novice a lab reagent kit may be in order. Using kits can prevent mistakes that are related to the reagent rather than the application. If you’re pretty advanced and haven’t found that specialty compound you’re searching for try talking to a Cole-Parmer technical expert. They often have information about chemicals not listed on the website but available for special order or by other means. Cole-Parmer also offers custom blending services for specific chemical mixes. With advances in technology selecting a lab reagent may not be as routine as originally thought so it’s great to have some help.

    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    Manufacturer number AC120180100





    Technical Resources
     
    Related Products for Gramine 99% 10g
    Image
      Product Detail
    SKU

    4'-Aminoacetophenone 99% (10g) (AC103090100)

    Category: Ametho to Aminob
    Description: 4'-Aminoacetophenone
    Brand: Cole-Parmer
    Model Number: AC103090100


    2-Hydroxybenzophenone 99% (10g) (AC146920100)

    Category: Hydroxy to Hypoxan
    Description: 2-Hydroxybenzophenone
    Brand: Cole-Parmer
    Model Number: AC146920100


    5-Nitro-m-xylene 99+% (10g) (AC160180100)

    Category: Nitroc to Nitroph
    Description: 5-Nitro-m-xylene
    Brand: Cole-Parmer
    Model Number: AC160180100


    Di-tert-butyl dicarbonate 99% (10g) (AC194670100)

    Category: Dirheni to Doctor
    Description: Di-tert-butyl dicarbonate
    Brand: Cole-Parmer
    Model Number: AC194670100


    Silver nitrite 99% (10g) (AC210690100)

    Category: Shikimi to Soda
    Description: Silver nitrite
    Brand: Cole-Parmer
    Model Number: AC210690100


    Silver chloride 99+% (10g) (AC211270100)

    Category: Shikimi to Soda
    Description: Silver Chloride
    Brand: Cole-Parmer
    Model Number: AC211270100


    D-Leucine 99% (10g) (AC226900100)

    Category: Leb to Lyxosem
    Description: D-Leucine
    Brand: Cole-Parmer
    Model Number: AC226900100


    3-Isobutoxy-2-cyclohexen-1-one 99% (10g) (AC291470100)

    Category: Iron to Isopent
    Description: 3-Isobutoxy-2-cyclohexen-1-one
    Brand: Cole-Parmer
    Model Number: AC291470100


    4-Acetamidobutyric acid 99+% (10g) (AC102290100)

    Category: A to Acetic
    Description: 4-Acetamidobutyric acid
    Brand: Cole-Parmer
    Model Number: AC102290100


    N-Acetyl-L-tyrosine ethyl ester monohydrate 99% (10g) (AC102750100)

    Category: Acetylf to Acetylz
    Description: N-Acetyl-L-tyrosine ethyl ester monohydrate
    Brand: Cole-Parmer
    Model Number: AC102750100


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews