Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Replacement pH Electrode for Hach SensIon Meters
Click to enlarge
More Product Details
Specifications
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-59005-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Accessories for pH/Ion Meters
Share
|
Replacement pH Electrode for Hach SensIon™ Meters
YO-59005-52
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Model
51910-00
Brand
Hach
Technical Resources
Instruction Manual (837 KB)
MSDS 1 (135 B)
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Replacement pH Electrode for Hach SensIon™ Meters
Hanna Hi1270 Replacement Ph Electrode
6mm Diameter Replacement Ph Electrode
Control Wizard Replacement Ph Electrode
Replacement Ph Electrode
Corning Replacement Ph Electrode
Hach do Meter
Ph Electrode Replaceable Fret
Corning Ph Electrode Cat 176155
Corning Ph Electrode 476157
Pinnacle Ph Electrodes (Formerly Corning
Related Products for Replacement pH Electrode for Hach SensIon™ Meters
Image
Product Detail
SKU
Replacement pH electrode, waterproof, for meter kits 58901-00 and -05
Category: pH/Ion Laboratory Electrodes
Description: Pinnacle pH Electrodes (formerly Corning Electrodes) and Accessories
Brand:
Corning
Model Number:
476086
YO-58900-50
More Details
pH Electrode, Wastewater, Bench
Category: pH/Ion Benchtop Meters
Description: Hach sensION
™
+ pH/mV Benchtop Meters
Brand:
Hach
YO-59004-54
More Details
Cole-Parmerlog General Purpose pH electrode, double-junction, 10-ft (YO-05994-22)
Category: Cole-Parmer 12-mm Glass Body pH and ORP Electrodes and Mounting Housings
Description: Cole-Parmer 12-mm Glass Body, General Purpose, Double-Junction pH Electrodes
Brand:
Cole-Parmer
YO-05994-22
More Details
Submersible pH Electrode, CPVC (YO-27102-00)
Category: Cole-Parmer Self-Cleaning Flat Surface, Cartridge-Style pH/ORP Electrodes
Description: Self-Cleaning Industrial pH Electrodes with Submersion Assemblies
Brand:
Cole-Parmer
YO-27102-00
More Details
Submersible pH Electrode, CPVC, 100Ω RTD (YO-27102-10)
Category: Cole-Parmer Self-Cleaning Flat Surface, Cartridge-Style pH/ORP Electrodes
Description: Self-Cleaning Industrial pH Electrodes with Submersion Assemblies
Brand:
Cole-Parmer
YO-27102-10
More Details
Submersible pH Electrode, CPVC, 1 kΩ RTD (YO-27102-20)
Category: Cole-Parmer Self-Cleaning Flat Surface, Cartridge-Style pH/ORP Electrodes
Description: Self-Cleaning Industrial pH Electrodes with Submersion Assemblies
Brand:
Cole-Parmer
YO-27102-20
More Details
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC (YO-27108-00)
Category: Cole-Parmer Self-Cleaning Flat Surface, Cartridge-Style pH/ORP Electrodes
Description: Cole-Parmer Fluoride-Resistant pH Electrodes
Brand:
Cole-Parmer
YO-27108-00
More Details
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC, 100Ω RT (YO-27108-02)
Category: Cole-Parmer Self-Cleaning Flat Surface, Cartridge-Style pH/ORP Electrodes
Description: Cole-Parmer Fluoride-Resistant pH Electrodes
Brand:
Cole-Parmer
YO-27108-02
More Details
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC, 1 kΩ RT (YO-27108-04)
Category: Cole-Parmer Self-Cleaning Flat Surface, Cartridge-Style pH/ORP Electrodes
Description: Cole-Parmer Fluoride-Resistant pH Electrodes
Brand:
Cole-Parmer
YO-27108-04
More Details
pH Electrode, Gel-filled for Portable meter
Category: Hach sensION + Waterproof pH/mV Meter
Description: Replacement Electrodes
Brand:
Hach
YO-59004-51
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Replacement pH Electrode for Hach SensIon™ Meters
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Replacement pH Electrode Cartridge for 27106-series (YO-27106-60)
|
Next: Replacement pH Electrode for Horiba Twin pH Testers
31125329-p-content_2 31125329pcontent2