This item is now discontinued. We recommend the following as suggested replacements: EW-35419-40

Accumet CPAB15 AB15 Basic pH/mV benchtop meter, 110/220 VAC 59330-00

Representative image only.
Accumet Mfr # CPAB15 – Item # EW-59330-00
Large display for easy-to-see setup and operation
  • Up to five-point calibration to ensure accuracy
  • Automatic recognition of three buffer sets
  • Selectable resolution to choose faster or more precise readings
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-1.99
  • Max pH (pH)19.99
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 5 points
  • Min mV (mV)-1800
  • Max mV (mV)1800
  • mV Resolution0.1 mV
  • mV Accuracy±0.1 mV
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceEthernet, RS-232, and USB
  • Width (cm)19.05
  • Height (cm)8.255
  • Width (in)7 1/2
  • Height (in)3 1/4
  • DescriptionAB15 Basic pH/mV benchtop meter, 110/220 VAC 59330-00
  • Warranty1 year
1 Year WarrantyISO9001:2000 Certified Supplier

Key Features

  • Up to five-point calibration to ensure accuracy
  • Automatic recognition of three buffer sets
  • Selectable resolution to choose faster or more precise readings
  • 1 year warranty

More About this Item

Access all meter functions, including push-button calibration and automatic electrode check, with five front-panel buttons. The large 3"W x 4-1/4"H (7.6 x 10.8 cm) LCD steps you through setup and operation with plain language prompts.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat