Home  >  Oa to Octynol >
  

Octyl aldehyde 99% (100ml) (AC129481000)

YO-88131-08
$84.29 AUD/EACH

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 124-13-0
    Form Liquid
    Formula weight 128.21
    Empirical formula C8H16O
    Linear formula CH3(CH2)6CHO
    Synonyms Capryl aldehyde, Octanal
    Quantity (mL) 100
    Manufacturer number AC129481000
    Quantity 100ml





    Technical Resources
     
    Related Products for Octyl aldehyde 99% (100ml) (AC129481000)
    Image
      Product Detail
    SKU

    Octyl aldehyde 99% (500ml) (AC129485000)

    Category: Oa to Octynol
    Description: Octyl aldehyde
    Brand: Cole-Parmer
    Model Number: AC129485000


    Decyl aldehyde 95% (100ml) (YO-88159-09)

    Category: Da to Diallyl
    Description: Decyl aldehyde
    Brand: Cole-Parmer


    4-Nitrophenyl octyl ether 99% (5g) (AC337480050)

    Category: Nitroc to Nitroph
    Description: 4-Nitrophenyl octyl ether
    Brand: Cole-Parmer
    Model Number: AC337480050


    4-Nitrophenyl octyl ether 99% (25g) (AC337480250)

    Category: Nitroc to Nitroph
    Description: 4-Nitrophenyl octyl ether
    Brand: Cole-Parmer
    Model Number: AC337480250


    2,4-Dichlorobenzyl chloride 99% (100ml) (AC134391000)

    Category: Dichlor
    Description: 2,4-Dichlorobenzyl chloride
    Brand: Cole-Parmer
    Model Number: AC134391000


    2-Aminobenzotrifluoride 99% (100ml) (AC103181000)

    Category: Ametho to Aminob
    Description: 2-Aminobenzotrifluoride
    Brand: Cole-Parmer
    Model Number: AC103181000


    3-Aminobenzotrifluoride 99+% (100ml) (AC103191000)

    Category: Ametho to Aminob
    Description: 3-Aminobenzotrifluoride
    Brand: Cole-Parmer
    Model Number: AC103191000


    2-Heptylamine 99% (100ml) (AC103771000)

    Category: Ha to Heptyno
    Description: 2-Heptylamine
    Brand: Cole-Parmer
    Model Number: AC103771000


    2-(Aminomethyl)pyridine 99% (100ml) (AC104071000)

    Category: Aminom
    Description: 2-(Aminomethyl)pyridine
    Brand: Cole-Parmer
    Model Number: AC104071000


    3-(Aminomethyl)pyridine 99+% (100ml) (AC104081000)

    Category: Aminom
    Description: 3-(Aminomethyl)pyridine
    Brand: Cole-Parmer
    Model Number: AC104081000


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Octyl aldehyde 99% (100ml) (AC129481000)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Octyl Acetate 99+% 100g (AC310631000)  |  Next: Octyl aldehyde 99% (500ml) (AC129485000)