Home  >  Trichlo to Triethyl >
  

Triethylborane, 1M solution in tetrahydrofuran (100ml) (AC176981000)

YO-88189-87
Loading price...

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 97-94-9
    Form Liquid
    Formula weight 98
    Empirical formula C6H15B
    Linear formula (C2H5)3B
    Hazard symbol Highly_Flammable, Corrosive
    Quantity (mL) 100
    Manufacturer number AC176981000
    Quantity 100ml





    Technical Resources
     
    Related Products for Triethylborane, 1M solution in tetrahydrofuran (100ml) (AC176981000)
    Image
      Product Detail
    SKU

    HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology) (500mL)

    Category: Ha to Heptyno
    Description: HEPES Buffer, 1M Solution
    Brand: Cole-Parmer


    Borane-tetrahydrofuran complex, 1M solution in tetrahydrofuran, stabilized (800ml)

    Category: BOCL to Brillia
    Description: Borane-tetrahydrofuran complex
    Brand: Cole-Parmer


    Potassium tri-sec-butylborohydride, 1 M solution in tetrahydrofuran (100ml)

    Category: Potassium
    Description: Potassium tri-sec-butylborohydride
    Brand: Cole-Parmer


    Tributylborane, 1M solution in tetrahydrofuran (800ml) (AC177108000)

    Category: Toluica to Tricarb
    Description: Tributylborane
    Brand: Cole-Parmer


    Tris Hydrochloride, 1M Solutions(100mL)ph 7.0 100ml

    Category: Tris
    Description: Tris Hydrochloride, 1M Solutions
    Brand: Cole-Parmer


    Tris Hydrochloride, 1M Solutions(100mL)ph 7.5 100ml

    Category: Tris
    Description: Tris Hydrochloride, 1M Solutions
    Brand: Cole-Parmer


    Tris Hydrochloride, 1M Solutions(100mL)ph 8.0 100ml

    Category: Tris
    Description: Tris Hydrochloride, 1M Solutions
    Brand: Cole-Parmer


    Tris Buffer, 2M Solution (Molecular Biology) (100mL)

    Category: Tris
    Description: Tris Buffer, 2M Solution
    Brand: Cole-Parmer


    (Tris)-1,10-Phenanthroline Iron (II) Sulfate Solution (0.025M/Certified) (100mL)

    Category: Tris
    Description: (tris)-1,10-Phenanthroline Iron (II) Sulfate Solution
    Brand: Cole-Parmer


    Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride (100ml)

    Category: Trichlo to Triethyl
    Description: Triethyloxonium tetrafluoroborate
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Triethylborane, 1M solution in tetrahydrofuran (100ml) (AC176981000)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Triethylamine trihydrofluoride 97% (25ml) (AC350620250)  |  Next: Triethylene Glycol (Laboratory) (4 liter) (T3464)