2-Naphthyl sulfate, potassium salt 99% (1g) (AC230250010)

YO-88246-77
Loading price...

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 21409-32-5
    Formula weight 262.32
    Empirical formula C10H5K.H2SO4
    Linear formula C10H7OSO3K
    Manufacturer number AC230250010
    Quantity 1g





    Technical Resources
     
    Related Products for 2-Naphthyl sulfate, potassium salt 99% (1g) (AC230250010)
    Image
      Product Detail
    SKU

    5-Bromo-4-chloro-3-indolyl sulfate potassium salt 99% (50mg) (YO-88307-50)

    Category: Bromoc to Bromoe
    Description: 5-Bromo-4-chloro-3-indolyl sulfate potassium salt
    Brand: Cole-Parmer


    4-Nitrophenyl sulfate, potassium saltmonohydrate 99+% (1g) (AC227490010)

    Category: Nitroc to Nitroph
    Description: 4-Nitrophenyl sulfate, potassium saltmonohydrate
    Brand: Cole-Parmer
    Model Number: AC227490010


    Methylsulfuric acid potassium salt 99% (100g) (AC414891000)

    Category: Methylr to Methylz
    Description: Methylsulfuric acid potassium salt
    Brand: Cole-Parmer
    Model Number: AC414891000


    Gluconic acid, potassium salt 99% (1kg) (AC229320010)

    Category: Ga to Glutath
    Description: Gluconic acid, potassium salt
    Brand: Cole-Parmer
    Model Number: AC229320010


    Gluconic acid, potassium salt 99% (250g) (AC229322500)

    Category: Ga to Glutath
    Description: Gluconic acid, potassium salt
    Brand: Cole-Parmer
    Model Number: AC229322500


    Dodecyl sulfate, sodium salt 99% (100g) (AC230421000)

    Category: Dodecah to Dyspros
    Description: Dodecyl sulfate, sodium salt
    Brand: Cole-Parmer
    Model Number: AC230421000


    Dodecyl sulfate, sodium salt 99% (500g) (YO-88246-97)

    Category: Dodecah to Dyspros
    Description: Dodecyl sulfate, sodium salt
    Brand: Cole-Parmer


    (2R,3S)-1-Carboxy-4-phenyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt 95% (1g)

    Category: Carboxy to Cesium
    Description: (2R,3S)-1-Carboxy-4-phenyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
    Brand: Cole-Parmer
    Model Number: AC271630010


    Isethionic acid, potassium salt 98% (25g) (AC412630250)

    Category: Iron to Isopent
    Description: Isethionic acid, potassium salt
    Brand: Cole-Parmer
    Model Number: AC412630250


    Polyvinylsulfuric acid potassium salt 90% (25g) (AC418140250)

    Category: Platina to Potash
    Description: Polyvinylsulfuric acid potassium salt
    Brand: Cole-Parmer
    Model Number: AC418140250


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    2-Naphthyl sulfate, potassium salt 99% (1g) (AC230250010)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: 2-Naphthyl phenyl ketone 98% (5g) (AC415420050)  |  Next: 2-Naphthyl-β-D-galactopyranoside (100mg)