Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
A bromo 2 3 difluorotoluene 1g
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-88285-53
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Chemicals, Reagents, & Lubricants
Share
|
A-BROMO-2 3-DIFLUOROTOLUENE 1G (YO-88285-53)
YO-88285-53
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
A lab reagent is a staple in chemical applications. It is a great way to test chemical reactions or to see if a reaction will occur. You can find one just about anywhere including Cole-Parmer’s catalog which features a broad range of laboratory reagents, cleaning chemicals and buffers commonly found in labs. But occasionally workers run across an application that requires a specialty lab reagent. You might need Malachite Green, a high purity biological stain. How do you go about selecting a lab reagent especially if there are new and more advanced compounds out there that you might want to try? For the novice a lab reagent kit may be in order. Using kits can prevent mistakes that are related to the reagent rather than the application. If you’re pretty advanced and haven’t found that specialty compound you’re searching for try talking to a Cole-Parmer technical expert. They often have information about chemicals not listed on the website but available for special order or by other means. Cole-Parmer also offers custom blending services for specific chemical mixes. With advances in technology selecting a lab reagent may not be as routine as originally thought so it’s great to have some help.
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Product Type
Chemical Reagents
Manufacturer number
AC311540010
Technical Resources
Related technical articles (Roll over for summary)
ChemicalsSpecial Ordering Information
Show Less...
Related Searches for A-BROMO-2 3-DIFLUOROTOLUENE 1G (YO-88285-53)
Cole-Parmer 1 2-Propanediol
Cole-Parmer 2-Nitrophenol
2 Quart Stainless Steel Funnel
Cole-Parmer Methoxy
Cole-Parmer Stainless Steel Funnels
Cole-Parmer Sterile Sutures and Needles
Carbon Filters for Fume Hood
Xpert Systems
Steel Funnels
Stainless Steel Funnels in Dallas Tx
Related Products for A-BROMO-2 3-DIFLUOROTOLUENE 1G (YO-88285-53)
Image
Product Detail
SKU
1-BROMO-2 3-DIFLUOROBENZENE 1G (YO-88283-33)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88283-33
More Details
2' 6'-DIFLUOROACETOPHENONE 1G (YO-88285-45)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-45
More Details
ALPHA-BROMO-2 5-DIFLUOROTO 10G (YO-88285-54)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-54
More Details
2 5-DIFLUOROPHENYLACETONITR 1G (YO-88285-63)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-63
More Details
2 6-DIFLUOROPHENYLACETONITR 1G (YO-88285-64)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-64
More Details
2 4-BIS(TRIFLUOROMETHYL)BRO 1G (YO-88285-74)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-74
More Details
2 3-DIFLUOROBENZONITRILE 1G (YO-88279-18)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88279-18
More Details
2-AMINO-5-BROMO-3-NITROPYRI 1G (YO-88281-84)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88281-84
More Details
1-BROMO-2 3 4-TRIFLUOROBENZ 1G (YO-88283-00)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88283-00
More Details
2-CHLORO-5-TRIFLUOROMETHYL 1G (YO-88285-28)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-28
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
A-BROMO-2 3-DIFLUOROTOLUENE 1G (YO-88285-53)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: A-314:Bleed Fitting
|
Next: A&D 220V Blade Adapater
31125329-p-content_2 31125329pcontent2