Home  >  Chemicals, Reagents, & Lubricants >
  

Platinum Powder 0.27-0 500mg (AC318855000)

YO-88290-44
Loading price...

Usually ships in 14 days.
Qty
    A lab reagent is a staple in chemical applications. It is a great way to test chemical reactions or to see if a reaction will occur. You can find one just about anywhere including Cole-Parmer’s catalog which features a broad range of laboratory reagents, cleaning chemicals and buffers commonly found in labs. But occasionally workers run across an application that requires a specialty lab reagent. You might need Malachite Green, a high purity biological stain. How do you go about selecting a lab reagent especially if there are new and more advanced compounds out there that you might want to try? For the novice a lab reagent kit may be in order. Using kits can prevent mistakes that are related to the reagent rather than the application. If you’re pretty advanced and haven’t found that specialty compound you’re searching for try talking to a Cole-Parmer technical expert. They often have information about chemicals not listed on the website but available for special order or by other means. Cole-Parmer also offers custom blending services for specific chemical mixes. With advances in technology selecting a lab reagent may not be as routine as originally thought so it’s great to have some help.

    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    Form Powder
    Manufacturer number AC318855000





    Technical Resources
     
    Related Products for Platinum Powder 0.27-0 500mg (AC318855000)
    Image
      Product Detail
    SKU

    Iridium, powder 99.9% (500mg) (YO-88211-92)

    Category: Iodo to Iridium
    Description: Iridium
    Brand: Cole-Parmer


    Fluke 27 II, Rugged IP67 Industrial Multimeter

    Category: Fluke Digital Multimeters
    Description: Fluke 27 II and 28 II Rugged IP67 Industrial Multimeters
    Brand: Fluke


    4-(2-Aminoethyl)benzenesulfonyl Fluoride HCl (White Powder) (500mg)

    Category: Aminod to Aminog
    Description: 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
    Brand: Cole-Parmer


    CHAPSO (White Powder) (500mg)

    Category: Cetyl to Chloroa
    Description: CHAPSO
    Brand: Cole-Parmer


    Sodium Potassium Tartrate (Powder/USP) (500g) (YO-88081-27)

    Category: Sodiump to Sodiumz
    Description: Sodium Potassium Tartrate
    Brand: Cole-Parmer


    Three-stop silicone (platinum cured) tubing, 1.30 mm ID, 6/pack (95603-32)

    Category: Three-Stop Color-Coded Autoanalysis/Microbore Tubing for Ismatec Pumps
    Description: Three-Stop Color-Coded Silicone (Platinum Cured) Autoanalysis Pump Tubing for Ismatec Pumps
    Brand: PharMed


    Three-stop silicone (platinum cured) tubing, 1.42 mm ID, 6/pack (95603-34)

    Category: Three-Stop Color-Coded Autoanalysis/Microbore Tubing for Ismatec Pumps
    Description: Three-Stop Color-Coded Silicone (Platinum Cured) Autoanalysis Pump Tubing for Ismatec Pumps
    Brand: PharMed


    Three-stop silicone (platinum cured) tubing, 2.54 mm ID, 6/pack (95603-46)

    Category: Three-Stop Color-Coded Autoanalysis/Microbore Tubing for Ismatec Pumps
    Description: Three-Stop Color-Coded Silicone (Platinum Cured) Autoanalysis Pump Tubing for Ismatec Pumps
    Brand: PharMed


    Masterflex platinum-cured silicone sanitary tubing, L/S 13, premolded ends for 1/2" connection, 5 ft.

    Category: L/S Pump Tubing with Sanitary Connectors for Masterflex L/S Pumps
    Description: Silicone (Platinum-Cured) Sanitary Tubing for Masterflex L/S Pumps
    Brand: Masterflex


    Masterflex platinum-cured silicone sanitary tubing, L/S 14, premolded ends for 1/2" connection, 5 ft.

    Category: L/S Pump Tubing with Sanitary Connectors for Masterflex L/S Pumps
    Description: Silicone (Platinum-Cured) Sanitary Tubing for Masterflex L/S Pumps
    Brand: Masterflex


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews