Home  >  Chemicals, Reagents, & Lubricants >
  

Phenoxathiin 10g (AC417260100)

YO-88354-45
Loading price...

Usually ships in 14 days.
Qty
    A lab reagent is a staple in chemical applications. It is a great way to test chemical reactions or to see if a reaction will occur. You can find one just about anywhere including Cole-Parmer’s catalog which features a broad range of laboratory reagents, cleaning chemicals and buffers commonly found in labs. But occasionally workers run across an application that requires a specialty lab reagent. You might need Malachite Green, a high purity biological stain. How do you go about selecting a lab reagent especially if there are new and more advanced compounds out there that you might want to try? For the novice a lab reagent kit may be in order. Using kits can prevent mistakes that are related to the reagent rather than the application. If you’re pretty advanced and haven’t found that specialty compound you’re searching for try talking to a Cole-Parmer technical expert. They often have information about chemicals not listed on the website but available for special order or by other means. Cole-Parmer also offers custom blending services for specific chemical mixes. With advances in technology selecting a lab reagent may not be as routine as originally thought so it’s great to have some help.

    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    Manufacturer number AC417260100





    Technical Resources
     
    Related Products for Phenoxathiin 10g (AC417260100)
    Image
      Product Detail
    SKU

    Phenoxathiin-4-boronic acid (100mg) (AC359631000)

    Category: Pepsin to Phenoxy
    Description: Phenoxathiin-4-boronic acid
    Brand: Cole-Parmer


    PM 620 Pressure Module -14.5 to 100 psi, gauge type (PM 620 10G)

    Category: Modules
    Description: PM 620 Pressure Modules
    Brand: GE Sensing


    Cole-Parmer 65-mm Correlated Flowmeter, Aluminum with Glass float; For Liquids and Gases, With Valve (P04/6-N012-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters with Standard Valves
    Brand: Cole-Parmer


    Cole-Parmer 65-mm Correlated Flowmeter, 316 SS Glass float; For Liquids and Gases, With Valve (P24/6-N012-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters with Standard Valves
    Brand: Cole-Parmer


    Cole-Parmer 65-mm Correlated Flowmeter, Aluminum with Glass float; For Liquids and Gases, Without Valve (P03/6-N012-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters without Valves
    Brand: Cole-Parmer


    Cole-Parmer 65-mm Correlated Flowmeter, 316 SS Glass float; For Liquids and Gases, Without Valve (P23/6-N012-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters without Valves
    Brand: Cole-Parmer


    Cole-Parmer 65-mm Correlated Flowmeter, Brass Glass float; For Liquids and Gases, With Valve (P14/6-N012-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters with Standard Valves
    Brand: Cole-Parmer


    Cole-Parmer 65-mm Correlated Flowmeter, Aluminum with Glass float; For Liquids and Gases, With High-Resolution Valve (P01/6-NO12-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters with High-Resolution Valves
    Brand: Cole-Parmer


    Cole-Parmer 65-mm Correlated Flowmeter, Brass Glass float; For Liquids and Gases, With High-Resolution Valve (P11/6-N012-10G)

    Category: Cole-Parmer 65-mm Correlated Flowmeters
    Description: Correlated Flowmeters with High-Resolution Valves
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews